Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61008392-61008882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD6
Entrez Gene ID
923
Gene Name
CD6 molecule
Gene Aliases
TP120
UniGene
Hs.744366
Gene Chromosome Location
Chr.11: 60971641-61020377 on Build GRCh38
Transcripts
NM_001254750.1 NM_001254751.1 NM_006725.4 NR_045638.1 XM_006718738.1 XM_006718739.1 XM_006718740.1 XM_006718741.1 XM_011545360.1 XM_011545362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623854 c.747C>T p.C249C Chr.11: 61008811-61008811 on Build GRCh38
6025171 c.781G>T p.E261* Chr.11: 61008845-61008845 on Build GRCh38
1355170 c.693delG p.A233fs*35 Chr.11: 61008757-61008757 on Build GRCh38
429378 c.771G>C p.A257A Chr.11: 61008835-61008835 on Build GRCh38
6623858 c.483G>A p.L161L Chr.11: 61008547-61008547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762653723 Chr.11: 61008560-61008560 on Build GRCh38
rs558747964 Chr.11: 61008855-61008855 on Build GRCh38
rs377259396 Chr.11: 61008592-61008592 on Build GRCh38
rs745462325 Chr.11: 61008803-61008803 on Build GRCh38
rs770784436 Chr.11: 61008787-61008787 on Build GRCh38
rs751671555 Chr.11: 61008725-61008725 on Build GRCh38
rs866089836 Chr.11: 61008634-61008634 on Build GRCh38
rs373242647 Chr.11: 61008744-61008744 on Build GRCh38
rs368664651 Chr.11: 61008724-61008724 on Build GRCh38
rs149536568 Chr.11: 61008718-61008718 on Build GRCh38
rs758957984 Chr.11: 61008583-61008583 on Build GRCh38
rs778119453 Chr.11: 61008488-61008488 on Build GRCh38
rs764652924 Chr.11: 61008542-61008542 on Build GRCh38
rs772164608 Chr.11: 61008527-61008527 on Build GRCh38
rs962786672 Chr.11: 61008638-61008638 on Build GRCh38
rs769741012 Chr.11: 61008672-61008672 on Build GRCh38
rs1049437100 Chr.11: 61008530-61008530 on Build GRCh38
rs747177213 Chr.11: 61008652-61008652 on Build GRCh38
rs970777157 Chr.11: 61008645-61008645 on Build GRCh38
rs766880687 Chr.11: 61008712-61008712 on Build GRCh38
rs751128467 Chr.11: 61008571-61008571 on Build GRCh38
rs924109496 Chr.11: 61008477-61008477 on Build GRCh38
rs769650039 Chr.11: 61008549-61008549 on Build GRCh38
rs386754116 Chr.11: 61008834-61008835 on Build GRCh38
rs1022451660 Chr.11: 61008458-61008458 on Build GRCh38
rs1048784478 Chr.11: 61008622-61008623 on Build GRCh38
rs766098012 Chr.11: 61008568-61008568 on Build GRCh38
rs2074225 Chr.11: 61008834-61008834 on Build GRCh38
rs929780266 Chr.11: 61008859-61008859 on Build GRCh38
rs756682765 Chr.11: 61008487-61008487 on Build GRCh38
rs766624887 Chr.11: 61008723-61008723 on Build GRCh38
rs1030306849 Chr.11: 61008556-61008556 on Build GRCh38
rs747030622 Chr.11: 61008528-61008528 on Build GRCh38
rs974598178 Chr.11: 61008693-61008693 on Build GRCh38
rs747982969 Chr.11: 61008670-61008670 on Build GRCh38
rs749161653 Chr.11: 61008767-61008767 on Build GRCh38
rs759092657 Chr.11: 61008706-61008706 on Build GRCh38
rs1052716025 Chr.11: 61008743-61008743 on Build GRCh38
rs768549272 Chr.11: 61008655-61008655 on Build GRCh38
rs982246577 Chr.11: 61008443-61008443 on Build GRCh38
rs779112498 Chr.11: 61008507-61008507 on Build GRCh38
rs1040815188 Chr.11: 61008566-61008566 on Build GRCh38
rs773180903 Chr.11: 61008689-61008689 on Build GRCh38
rs866112423 Chr.11: 61008647-61008647 on Build GRCh38
rs754288553 Chr.11: 61008503-61008503 on Build GRCh38
rs761670188 Chr.11: 61008547-61008547 on Build GRCh38
rs760158878 Chr.11: 61008833-61008833 on Build GRCh38
rs757839810 Chr.11: 61008858-61008858 on Build GRCh38
rs781198543 Chr.11: 61008656-61008656 on Build GRCh38
rs757560492 Chr.11: 61008506-61008506 on Build GRCh38
rs762271208 Chr.11: 61008716-61008716 on Build GRCh38
rs774907838 Chr.11: 61008832-61008832 on Build GRCh38
rs112852289 Chr.11: 61008833-61008834 on Build GRCh38
rs750744972 Chr.11: 61008779-61008779 on Build GRCh38
rs754232003 Chr.11: 61008620-61008620 on Build GRCh38
rs746032180 Chr.11: 61008526-61008526 on Build GRCh38
rs369823651 Chr.11: 61008622-61008622 on Build GRCh38
rs776424888 Chr.11: 61008544-61008544 on Build GRCh38
rs61899223 Chr.11: 61008631-61008631 on Build GRCh38
rs781088990 Chr.11: 61008738-61008738 on Build GRCh38
rs140597355 Chr.11: 61008603-61008603 on Build GRCh38
rs773017397 Chr.11: 61008554-61008554 on Build GRCh38
rs892835573 Chr.11: 61008806-61008806 on Build GRCh38
rs780235835 Chr.11: 61008650-61008650 on Build GRCh38
rs750839213 Chr.11: 61008719-61008719 on Build GRCh38
rs750679829 Chr.11: 61008626-61008626 on Build GRCh38
rs955623862 Chr.11: 61008621-61008621 on Build GRCh38
rs777837785 Chr.11: 61008757-61008757 on Build GRCh38
rs771646536 Chr.11: 61008826-61008826 on Build GRCh38
rs758752633 Chr.11: 61008722-61008722 on Build GRCh38
rs990131561 Chr.11: 61008498-61008498 on Build GRCh38
rs79848107 Chr.11: 61008835-61008835 on Build GRCh38
rs947026108 Chr.11: 61008529-61008529 on Build GRCh38
rs768691551 Chr.11: 61008537-61008537 on Build GRCh38
rs779203131 Chr.11: 61008623-61008623 on Build GRCh38
rs773872697 Chr.11: 61008698-61008698 on Build GRCh38
rs11230563 Chr.11: 61008737-61008737 on Build GRCh38
rs72926536 Chr.11: 61008861-61008861 on Build GRCh38
rs1003638548 Chr.11: 61008651-61008651 on Build GRCh38
rs148197209 Chr.11: 61008829-61008829 on Build GRCh38
rs2905518 Chr.11: 61008862-61008862 on Build GRCh38
rs981309422 Chr.11: 61008717-61008717 on Build GRCh38
rs756168861 Chr.11: 61008756-61008756 on Build GRCh38
rs11230562 Chr.11: 61008714-61008714 on Build GRCh38
rs556607637 Chr.11: 61008790-61008790 on Build GRCh38

View Full Product Details