Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61299751-61300244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB1
Entrez Gene ID
1642
Gene Name
damage specific DNA binding protein 1
Gene Aliases
DDBA, UV-DDB1, XAP1, XPCE, XPE, XPE-BF
UniGene
Hs.290758
Gene Chromosome Location
Chr.11: 61299447-61333212 on Build GRCh38
Transcripts
NM_001923.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7264451 c.3345C>T p.D1115D Chr.11: 61300214-61300214 on Build GRCh38
2037288 c.3365G>A p.R1122Q Chr.11: 61300194-61300194 on Build GRCh38
4676654 c.3412C>T p.R1138W Chr.11: 61300147-61300147 on Build GRCh38
689333 c.3413G>A p.R1138Q Chr.11: 61300146-61300146 on Build GRCh38
1298280 c.3343G>A p.D1115N Chr.11: 61300216-61300216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030025388 Chr.11: 61299798-61299798 on Build GRCh38
rs774190562 Chr.11: 61300111-61300111 on Build GRCh38
rs763102052 Chr.11: 61299783-61299783 on Build GRCh38
rs999566980 Chr.11: 61300061-61300061 on Build GRCh38
rs750936948 Chr.11: 61300167-61300167 on Build GRCh38
rs375160067 Chr.11: 61300166-61300166 on Build GRCh38
rs534974673 Chr.11: 61300047-61300047 on Build GRCh38
rs547082121 Chr.11: 61299885-61299885 on Build GRCh38
rs538485460 Chr.11: 61299945-61299945 on Build GRCh38
rs963243863 Chr.11: 61299838-61299838 on Build GRCh38
rs759244080 Chr.11: 61300114-61300114 on Build GRCh38
rs918541537 Chr.11: 61300106-61300106 on Build GRCh38
rs375900124 Chr.11: 61299810-61299811 on Build GRCh38
rs771804044 Chr.11: 61300122-61300122 on Build GRCh38
rs554677936 Chr.11: 61300060-61300060 on Build GRCh38
rs886143445 Chr.11: 61300085-61300085 on Build GRCh38
rs748216785 Chr.11: 61300090-61300090 on Build GRCh38
rs80012527 Chr.11: 61300118-61300118 on Build GRCh38
rs28720363 Chr.11: 61300065-61300065 on Build GRCh38
rs909543791 Chr.11: 61300153-61300153 on Build GRCh38
rs1016423846 Chr.11: 61299836-61299836 on Build GRCh38
rs763266907 Chr.11: 61300169-61300169 on Build GRCh38
rs753797966 Chr.11: 61300107-61300107 on Build GRCh38
rs796799077 Chr.11: 61299781-61299782 on Build GRCh38
rs934259840 Chr.11: 61300073-61300073 on Build GRCh38
rs373167984 Chr.11: 61300135-61300135 on Build GRCh38
rs925997619 Chr.11: 61300040-61300040 on Build GRCh38
rs751815287 Chr.11: 61299798-61299799 on Build GRCh38
rs3209377 Chr.11: 61299931-61299931 on Build GRCh38
rs28720364 Chr.11: 61299796-61299796 on Build GRCh38
rs561196813 Chr.11: 61299908-61299908 on Build GRCh38
rs946166022 Chr.11: 61299780-61299780 on Build GRCh38
rs138674997 Chr.11: 61300214-61300214 on Build GRCh38
rs751870731 Chr.11: 61300187-61300187 on Build GRCh38
rs757179069 Chr.11: 61299783-61299784 on Build GRCh38
rs886091221 Chr.11: 61299825-61299825 on Build GRCh38
rs1004667095 Chr.11: 61299827-61299827 on Build GRCh38
rs373562495 Chr.11: 61300103-61300103 on Build GRCh38
rs929669689 Chr.11: 61299852-61299852 on Build GRCh38
rs937001154 Chr.11: 61300174-61300174 on Build GRCh38
rs775172388 Chr.11: 61300123-61300123 on Build GRCh38
rs946048188 Chr.11: 61299985-61299985 on Build GRCh38
rs754870308 Chr.11: 61300120-61300120 on Build GRCh38
rs146394965 Chr.11: 61299797-61299798 on Build GRCh38
rs868847308 Chr.11: 61300165-61300165 on Build GRCh38
rs550076638 Chr.11: 61299918-61299918 on Build GRCh38
rs767656071 Chr.11: 61300218-61300218 on Build GRCh38
rs959859422 Chr.11: 61300213-61300213 on Build GRCh38
rs537325146 Chr.11: 61300083-61300083 on Build GRCh38
rs545112273 Chr.11: 61299816-61299816 on Build GRCh38
rs552249276 Chr.11: 61299952-61299952 on Build GRCh38
rs985966641 Chr.11: 61300072-61300072 on Build GRCh38
rs1025261125 Chr.11: 61300051-61300051 on Build GRCh38
rs911280777 Chr.11: 61300079-61300079 on Build GRCh38
rs1055508223 Chr.11: 61299984-61299984 on Build GRCh38
rs769620515 Chr.11: 61300098-61300098 on Build GRCh38
rs770759265 Chr.11: 61300109-61300109 on Build GRCh38
rs190309211 Chr.11: 61299853-61299853 on Build GRCh38
rs527281127 Chr.11: 61299861-61299861 on Build GRCh38
rs529961119 Chr.11: 61299915-61299915 on Build GRCh38
rs1016021115 Chr.11: 61300105-61300105 on Build GRCh38
rs1132797 Chr.11: 61299782-61299782 on Build GRCh38
rs777704235 Chr.11: 61300099-61300099 on Build GRCh38
rs572476486 Chr.11: 61299777-61299790 on Build GRCh38
rs1040251951 Chr.11: 61300050-61300050 on Build GRCh38
rs1013727449 Chr.11: 61300048-61300048 on Build GRCh38
rs61895891 Chr.11: 61300195-61300195 on Build GRCh38
rs766707297 Chr.11: 61300178-61300178 on Build GRCh38
rs1034627868 Chr.11: 61300183-61300183 on Build GRCh38
rs992694051 Chr.11: 61299937-61299937 on Build GRCh38
rs918491081 Chr.11: 61299815-61299815 on Build GRCh38
rs1032336438 Chr.11: 61300064-61300064 on Build GRCh38
rs565836116 Chr.11: 61299991-61299991 on Build GRCh38

View Full Product Details