Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61310205-61310708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB1
Entrez Gene ID
1642
Gene Name
damage specific DNA binding protein 1
Gene Aliases
DDBA, UV-DDB1, XAP1, XPCE, XPE, XPE-BF
UniGene
Hs.290758
Gene Chromosome Location
Chr.11: 61299447-61333212 on Build GRCh38
Transcripts
NM_001923.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416591 c.2360A>G p.E787G Chr.11: 61310336-61310336 on Build GRCh38
232140 c.2342C>T p.S781F Chr.11: 61310354-61310354 on Build GRCh38
7363912 c.2350G>A p.E784K Chr.11: 61310346-61310346 on Build GRCh38
6345275 c.2285C>A p.S762Y Chr.11: 61310411-61310411 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs890375332 Chr.11: 61310308-61310308 on Build GRCh38
rs529564907 Chr.11: 61310618-61310618 on Build GRCh38
rs749327518 Chr.11: 61310435-61310435 on Build GRCh38
rs867898499 Chr.11: 61310415-61310415 on Build GRCh38
rs564289747 Chr.11: 61310458-61310460 on Build GRCh38
rs926030044 Chr.11: 61310542-61310542 on Build GRCh38
rs890270041 Chr.11: 61310673-61310673 on Build GRCh38
rs763537975 Chr.11: 61310369-61310369 on Build GRCh38
rs781613654 Chr.11: 61310424-61310424 on Build GRCh38
rs1038481283 Chr.11: 61310683-61310683 on Build GRCh38
rs923453709 Chr.11: 61310552-61310552 on Build GRCh38
rs927452457 Chr.11: 61310472-61310472 on Build GRCh38
rs776981352 Chr.11: 61310278-61310278 on Build GRCh38
rs866009428 Chr.11: 61310254-61310254 on Build GRCh38
rs555624580 Chr.11: 61310359-61310359 on Build GRCh38
rs769737920 Chr.11: 61310288-61310288 on Build GRCh38
rs80208062 Chr.11: 61310533-61310533 on Build GRCh38
rs759310673 Chr.11: 61310328-61310328 on Build GRCh38
rs542940198 Chr.11: 61310670-61310670 on Build GRCh38
rs753352620 Chr.11: 61310371-61310371 on Build GRCh38
rs1053908 Chr.11: 61310329-61310329 on Build GRCh38
rs1002425282 Chr.11: 61310582-61310582 on Build GRCh38
rs759660492 Chr.11: 61310332-61310332 on Build GRCh38
rs758893429 Chr.11: 61310264-61310264 on Build GRCh38
rs28720326 Chr.11: 61310416-61310416 on Build GRCh38
rs373866581 Chr.11: 61310319-61310319 on Build GRCh38
rs369074861 Chr.11: 61310277-61310277 on Build GRCh38
rs552487833 Chr.11: 61310503-61310503 on Build GRCh38
rs369683404 Chr.11: 61310407-61310407 on Build GRCh38
rs41553012 Chr.11: 61310323-61310323 on Build GRCh38
rs11556192 Chr.11: 61310380-61310380 on Build GRCh38
rs527507882 Chr.11: 61310325-61310325 on Build GRCh38
rs752780298 Chr.11: 61310430-61310430 on Build GRCh38
rs560335578 Chr.11: 61310521-61310521 on Build GRCh38
rs780314376 Chr.11: 61310269-61310269 on Build GRCh38
rs758833977 Chr.11: 61310395-61310395 on Build GRCh38
rs913250111 Chr.11: 61310491-61310491 on Build GRCh38
rs770828280 Chr.11: 61310441-61310441 on Build GRCh38
rs932048589 Chr.11: 61310546-61310546 on Build GRCh38
rs755673027 Chr.11: 61310364-61310364 on Build GRCh38
rs748275911 Chr.11: 61310284-61310284 on Build GRCh38
rs1053100 Chr.11: 61310334-61310334 on Build GRCh38
rs553186063 Chr.11: 61310238-61310238 on Build GRCh38
rs1047865129 Chr.11: 61310568-61310568 on Build GRCh38
rs575540605 Chr.11: 61310401-61310401 on Build GRCh38
rs746341823 Chr.11: 61310262-61310262 on Build GRCh38
rs28720325 Chr.11: 61310576-61310576 on Build GRCh38
rs141156606 Chr.11: 61310483-61310483 on Build GRCh38
rs767220802 Chr.11: 61310353-61310353 on Build GRCh38
rs28720327 Chr.11: 61310261-61310261 on Build GRCh38
rs960572326 Chr.11: 61310478-61310478 on Build GRCh38
rs969575466 Chr.11: 61310276-61310276 on Build GRCh38
rs573926742 Chr.11: 61310561-61310562 on Build GRCh38
rs1006463077 Chr.11: 61310617-61310617 on Build GRCh38
rs755619744 Chr.11: 61310260-61310260 on Build GRCh38
rs376921474 Chr.11: 61310428-61310428 on Build GRCh38
rs773293195 Chr.11: 61310318-61310318 on Build GRCh38
rs904387236 Chr.11: 61310241-61310241 on Build GRCh38

View Full Product Details