Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61311542-61312037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB1
Entrez Gene ID
1642
Gene Name
damage specific DNA binding protein 1
Gene Aliases
DDBA, UV-DDB1, XAP1, XPCE, XPE, XPE-BF
UniGene
Hs.290758
Gene Chromosome Location
Chr.11: 61299447-61333212 on Build GRCh38
Transcripts
NM_001923.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1661403 c.2256G>C p.L752F Chr.11: 61311805-61311805 on Build GRCh38
4034579 c.2246C>T p.T749M Chr.11: 61311815-61311815 on Build GRCh38
4629778 c.2206C>A p.L736I Chr.11: 61311855-61311855 on Build GRCh38
1322314 c.2277_2277+1GG>TT p.? Chr.11: 61311783-61311784 on Build GRCh38
77568 c.2162C>T p.P721L Chr.11: 61311992-61311992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764543068 Chr.11: 61312006-61312006 on Build GRCh38
rs760461879 Chr.11: 61311837-61311837 on Build GRCh38
rs757475414 Chr.11: 61312015-61312015 on Build GRCh38
rs28720315 Chr.11: 61311926-61311926 on Build GRCh38
rs754071831 Chr.11: 61311917-61311917 on Build GRCh38
rs149076473 Chr.11: 61311790-61311790 on Build GRCh38
rs753057563 Chr.11: 61311901-61311901 on Build GRCh38
rs760992062 Chr.11: 61312003-61312003 on Build GRCh38
rs993612613 Chr.11: 61311695-61311695 on Build GRCh38
rs755475840 Chr.11: 61311760-61311763 on Build GRCh38
rs780029105 Chr.11: 61311875-61311875 on Build GRCh38
rs28720316 Chr.11: 61311796-61311796 on Build GRCh38
rs775219838 Chr.11: 61311764-61311764 on Build GRCh38
rs776344314 Chr.11: 61311843-61311843 on Build GRCh38
rs761373922 Chr.11: 61311781-61311781 on Build GRCh38
rs775273250 Chr.11: 61311830-61311830 on Build GRCh38
rs966007105 Chr.11: 61311666-61311666 on Build GRCh38
rs11230667 Chr.11: 61311703-61311703 on Build GRCh38
rs776237837 Chr.11: 61311974-61311974 on Build GRCh38
rs772861659 Chr.11: 61311982-61311982 on Build GRCh38
rs749911180 Chr.11: 61311856-61311856 on Build GRCh38
rs11601052 Chr.11: 61311946-61311946 on Build GRCh38
rs755492633 Chr.11: 61311793-61311793 on Build GRCh38
rs749392198 Chr.11: 61311814-61311814 on Build GRCh38
rs1008186666 Chr.11: 61311612-61311612 on Build GRCh38
rs754113357 Chr.11: 61312012-61312012 on Build GRCh38
rs925026585 Chr.11: 61311955-61311955 on Build GRCh38
rs58597752 Chr.11: 61311912-61311912 on Build GRCh38
rs556400194 Chr.11: 61311718-61311718 on Build GRCh38
rs936435343 Chr.11: 61311846-61311846 on Build GRCh38
rs190054116 Chr.11: 61311845-61311845 on Build GRCh38
rs771008830 Chr.11: 61311819-61311819 on Build GRCh38
rs774265530 Chr.11: 61311746-61311746 on Build GRCh38
rs778040253 Chr.11: 61311802-61311802 on Build GRCh38
rs1018966232 Chr.11: 61311650-61311650 on Build GRCh38
rs753044755 Chr.11: 61311799-61311799 on Build GRCh38
rs763865068 Chr.11: 61312001-61312001 on Build GRCh38
rs28720314 Chr.11: 61311943-61311943 on Build GRCh38
rs372313326 Chr.11: 61311826-61311826 on Build GRCh38
rs771961884 Chr.11: 61311827-61311827 on Build GRCh38
rs765750573 Chr.11: 61311762-61311763 on Build GRCh38
rs764261182 Chr.11: 61311913-61311913 on Build GRCh38
rs936391477 Chr.11: 61311965-61311965 on Build GRCh38
rs779954982 Chr.11: 61311942-61311942 on Build GRCh38
rs951901226 Chr.11: 61311760-61311760 on Build GRCh38
rs745885740 Chr.11: 61311936-61311936 on Build GRCh38
rs750352229 Chr.11: 61311625-61311625 on Build GRCh38
rs147201675 Chr.11: 61311761-61311761 on Build GRCh38
rs992454973 Chr.11: 61311876-61311876 on Build GRCh38
rs368339659 Chr.11: 61311751-61311751 on Build GRCh38
rs34519360 Chr.11: 61311791-61311791 on Build GRCh38
rs762537993 Chr.11: 61311985-61311985 on Build GRCh38
rs560290613 Chr.11: 61311923-61311923 on Build GRCh38
rs756300337 Chr.11: 61311801-61311801 on Build GRCh38
rs960165684 Chr.11: 61311684-61311684 on Build GRCh38
rs760125089 Chr.11: 61311862-61311862 on Build GRCh38
rs868063554 Chr.11: 61311855-61311855 on Build GRCh38
rs760280947 Chr.11: 61311768-61311768 on Build GRCh38
rs1004865656 Chr.11: 61311568-61311568 on Build GRCh38
rs768468919 Chr.11: 61311963-61311963 on Build GRCh38
rs778873432 Chr.11: 61311823-61311823 on Build GRCh38
rs766864507 Chr.11: 61311787-61311787 on Build GRCh38
rs759368261 Chr.11: 61311753-61311753 on Build GRCh38
rs758830038 Chr.11: 61311944-61311944 on Build GRCh38
rs753169593 Chr.11: 61311817-61311818 on Build GRCh38
rs993097394 Chr.11: 61311685-61311685 on Build GRCh38
rs758346327 Chr.11: 61311940-61311940 on Build GRCh38
rs768204727 Chr.11: 61311993-61311993 on Build GRCh38
rs747864801 Chr.11: 61311976-61311976 on Build GRCh38
rs545558847 Chr.11: 61311762-61311762 on Build GRCh38
rs889261586 Chr.11: 61312011-61312011 on Build GRCh38
rs553889793 Chr.11: 61311570-61311570 on Build GRCh38
rs757375979 Chr.11: 61311918-61311918 on Build GRCh38
rs573696112 Chr.11: 61311672-61311672 on Build GRCh38
rs746848371 Chr.11: 61311945-61311945 on Build GRCh38
rs753302742 Chr.11: 61311778-61311778 on Build GRCh38
rs768252620 Chr.11: 61311842-61311842 on Build GRCh38
rs963529124 Chr.11: 61311583-61311583 on Build GRCh38
rs769504067 Chr.11: 61311979-61311979 on Build GRCh38
rs917618830 Chr.11: 61311891-61311891 on Build GRCh38
rs764861108 Chr.11: 61311850-61311850 on Build GRCh38
rs763735628 Chr.11: 61311773-61311773 on Build GRCh38

View Full Product Details