Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61349234-61349722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB561A3
Entrez Gene ID
220002
Gene Name
cytochrome b561 family member A3
Gene Aliases
CYBASC3, LCYTB
UniGene
Hs.22546
Gene Chromosome Location
Chr.11: 61348748-61362299 on Build GRCh38
Transcripts
NM_001161452.1 NM_001161454.1 NM_001300763.1 NM_153611.4 XM_011544821.1 XM_011544823.1 XM_017017345.1 XM_017017346.1 XM_017017347.1 XM_017017518.1 XM_017017522.1

Gene Symbol
TKFC
Entrez Gene ID
26007
Gene Name
triokinase and FMN cyclase
Gene Aliases
DAK, NET45
UniGene
Hs.6278
Gene Chromosome Location
Chr.11: 61333182-61353426 on Build GRCh38
Transcripts
NM_001161452.1 NM_001161454.1 NM_001300763.1 NM_153611.4 XM_011544821.1 XM_011544823.1 XM_017017345.1 XM_017017346.1 XM_017017347.1 XM_017017518.1 XM_017017522.1

SNP Detail

SNP ID Chromosome Location
rs535785719 Chr.11: 61349661-61349661 on Build GRCh38
rs747320182 Chr.11: 61349584-61349584 on Build GRCh38
rs972776949 Chr.11: 61349537-61349537 on Build GRCh38
rs996944938 Chr.11: 61349512-61349512 on Build GRCh38
rs773226751 Chr.11: 61349618-61349618 on Build GRCh38
rs780247040 Chr.11: 61349548-61349548 on Build GRCh38
rs770223455 Chr.11: 61349357-61349357 on Build GRCh38
rs531580893 Chr.11: 61349440-61349440 on Build GRCh38
rs964666399 Chr.11: 61349470-61349470 on Build GRCh38
rs948563178 Chr.11: 61349278-61349278 on Build GRCh38
rs908659080 Chr.11: 61349577-61349577 on Build GRCh38
rs892785769 Chr.11: 61349638-61349638 on Build GRCh38
rs961560585 Chr.11: 61349528-61349528 on Build GRCh38
rs774014018 Chr.11: 61349665-61349665 on Build GRCh38
rs371157549 Chr.11: 61349615-61349615 on Build GRCh38
rs550723512 Chr.11: 61349429-61349429 on Build GRCh38
rs1011695426 Chr.11: 61349442-61349442 on Build GRCh38
rs982832180 Chr.11: 61349538-61349538 on Build GRCh38
rs1045610157 Chr.11: 61349282-61349282 on Build GRCh38
rs7949479 Chr.11: 61349308-61349308 on Build GRCh38
rs1022530228 Chr.11: 61349443-61349443 on Build GRCh38
rs539405227 Chr.11: 61349591-61349591 on Build GRCh38
rs1052797121 Chr.11: 61349634-61349634 on Build GRCh38
rs953856775 Chr.11: 61349511-61349511 on Build GRCh38
rs190949677 Chr.11: 61349595-61349595 on Build GRCh38
rs768848652 Chr.11: 61349613-61349613 on Build GRCh38
rs548393372 Chr.11: 61349596-61349596 on Build GRCh38
rs367724244 Chr.11: 61349346-61349346 on Build GRCh38
rs1011364907 Chr.11: 61349646-61349646 on Build GRCh38
rs1019530377 Chr.11: 61349516-61349516 on Build GRCh38
rs892892248 Chr.11: 61349416-61349416 on Build GRCh38
rs570583486 Chr.11: 61349532-61349532 on Build GRCh38
rs994455889 Chr.11: 61349694-61349694 on Build GRCh38
rs1008076337 Chr.11: 61349664-61349664 on Build GRCh38
rs746308761 Chr.11: 61349317-61349317 on Build GRCh38
rs1029796775 Chr.11: 61349530-61349530 on Build GRCh38
rs764286283 Chr.11: 61349660-61349660 on Build GRCh38
rs769830027 Chr.11: 61349616-61349616 on Build GRCh38
rs774479891 Chr.11: 61349676-61349676 on Build GRCh38
rs187383096 Chr.11: 61349321-61349321 on Build GRCh38

View Full Product Details