Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61356425-61356922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB561A3
Entrez Gene ID
220002
Gene Name
cytochrome b561 family member A3
Gene Aliases
CYBASC3, LCYTB
UniGene
Hs.22546
Gene Chromosome Location
Chr.11: 61348748-61362299 on Build GRCh38
Transcripts
NM_001161452.1 NM_001161454.1 NM_001300763.1 NM_153611.4 XM_011544821.1 XM_011544823.1 XM_011544824.1 XM_017017345.1 XM_017017346.1 XM_017017347.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1585865 c.145C>T p.R49C Chr.11: 61356620-61356620 on Build GRCh38
3809744 c.78C>T p.S26S Chr.11: 61356687-61356687 on Build GRCh38
7158036 c.73A>G p.I25V Chr.11: 61356641-61356641 on Build GRCh38
4135690 c.55A>T p.M19L Chr.11: 61356659-61356659 on Build GRCh38
5429144 c.65G>A p.R22Q Chr.11: 61356700-61356700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780362419 Chr.11: 61356620-61356620 on Build GRCh38
rs535593091 Chr.11: 61356617-61356617 on Build GRCh38
rs774864809 Chr.11: 61356694-61356694 on Build GRCh38
rs367786985 Chr.11: 61356719-61356719 on Build GRCh38
rs972856023 Chr.11: 61356894-61356894 on Build GRCh38
rs150562601 Chr.11: 61356701-61356701 on Build GRCh38
rs762142933 Chr.11: 61356737-61356739 on Build GRCh38
rs912827998 Chr.11: 61356488-61356488 on Build GRCh38
rs200238994 Chr.11: 61356495-61356495 on Build GRCh38
rs757249515 Chr.11: 61356772-61356772 on Build GRCh38
rs528972385 Chr.11: 61356503-61356503 on Build GRCh38
rs567135754 Chr.11: 61356524-61356527 on Build GRCh38
rs767204912 Chr.11: 61356538-61356538 on Build GRCh38
rs762692032 Chr.11: 61356505-61356505 on Build GRCh38
rs566667066 Chr.11: 61356607-61356607 on Build GRCh38
rs749254249 Chr.11: 61356771-61356771 on Build GRCh38
rs929861010 Chr.11: 61356868-61356868 on Build GRCh38
rs201907044 Chr.11: 61356678-61356678 on Build GRCh38
rs752606159 Chr.11: 61356753-61356755 on Build GRCh38
rs2290388 Chr.11: 61356831-61356831 on Build GRCh38
rs371064566 Chr.11: 61356526-61356529 on Build GRCh38
rs1019834723 Chr.11: 61356883-61356883 on Build GRCh38
rs773629309 Chr.11: 61356688-61356688 on Build GRCh38
rs530605885 Chr.11: 61356875-61356875 on Build GRCh38
rs756183303 Chr.11: 61356644-61356644 on Build GRCh38
rs1045168604 Chr.11: 61356879-61356879 on Build GRCh38
rs577994166 Chr.11: 61356829-61356829 on Build GRCh38
rs374372315 Chr.11: 61356516-61356516 on Build GRCh38
rs927310097 Chr.11: 61356619-61356619 on Build GRCh38
rs919973325 Chr.11: 61356549-61356549 on Build GRCh38
rs777882043 Chr.11: 61356741-61356741 on Build GRCh38
rs748185091 Chr.11: 61356642-61356642 on Build GRCh38
rs199598320 Chr.11: 61356656-61356656 on Build GRCh38
rs766652171 Chr.11: 61356491-61356491 on Build GRCh38
rs560804273 Chr.11: 61356843-61356843 on Build GRCh38
rs371810863 Chr.11: 61356687-61356687 on Build GRCh38
rs777597647 Chr.11: 61356651-61356651 on Build GRCh38
rs774196828 Chr.11: 61356519-61356519 on Build GRCh38
rs752160353 Chr.11: 61356546-61356546 on Build GRCh38
rs759621508 Chr.11: 61356562-61356563 on Build GRCh38
rs755177003 Chr.11: 61356624-61356624 on Build GRCh38
rs961630228 Chr.11: 61356888-61356888 on Build GRCh38
rs371792294 Chr.11: 61356479-61356479 on Build GRCh38
rs368860318 Chr.11: 61356504-61356504 on Build GRCh38
rs770800771 Chr.11: 61356511-61356511 on Build GRCh38
rs904243408 Chr.11: 61356813-61356813 on Build GRCh38
rs201818305 Chr.11: 61356499-61356499 on Build GRCh38
rs749227299 Chr.11: 61356655-61356655 on Build GRCh38
rs765693705 Chr.11: 61356588-61356588 on Build GRCh38
rs373218798 Chr.11: 61356566-61356566 on Build GRCh38
rs370931902 Chr.11: 61356722-61356722 on Build GRCh38
rs763442436 Chr.11: 61356696-61356696 on Build GRCh38
rs543197868 Chr.11: 61356863-61356863 on Build GRCh38
rs372026923 Chr.11: 61356725-61356725 on Build GRCh38
rs368201525 Chr.11: 61356724-61356724 on Build GRCh38
rs534210430 Chr.11: 61356652-61356653 on Build GRCh38
rs200823476 Chr.11: 61356602-61356602 on Build GRCh38
rs374580223 Chr.11: 61356723-61356723 on Build GRCh38
rs760125644 Chr.11: 61356684-61356684 on Build GRCh38
rs565633437 Chr.11: 61356641-61356641 on Build GRCh38
rs774285181 Chr.11: 61356661-61356661 on Build GRCh38
rs1453319 Chr.11: 61356493-61356493 on Build GRCh38
rs954382350 Chr.11: 61356872-61356872 on Build GRCh38
rs891733209 Chr.11: 61356700-61356700 on Build GRCh38
rs12291349 Chr.11: 61356874-61356874 on Build GRCh38
rs115940701 Chr.11: 61356537-61356537 on Build GRCh38
rs547657560 Chr.11: 61356857-61356857 on Build GRCh38
rs575330098 Chr.11: 61356670-61356670 on Build GRCh38
rs781336830 Chr.11: 61356630-61356630 on Build GRCh38
rs931387193 Chr.11: 61356592-61356592 on Build GRCh38
rs751937853 Chr.11: 61356704-61356704 on Build GRCh38

View Full Product Details