Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61799538-61800041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS1
Entrez Gene ID
3992
Gene Name
fatty acid desaturase 1
Gene Aliases
D5D, FADS6, FADSD5, LLCDL1, TU12
UniGene
Hs.503546
Gene Chromosome Location
Chr.11: 61799625-61817057 on Build GRCh38
Transcripts
NM_013402.4

SNP Detail

SNP ID Chromosome Location
rs946490572 Chr.11: 61799743-61799744 on Build GRCh38
rs943009601 Chr.11: 61799817-61799817 on Build GRCh38
rs188952822 Chr.11: 61799668-61799668 on Build GRCh38
rs14536 Chr.11: 61799662-61799662 on Build GRCh38
rs981814129 Chr.11: 61799616-61799616 on Build GRCh38
rs150861621 Chr.11: 61799686-61799686 on Build GRCh38
rs1035389713 Chr.11: 61799584-61799584 on Build GRCh38
rs918736518 Chr.11: 61799681-61799681 on Build GRCh38
rs778668667 Chr.11: 61799644-61799645 on Build GRCh38
rs1051344734 Chr.11: 61799769-61799769 on Build GRCh38
rs943065153 Chr.11: 61799813-61799813 on Build GRCh38
rs1003821603 Chr.11: 61799991-61799991 on Build GRCh38
rs1038735877 Chr.11: 61799853-61799853 on Build GRCh38
rs750801284 Chr.11: 61799648-61799648 on Build GRCh38
rs879339918 Chr.11: 61799642-61799642 on Build GRCh38
rs893529336 Chr.11: 61799863-61799863 on Build GRCh38
rs991236998 Chr.11: 61799598-61799598 on Build GRCh38
rs757960888 Chr.11: 61799794-61799794 on Build GRCh38
rs568437228 Chr.11: 61799866-61799866 on Build GRCh38
rs529024824 Chr.11: 61799563-61799563 on Build GRCh38
rs993807067 Chr.11: 61799860-61799860 on Build GRCh38
rs914736100 Chr.11: 61799727-61799727 on Build GRCh38
rs374878388 Chr.11: 61799594-61799594 on Build GRCh38
rs118171418 Chr.11: 61799880-61799880 on Build GRCh38
rs966565305 Chr.11: 61799562-61799562 on Build GRCh38
rs376524567 Chr.11: 61799823-61799823 on Build GRCh38
rs902347322 Chr.11: 61799985-61799985 on Build GRCh38
rs531599872 Chr.11: 61799861-61799861 on Build GRCh38
rs547998922 Chr.11: 61800015-61800015 on Build GRCh38
rs909932098 Chr.11: 61799604-61799604 on Build GRCh38
rs571947776 Chr.11: 61799905-61799905 on Build GRCh38
rs527964264 Chr.11: 61799960-61799960 on Build GRCh38
rs1020743263 Chr.11: 61799964-61799964 on Build GRCh38
rs887608651 Chr.11: 61799910-61799910 on Build GRCh38
rs756523214 Chr.11: 61799787-61799787 on Build GRCh38
rs747702965 Chr.11: 61799738-61799738 on Build GRCh38
rs369160971 Chr.11: 61799938-61799938 on Build GRCh38

View Full Product Details