Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61800139-61800685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS1
Entrez Gene ID
3992
Gene Name
fatty acid desaturase 1
Gene Aliases
D5D, FADS6, FADSD5, LLCDL1, TU12
UniGene
Hs.503546
Gene Chromosome Location
Chr.11: 61799625-61817057 on Build GRCh38
Transcripts
NM_013402.4 XM_011545022.2

SNP Detail

SNP ID Chromosome Location
rs992206359 Chr.11: 61800227-61800227 on Build GRCh38
rs758102662 Chr.11: 61800383-61800383 on Build GRCh38
rs1012106797 Chr.11: 61800268-61800268 on Build GRCh38
rs139299536 Chr.11: 61800260-61800260 on Build GRCh38
rs508187 Chr.11: 61800210-61800210 on Build GRCh38
rs527762120 Chr.11: 61800598-61800598 on Build GRCh38
rs1056720719 Chr.11: 61800261-61800261 on Build GRCh38
rs149991368 Chr.11: 61800623-61800623 on Build GRCh38
rs1035733740 Chr.11: 61800475-61800475 on Build GRCh38
rs1054739314 Chr.11: 61800197-61800197 on Build GRCh38
rs185398735 Chr.11: 61800229-61800229 on Build GRCh38
rs911452235 Chr.11: 61800170-61800170 on Build GRCh38
rs552883766 Chr.11: 61800520-61800520 on Build GRCh38
rs372120634 Chr.11: 61800192-61800192 on Build GRCh38
rs553613646 Chr.11: 61800267-61800267 on Build GRCh38
rs909150354 Chr.11: 61800640-61800640 on Build GRCh38
rs746960326 Chr.11: 61800219-61800219 on Build GRCh38
rs536808877 Chr.11: 61800182-61800182 on Build GRCh38
rs964524706 Chr.11: 61800522-61800522 on Build GRCh38
rs986956417 Chr.11: 61800477-61800477 on Build GRCh38
rs990400430 Chr.11: 61800495-61800495 on Build GRCh38
rs1025642981 Chr.11: 61800354-61800354 on Build GRCh38
rs369065043 Chr.11: 61800532-61800532 on Build GRCh38
rs905977186 Chr.11: 61800389-61800389 on Build GRCh38
rs946332151 Chr.11: 61800599-61800599 on Build GRCh38
rs1003362289 Chr.11: 61800422-61800422 on Build GRCh38
rs746804609 Chr.11: 61800592-61800592 on Build GRCh38
rs1047639759 Chr.11: 61800269-61800269 on Build GRCh38
rs573506611 Chr.11: 61800277-61800277 on Build GRCh38
rs174544 Chr.11: 61800281-61800281 on Build GRCh38
rs200711264 Chr.11: 61800197-61800198 on Build GRCh38
rs942956270 Chr.11: 61800196-61800196 on Build GRCh38
rs968936752 Chr.11: 61800494-61800494 on Build GRCh38
rs545419144 Chr.11: 61800533-61800533 on Build GRCh38
rs535851459 Chr.11: 61800319-61800319 on Build GRCh38
rs1127959 Chr.11: 61800254-61800254 on Build GRCh38
rs911364741 Chr.11: 61800478-61800478 on Build GRCh38
rs917933392 Chr.11: 61800246-61800246 on Build GRCh38
rs181067085 Chr.11: 61800483-61800483 on Build GRCh38
rs8448 Chr.11: 61800465-61800465 on Build GRCh38
rs994136126 Chr.11: 61800324-61800324 on Build GRCh38
rs193250731 Chr.11: 61800198-61800198 on Build GRCh38
rs1022729029 Chr.11: 61800351-61800351 on Build GRCh38
rs562688177 Chr.11: 61800480-61800480 on Build GRCh38
rs190592521 Chr.11: 61800249-61800249 on Build GRCh38
rs568928148 Chr.11: 61800199-61800199 on Build GRCh38
rs946407174 Chr.11: 61800239-61800239 on Build GRCh38
rs914789857 Chr.11: 61800191-61800193 on Build GRCh38
rs955660022 Chr.11: 61800444-61800444 on Build GRCh38
rs923005906 Chr.11: 61800577-61800578 on Build GRCh38
rs976232688 Chr.11: 61800562-61800562 on Build GRCh38
rs989060240 Chr.11: 61800164-61800164 on Build GRCh38
rs557460309 Chr.11: 61800247-61800247 on Build GRCh38

View Full Product Details