Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61837698-61838193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS2
Entrez Gene ID
9415
Gene Name
fatty acid desaturase 2
Gene Aliases
D6D, DES6, FADSD6, LLCDL2, SLL0262, TU13
UniGene
Hs.502745
Gene Chromosome Location
Chr.11: 61816203-61867354 on Build GRCh38
Transcripts
NM_001281501.1 NM_001281502.1 NM_004265.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5373027 c.163C>T p.P55S Chr.11: 61837799-61837799 on Build GRCh38
4486090 c.300C>T p.S100S Chr.11: 61837870-61837870 on Build GRCh38
70678 c.230C>G p.P77R Chr.11: 61837800-61837800 on Build GRCh38
1205979 c.189C>A p.F63L Chr.11: 61837825-61837825 on Build GRCh38
5838988 c.245G>A p.G82D Chr.11: 61837881-61837881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753961325 Chr.11: 61837855-61837855 on Build GRCh38
rs748828482 Chr.11: 61837908-61837908 on Build GRCh38
rs917692375 Chr.11: 61837934-61837934 on Build GRCh38
rs570931348 Chr.11: 61838085-61838085 on Build GRCh38
rs774470752 Chr.11: 61838043-61838043 on Build GRCh38
rs373144076 Chr.11: 61837893-61837893 on Build GRCh38
rs577282649 Chr.11: 61837754-61837754 on Build GRCh38
rs377238578 Chr.11: 61837813-61837813 on Build GRCh38
rs1004420822 Chr.11: 61838036-61838036 on Build GRCh38
rs748559660 Chr.11: 61837760-61837760 on Build GRCh38
rs545952540 Chr.11: 61837789-61837789 on Build GRCh38
rs761842216 Chr.11: 61838049-61838049 on Build GRCh38
rs937866666 Chr.11: 61837798-61837798 on Build GRCh38
rs174578 Chr.11: 61838027-61838027 on Build GRCh38
rs889946870 Chr.11: 61837977-61837977 on Build GRCh38
rs761610364 Chr.11: 61837834-61837834 on Build GRCh38
rs959001360 Chr.11: 61838093-61838093 on Build GRCh38
rs936919686 Chr.11: 61837996-61837996 on Build GRCh38
rs752276658 Chr.11: 61838048-61838048 on Build GRCh38
rs942842913 Chr.11: 61838149-61838149 on Build GRCh38
rs140889931 Chr.11: 61837880-61837880 on Build GRCh38
rs373804840 Chr.11: 61837790-61837790 on Build GRCh38
rs2072114 Chr.11: 61837743-61837743 on Build GRCh38
rs760469083 Chr.11: 61837927-61837927 on Build GRCh38
rs181147340 Chr.11: 61837858-61837858 on Build GRCh38
rs1033220945 Chr.11: 61838002-61838002 on Build GRCh38
rs750294408 Chr.11: 61837848-61837848 on Build GRCh38
rs772777387 Chr.11: 61837769-61837769 on Build GRCh38
rs147302709 Chr.11: 61837783-61837783 on Build GRCh38
rs761491740 Chr.11: 61837912-61837912 on Build GRCh38
rs1042567436 Chr.11: 61837961-61837961 on Build GRCh38
rs75992720 Chr.11: 61838106-61838106 on Build GRCh38
rs771484619 Chr.11: 61837889-61837889 on Build GRCh38
rs772645024 Chr.11: 61837763-61837763 on Build GRCh38
rs773114496 Chr.11: 61837925-61837925 on Build GRCh38
rs369021206 Chr.11: 61837775-61837775 on Build GRCh38
rs1007503664 Chr.11: 61837979-61837979 on Build GRCh38
rs755180012 Chr.11: 61837857-61837857 on Build GRCh38
rs750670888 Chr.11: 61838092-61838092 on Build GRCh38
rs1000786193 Chr.11: 61837739-61837739 on Build GRCh38
rs761308406 Chr.11: 61837780-61837780 on Build GRCh38
rs369037848 Chr.11: 61838063-61838063 on Build GRCh38
rs562022986 Chr.11: 61837909-61837909 on Build GRCh38
rs945506406 Chr.11: 61837949-61837950 on Build GRCh38
rs776490403 Chr.11: 61838003-61838003 on Build GRCh38
rs773969818 Chr.11: 61837911-61837911 on Build GRCh38
rs1020590906 Chr.11: 61838024-61838024 on Build GRCh38
rs531049028 Chr.11: 61838015-61838015 on Build GRCh38
rs756136818 Chr.11: 61837734-61837734 on Build GRCh38
rs767209852 Chr.11: 61837917-61837917 on Build GRCh38
rs770713689 Chr.11: 61837722-61837722 on Build GRCh38
rs746791103 Chr.11: 61837913-61837913 on Build GRCh38
rs926274275 Chr.11: 61837903-61837903 on Build GRCh38
rs1054560566 Chr.11: 61838151-61838151 on Build GRCh38
rs1039293493 Chr.11: 61837997-61837997 on Build GRCh38
rs780249421 Chr.11: 61837742-61837742 on Build GRCh38
rs769052376 Chr.11: 61837753-61837753 on Build GRCh38
rs973695547 Chr.11: 61837895-61837895 on Build GRCh38
rs117750625 Chr.11: 61838136-61838136 on Build GRCh38
rs982324782 Chr.11: 61837788-61837788 on Build GRCh38
rs900500175 Chr.11: 61838000-61838000 on Build GRCh38
rs174579 Chr.11: 61838141-61838141 on Build GRCh38
rs900547709 Chr.11: 61837755-61837755 on Build GRCh38
rs1051638179 Chr.11: 61837945-61837945 on Build GRCh38
rs989145495 Chr.11: 61837751-61837751 on Build GRCh38
rs774065594 Chr.11: 61837832-61837832 on Build GRCh38
rs909837590 Chr.11: 61837838-61837838 on Build GRCh38
rs111760062 Chr.11: 61837776-61837776 on Build GRCh38
rs934107444 Chr.11: 61837924-61837924 on Build GRCh38
rs769154001 Chr.11: 61837787-61837787 on Build GRCh38
rs748433335 Chr.11: 61837879-61837879 on Build GRCh38
rs972702742 Chr.11: 61838094-61838094 on Build GRCh38
rs934013156 Chr.11: 61838112-61838112 on Build GRCh38
rs981867418 Chr.11: 61838029-61838029 on Build GRCh38
rs774797493 Chr.11: 61837906-61837906 on Build GRCh38
rs370195926 Chr.11: 61837850-61837850 on Build GRCh38
rs144799113 Chr.11: 61837888-61837888 on Build GRCh38
rs766352094 Chr.11: 61837851-61837851 on Build GRCh38
rs200220738 Chr.11: 61837811-61837811 on Build GRCh38
rs767777621 Chr.11: 61838070-61838070 on Build GRCh38

View Full Product Details