Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61848078-61848577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS2
Entrez Gene ID
9415
Gene Name
fatty acid desaturase 2
Gene Aliases
D6D, DES6, FADSD6, LLCDL2, SLL0262, TU13
UniGene
Hs.502745
Gene Chromosome Location
Chr.11: 61816203-61867354 on Build GRCh38
Transcripts
NM_001281501.1 NM_001281502.1 NM_004265.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
139902 c.649C>T p.H217Y Chr.11: 61848189-61848189 on Build GRCh38
5050949 c.708G>A p.L236L Chr.11: 61848248-61848248 on Build GRCh38
5125258 c.678+4C>T p.? Chr.11: 61848288-61848288 on Build GRCh38
6489782 c.636C>A p.N212K Chr.11: 61848242-61848242 on Build GRCh38
1205977 c.581G>A p.R194H Chr.11: 61848187-61848187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879074102 Chr.11: 61848405-61848406 on Build GRCh38
rs118041921 Chr.11: 61848288-61848288 on Build GRCh38
rs1027627757 Chr.11: 61848347-61848347 on Build GRCh38
rs775830458 Chr.11: 61848151-61848151 on Build GRCh38
rs35195180 Chr.11: 61848113-61848114 on Build GRCh38
rs966176905 Chr.11: 61848375-61848375 on Build GRCh38
rs534536802 Chr.11: 61848411-61848411 on Build GRCh38
rs2851682 Chr.11: 61848540-61848540 on Build GRCh38
rs556445068 Chr.11: 61848492-61848492 on Build GRCh38
rs116757889 Chr.11: 61848431-61848431 on Build GRCh38
rs770049599 Chr.11: 61848296-61848296 on Build GRCh38
rs1028140873 Chr.11: 61848101-61848101 on Build GRCh38
rs910359042 Chr.11: 61848387-61848387 on Build GRCh38
rs199900021 Chr.11: 61848204-61848204 on Build GRCh38
rs1049430775 Chr.11: 61848227-61848227 on Build GRCh38
rs778805012 Chr.11: 61848251-61848251 on Build GRCh38
rs770243987 Chr.11: 61848147-61848147 on Build GRCh38
rs144659223 Chr.11: 61848406-61848406 on Build GRCh38
rs766641933 Chr.11: 61848517-61848517 on Build GRCh38
rs174589 Chr.11: 61848331-61848331 on Build GRCh38
rs925606614 Chr.11: 61848420-61848420 on Build GRCh38
rs905807999 Chr.11: 61848330-61848330 on Build GRCh38
rs538650221 Chr.11: 61848418-61848418 on Build GRCh38
rs376130983 Chr.11: 61848346-61848346 on Build GRCh38
rs777016666 Chr.11: 61848143-61848143 on Build GRCh38
rs753617407 Chr.11: 61848233-61848233 on Build GRCh38
rs1047845056 Chr.11: 61848538-61848538 on Build GRCh38
rs763495959 Chr.11: 61848299-61848299 on Build GRCh38
rs747180677 Chr.11: 61848137-61848137 on Build GRCh38
rs754703896 Chr.11: 61848240-61848240 on Build GRCh38
rs746142255 Chr.11: 61848295-61848295 on Build GRCh38
rs558325682 Chr.11: 61848298-61848298 on Build GRCh38
rs913019719 Chr.11: 61848505-61848505 on Build GRCh38
rs878936101 Chr.11: 61848389-61848389 on Build GRCh38
rs769289525 Chr.11: 61848324-61848324 on Build GRCh38
rs749964341 Chr.11: 61848167-61848167 on Build GRCh38
rs1006367574 Chr.11: 61848541-61848541 on Build GRCh38
rs560131316 Chr.11: 61848145-61848145 on Build GRCh38
rs536835787 Chr.11: 61848442-61848442 on Build GRCh38
rs752621339 Chr.11: 61848252-61848252 on Build GRCh38
rs949849907 Chr.11: 61848533-61848533 on Build GRCh38
rs867184224 Chr.11: 61848289-61848289 on Build GRCh38
rs368031408 Chr.11: 61848118-61848118 on Build GRCh38
rs1015002644 Chr.11: 61848365-61848365 on Build GRCh38
rs771065351 Chr.11: 61848139-61848139 on Build GRCh38
rs748127745 Chr.11: 61848128-61848128 on Build GRCh38
rs922244830 Chr.11: 61848168-61848168 on Build GRCh38
rs376981471 Chr.11: 61848332-61848332 on Build GRCh38
rs768595850 Chr.11: 61848457-61848463 on Build GRCh38
rs1014134925 Chr.11: 61848303-61848303 on Build GRCh38
rs922161056 Chr.11: 61848402-61848402 on Build GRCh38
rs777891525 Chr.11: 61848136-61848136 on Build GRCh38
rs371340901 Chr.11: 61848110-61848110 on Build GRCh38
rs952140270 Chr.11: 61848103-61848103 on Build GRCh38
rs375947275 Chr.11: 61848152-61848152 on Build GRCh38
rs567790006 Chr.11: 61848433-61848433 on Build GRCh38
rs993461156 Chr.11: 61848518-61848518 on Build GRCh38
rs771137353 Chr.11: 61848279-61848279 on Build GRCh38
rs1045592097 Chr.11: 61848535-61848535 on Build GRCh38
rs149777687 Chr.11: 61848203-61848203 on Build GRCh38
rs1004883798 Chr.11: 61848353-61848353 on Build GRCh38
rs747125681 Chr.11: 61848269-61848269 on Build GRCh38
rs139762727 Chr.11: 61848507-61848507 on Build GRCh38
rs540099308 Chr.11: 61848134-61848134 on Build GRCh38
rs966185204 Chr.11: 61848142-61848142 on Build GRCh38
rs947445499 Chr.11: 61848358-61848358 on Build GRCh38
rs776612441 Chr.11: 61848544-61848545 on Build GRCh38
rs750572087 Chr.11: 61848428-61848428 on Build GRCh38
rs551820933 Chr.11: 61848361-61848361 on Build GRCh38
rs529084714 Chr.11: 61848154-61848154 on Build GRCh38
rs758350373 Chr.11: 61848266-61848266 on Build GRCh38
rs952056994 Chr.11: 61848350-61848350 on Build GRCh38
rs370618898 Chr.11: 61848483-61848483 on Build GRCh38
rs1052851058 Chr.11: 61848493-61848493 on Build GRCh38
rs765667632 Chr.11: 61848426-61848426 on Build GRCh38
rs34883317 Chr.11: 61848136-61848137 on Build GRCh38
rs553426992 Chr.11: 61848267-61848267 on Build GRCh38

View Full Product Details