Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61967568-61968068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTH1
Entrez Gene ID
2495
Gene Name
ferritin heavy chain 1
Gene Aliases
FHC, FTH, FTHL6, HFE5, PIG15, PLIF
UniGene
Hs.524910
Gene Chromosome Location
Chr.11: 61964285-61967660 on Build GRCh38
Transcripts
NM_002032.2

Gene Symbol
LOC399900
Entrez Gene ID
399900
Gene Name
uncharacterized LOC399900
Gene Aliases
-
UniGene
Hs.528437
Gene Chromosome Location
Chr.11: 61967798-61969275 on Build GRCh38
Transcripts
NM_002032.2

SNP Detail

SNP ID Chromosome Location
rs777594256 Chr.11: 61967608-61967608 on Build GRCh38
rs35224707 Chr.11: 61968014-61968014 on Build GRCh38
rs903885051 Chr.11: 61967811-61967811 on Build GRCh38
rs959130699 Chr.11: 61967884-61967884 on Build GRCh38
rs976154463 Chr.11: 61967611-61967611 on Build GRCh38
rs979629275 Chr.11: 61967780-61967780 on Build GRCh38
rs896709794 Chr.11: 61967807-61967807 on Build GRCh38
rs886048433 Chr.11: 61967629-61967629 on Build GRCh38
rs966179424 Chr.11: 61967605-61967605 on Build GRCh38
rs746763203 Chr.11: 61967642-61967642 on Build GRCh38
rs887596455 Chr.11: 61967910-61967910 on Build GRCh38
rs776621212 Chr.11: 61967649-61967649 on Build GRCh38
rs968705368 Chr.11: 61967643-61967643 on Build GRCh38
rs1005191081 Chr.11: 61967754-61967754 on Build GRCh38
rs1001338219 Chr.11: 61967919-61967919 on Build GRCh38
rs927080078 Chr.11: 61967877-61967877 on Build GRCh38
rs387906549 Chr.11: 61967589-61967589 on Build GRCh38
rs929006617 Chr.11: 61967978-61967978 on Build GRCh38
rs570386508 Chr.11: 61967612-61967612 on Build GRCh38
rs555788021 Chr.11: 61967828-61967828 on Build GRCh38
rs1032837210 Chr.11: 61967707-61967707 on Build GRCh38
rs926792862 Chr.11: 61967776-61967776 on Build GRCh38
rs899653989 Chr.11: 61967856-61967856 on Build GRCh38
rs937778665 Chr.11: 61967917-61967917 on Build GRCh38
rs764392577 Chr.11: 61967679-61967679 on Build GRCh38
rs540799673 Chr.11: 61967925-61967925 on Build GRCh38
rs114778979 Chr.11: 61967704-61967704 on Build GRCh38
rs953531326 Chr.11: 61967779-61967779 on Build GRCh38
rs1045434304 Chr.11: 61967694-61967694 on Build GRCh38
rs945639038 Chr.11: 61968025-61968025 on Build GRCh38
rs775906846 Chr.11: 61967640-61967640 on Build GRCh38
rs1028988413 Chr.11: 61967916-61967916 on Build GRCh38
rs988784899 Chr.11: 61967825-61967825 on Build GRCh38
rs937091385 Chr.11: 61967882-61967882 on Build GRCh38
rs868158002 Chr.11: 61967715-61967715 on Build GRCh38
rs377693938 Chr.11: 61967832-61967832 on Build GRCh38
rs912870540 Chr.11: 61967656-61967656 on Build GRCh38
rs555401255 Chr.11: 61967810-61967810 on Build GRCh38
rs970946609 Chr.11: 61968031-61968031 on Build GRCh38
rs979681287 Chr.11: 61967635-61967635 on Build GRCh38
rs185835490 Chr.11: 61967809-61967809 on Build GRCh38
rs1036663252 Chr.11: 61967913-61967913 on Build GRCh38
rs114865849 Chr.11: 61967915-61967915 on Build GRCh38
rs988413980 Chr.11: 61967989-61967989 on Build GRCh38
rs34316319 Chr.11: 61967913-61967914 on Build GRCh38
rs1021241587 Chr.11: 61967634-61967634 on Build GRCh38
rs991892376 Chr.11: 61967956-61967956 on Build GRCh38
rs200583963 Chr.11: 61967918-61967919 on Build GRCh38
rs763987917 Chr.11: 61967909-61967910 on Build GRCh38
rs747618890 Chr.11: 61967594-61967594 on Build GRCh38
rs868452793 Chr.11: 61967616-61967616 on Build GRCh38
rs578245779 Chr.11: 61967918-61967918 on Build GRCh38
rs941127151 Chr.11: 61967629-61967629 on Build GRCh38
rs908258843 Chr.11: 61967598-61967598 on Build GRCh38
rs763318260 Chr.11: 61967646-61967646 on Build GRCh38
rs982446384 Chr.11: 61967752-61967752 on Build GRCh38
rs922106202 Chr.11: 61967617-61967617 on Build GRCh38
rs949684348 Chr.11: 61967837-61967837 on Build GRCh38
rs955486207 Chr.11: 61967744-61967744 on Build GRCh38
rs756637953 Chr.11: 61967587-61967587 on Build GRCh38
rs962395685 Chr.11: 61967914-61967915 on Build GRCh38
rs878918644 Chr.11: 61967654-61967654 on Build GRCh38
rs953501245 Chr.11: 61967630-61967630 on Build GRCh38
rs143760071 Chr.11: 61967910-61967911 on Build GRCh38
rs935573685 Chr.11: 61967644-61967644 on Build GRCh38
rs901814361 Chr.11: 61967763-61967763 on Build GRCh38
rs546464863 Chr.11: 61967774-61967774 on Build GRCh38
rs955280735 Chr.11: 61967911-61967912 on Build GRCh38
rs973830487 Chr.11: 61967766-61967766 on Build GRCh38
rs60961005 Chr.11: 61967919-61967920 on Build GRCh38
rs963912142 Chr.11: 61967709-61967709 on Build GRCh38
rs1049362478 Chr.11: 61967712-61967712 on Build GRCh38
rs886048434 Chr.11: 61967656-61967656 on Build GRCh38
rs1056630007 Chr.11: 61967796-61967796 on Build GRCh38
rs1021391103 Chr.11: 61967681-61967681 on Build GRCh38
rs1029039740 Chr.11: 61967778-61967778 on Build GRCh38
rs937142161 Chr.11: 61967705-61967705 on Build GRCh38
rs1009540087 Chr.11: 61967680-61967680 on Build GRCh38
rs796258363 Chr.11: 61967914-61967914 on Build GRCh38
rs73493231 Chr.11: 61967949-61967949 on Build GRCh38
rs758012761 Chr.11: 61967599-61967599 on Build GRCh38
rs866119662 Chr.11: 61967627-61967627 on Build GRCh38
rs917651404 Chr.11: 61967961-61967961 on Build GRCh38
rs908208583 Chr.11: 61967757-61967757 on Build GRCh38
rs1017725843 Chr.11: 61967843-61967843 on Build GRCh38
rs896461553 Chr.11: 61967658-61967658 on Build GRCh38
rs981165495 Chr.11: 61967855-61967855 on Build GRCh38
rs766985190 Chr.11: 61967938-61967938 on Build GRCh38
rs1025614461 Chr.11: 61968027-61968027 on Build GRCh38
rs997618346 Chr.11: 61967777-61967777 on Build GRCh38
rs371399327 Chr.11: 61967864-61967864 on Build GRCh38
rs745801010 Chr.11: 61967650-61967650 on Build GRCh38
rs1014957142 Chr.11: 61967760-61967760 on Build GRCh38
rs957061850 Chr.11: 61967957-61967957 on Build GRCh38
rs571926915 Chr.11: 61967912-61967912 on Build GRCh38
rs778481941 Chr.11: 61967592-61967592 on Build GRCh38
rs949831394 Chr.11: 61967674-61967674 on Build GRCh38

View Full Product Details