Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62625165-62625660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GANAB
Entrez Gene ID
23193
Gene Name
glucosidase II alpha subunit
Gene Aliases
G2AN, GIIA, GLUII
UniGene
Hs.595071
Gene Chromosome Location
Chr.11: 62624826-62646726 on Build GRCh38
Transcripts
NM_001278192.1 NM_001278193.1 NM_001278194.1 NM_198334.2 NM_198335.3 XM_017017412.1 XM_017017413.1 XM_017017414.1

SNP Detail

SNP ID Chromosome Location
rs1041016691 Chr.11: 62625245-62625245 on Build GRCh38
rs763336696 Chr.11: 62625365-62625365 on Build GRCh38
rs768066030 Chr.11: 62625574-62625574 on Build GRCh38
rs180937079 Chr.11: 62625552-62625552 on Build GRCh38
rs764374413 Chr.11: 62625410-62625410 on Build GRCh38
rs746302915 Chr.11: 62625293-62625293 on Build GRCh38
rs779598404 Chr.11: 62625635-62625635 on Build GRCh38
rs568309359 Chr.11: 62625191-62625191 on Build GRCh38
rs917536739 Chr.11: 62625228-62625228 on Build GRCh38
rs992957435 Chr.11: 62625499-62625499 on Build GRCh38
rs1815835 Chr.11: 62625229-62625229 on Build GRCh38
rs968865554 Chr.11: 62625414-62625414 on Build GRCh38
rs6916 Chr.11: 62625404-62625404 on Build GRCh38
rs757819322 Chr.11: 62625570-62625570 on Build GRCh38
rs993031160 Chr.11: 62625227-62625227 on Build GRCh38
rs1049827273 Chr.11: 62625216-62625216 on Build GRCh38
rs542383941 Chr.11: 62625546-62625546 on Build GRCh38
rs944107950 Chr.11: 62625451-62625451 on Build GRCh38
rs1047579516 Chr.11: 62625289-62625289 on Build GRCh38
rs907961710 Chr.11: 62625588-62625588 on Build GRCh38
rs765423503 Chr.11: 62625214-62625214 on Build GRCh38
rs111241392 Chr.11: 62625306-62625306 on Build GRCh38
rs770321873 Chr.11: 62625295-62625295 on Build GRCh38
rs564470422 Chr.11: 62625595-62625595 on Build GRCh38
rs929255748 Chr.11: 62625517-62625517 on Build GRCh38
rs996757143 Chr.11: 62625619-62625619 on Build GRCh38
rs1050135407 Chr.11: 62625624-62625624 on Build GRCh38
rs556867025 Chr.11: 62625292-62625292 on Build GRCh38
rs911304334 Chr.11: 62625441-62625441 on Build GRCh38
rs557002169 Chr.11: 62625594-62625594 on Build GRCh38
rs980787363 Chr.11: 62625572-62625572 on Build GRCh38
rs917472962 Chr.11: 62625547-62625547 on Build GRCh38
rs934858110 Chr.11: 62625224-62625224 on Build GRCh38
rs1044201524 Chr.11: 62625584-62625584 on Build GRCh38
rs528749214 Chr.11: 62625319-62625319 on Build GRCh38
rs901098228 Chr.11: 62625349-62625349 on Build GRCh38
rs1010526508 Chr.11: 62625628-62625628 on Build GRCh38
rs551774837 Chr.11: 62625523-62625523 on Build GRCh38
rs573076836 Chr.11: 62625459-62625459 on Build GRCh38
rs117685098 Chr.11: 62625583-62625583 on Build GRCh38
rs969792175 Chr.11: 62625296-62625297 on Build GRCh38
rs553182716 Chr.11: 62625429-62625429 on Build GRCh38
rs772801273 Chr.11: 62625337-62625337 on Build GRCh38
rs776585548 Chr.11: 62625351-62625351 on Build GRCh38
rs993352744 Chr.11: 62625267-62625267 on Build GRCh38
rs968811908 Chr.11: 62625395-62625395 on Build GRCh38
rs574453455 Chr.11: 62625420-62625422 on Build GRCh38
rs575855623 Chr.11: 62625568-62625568 on Build GRCh38
rs370839982 Chr.11: 62625335-62625335 on Build GRCh38
rs759626724 Chr.11: 62625189-62625189 on Build GRCh38
rs111294759 Chr.11: 62625350-62625350 on Build GRCh38
rs771117093 Chr.11: 62625329-62625329 on Build GRCh38
rs1031415130 Chr.11: 62625444-62625444 on Build GRCh38
rs914771881 Chr.11: 62625371-62625371 on Build GRCh38
rs1815836 Chr.11: 62625226-62625226 on Build GRCh38
rs1044417980 Chr.11: 62625254-62625254 on Build GRCh38
rs749943092 Chr.11: 62625587-62625587 on Build GRCh38
rs766091367 Chr.11: 62625351-62625351 on Build GRCh38
rs944039780 Chr.11: 62625241-62625241 on Build GRCh38
rs114214376 Chr.11: 62625221-62625221 on Build GRCh38
rs760490276 Chr.11: 62625339-62625339 on Build GRCh38
rs185154133 Chr.11: 62625613-62625613 on Build GRCh38
rs576575660 Chr.11: 62625299-62625299 on Build GRCh38
rs891453680 Chr.11: 62625628-62625630 on Build GRCh38
rs752073689 Chr.11: 62625437-62625437 on Build GRCh38
rs1028006567 Chr.11: 62625367-62625367 on Build GRCh38
rs901771165 Chr.11: 62625257-62625257 on Build GRCh38
rs948687116 Chr.11: 62625238-62625238 on Build GRCh38

View Full Product Details