Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62625558-62626059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GANAB
Entrez Gene ID
23193
Gene Name
glucosidase II alpha subunit
Gene Aliases
G2AN, GIIA, GLUII
UniGene
Hs.595071
Gene Chromosome Location
Chr.11: 62624826-62646726 on Build GRCh38
Transcripts
NM_001278192.1 NM_001278193.1 NM_001278194.1 NM_198334.2 NM_198335.3 XM_017017412.1 XM_017017413.1 XM_017017414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5892128 c.2839G>T p.V947L Chr.11: 62625877-62625877 on Build GRCh38
5616540 c.2726-9C>T p.? Chr.11: 62625933-62625933 on Build GRCh38
3700110 c.2822A>G p.H941R Chr.11: 62625894-62625894 on Build GRCh38
3451047 c.2760C>T p.D920D Chr.11: 62625890-62625890 on Build GRCh38
5892129 c.2773G>T p.V925L Chr.11: 62625877-62625877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747679741 Chr.11: 62625842-62625842 on Build GRCh38
rs535683452 Chr.11: 62625983-62625983 on Build GRCh38
rs200956498 Chr.11: 62625804-62625804 on Build GRCh38
rs763042174 Chr.11: 62626018-62626018 on Build GRCh38
rs1024511087 Chr.11: 62625989-62625989 on Build GRCh38
rs781470089 Chr.11: 62625820-62625820 on Build GRCh38
rs779598404 Chr.11: 62625635-62625635 on Build GRCh38
rs547099748 Chr.11: 62625676-62625676 on Build GRCh38
rs750799764 Chr.11: 62625930-62625930 on Build GRCh38
rs1021882378 Chr.11: 62625683-62625683 on Build GRCh38
rs770553981 Chr.11: 62625868-62625868 on Build GRCh38
rs866625049 Chr.11: 62626009-62626009 on Build GRCh38
rs976850831 Chr.11: 62625736-62625736 on Build GRCh38
rs141471770 Chr.11: 62625745-62625745 on Build GRCh38
rs754185984 Chr.11: 62626034-62626034 on Build GRCh38
rs976188802 Chr.11: 62625849-62625849 on Build GRCh38
rs573683091 Chr.11: 62625644-62625644 on Build GRCh38
rs767655408 Chr.11: 62626022-62626022 on Build GRCh38
rs757621446 Chr.11: 62625912-62625912 on Build GRCh38
rs772852228 Chr.11: 62625859-62625859 on Build GRCh38
rs144153823 Chr.11: 62625921-62625921 on Build GRCh38
rs765191082 Chr.11: 62625794-62625794 on Build GRCh38
rs796525964 Chr.11: 62625757-62625757 on Build GRCh38
rs968925124 Chr.11: 62625700-62625700 on Build GRCh38
rs1044641546 Chr.11: 62625809-62625809 on Build GRCh38
rs765193755 Chr.11: 62625788-62625788 on Build GRCh38
rs920885991 Chr.11: 62625762-62625762 on Build GRCh38
rs907961710 Chr.11: 62625588-62625588 on Build GRCh38
rs564470422 Chr.11: 62625595-62625595 on Build GRCh38
rs762854787 Chr.11: 62625942-62625942 on Build GRCh38
rs749724558 Chr.11: 62625942-62625942 on Build GRCh38
rs756451391 Chr.11: 62625934-62625934 on Build GRCh38
rs996757143 Chr.11: 62625619-62625619 on Build GRCh38
rs1050135407 Chr.11: 62625624-62625624 on Build GRCh38
rs575897355 Chr.11: 62626021-62626021 on Build GRCh38
rs751678133 Chr.11: 62625910-62625910 on Build GRCh38
rs557002169 Chr.11: 62625594-62625594 on Build GRCh38
rs760261107 Chr.11: 62625864-62625864 on Build GRCh38
rs891507158 Chr.11: 62625846-62625846 on Build GRCh38
rs746240475 Chr.11: 62625824-62625824 on Build GRCh38
rs986046189 Chr.11: 62625707-62625707 on Build GRCh38
rs770241716 Chr.11: 62625825-62625825 on Build GRCh38
rs202077418 Chr.11: 62626036-62626036 on Build GRCh38
rs775449009 Chr.11: 62625938-62625939 on Build GRCh38
rs1044201524 Chr.11: 62625584-62625584 on Build GRCh38
rs952041936 Chr.11: 62625764-62625764 on Build GRCh38
rs145162136 Chr.11: 62625969-62625969 on Build GRCh38
rs775583375 Chr.11: 62626015-62626015 on Build GRCh38
rs1010526508 Chr.11: 62625628-62625628 on Build GRCh38
rs117685098 Chr.11: 62625583-62625583 on Build GRCh38
rs752771425 Chr.11: 62625792-62625792 on Build GRCh38
rs371161821 Chr.11: 62625844-62625844 on Build GRCh38
rs146543067 Chr.11: 62625734-62625734 on Build GRCh38
rs145576494 Chr.11: 62625851-62625851 on Build GRCh38
rs932510165 Chr.11: 62625889-62625889 on Build GRCh38
rs1031341434 Chr.11: 62625702-62625702 on Build GRCh38
rs536835261 Chr.11: 62625701-62625701 on Build GRCh38
rs181231903 Chr.11: 62625973-62625973 on Build GRCh38
rs757913089 Chr.11: 62625955-62625955 on Build GRCh38
rs185278436 Chr.11: 62626026-62626026 on Build GRCh38
rs763854072 Chr.11: 62625941-62625942 on Build GRCh38
rs759356524 Chr.11: 62625880-62625880 on Build GRCh38
rs369751066 Chr.11: 62626014-62626014 on Build GRCh38
rs904179937 Chr.11: 62625678-62625678 on Build GRCh38
rs149726248 Chr.11: 62625819-62625819 on Build GRCh38
rs756435491 Chr.11: 62626031-62626031 on Build GRCh38
rs374003721 Chr.11: 62625668-62625668 on Build GRCh38
rs965565674 Chr.11: 62626024-62626024 on Build GRCh38
rs559615967 Chr.11: 62625686-62625686 on Build GRCh38
rs753825555 Chr.11: 62625769-62625769 on Build GRCh38
rs1013947232 Chr.11: 62625984-62625984 on Build GRCh38
rs189567845 Chr.11: 62625735-62625735 on Build GRCh38
rs776300183 Chr.11: 62625970-62625970 on Build GRCh38
rs749943092 Chr.11: 62625587-62625587 on Build GRCh38
rs140322818 Chr.11: 62625909-62625909 on Build GRCh38
rs777319548 Chr.11: 62625956-62625956 on Build GRCh38
rs766704400 Chr.11: 62626033-62626033 on Build GRCh38
rs572863695 Chr.11: 62625981-62625981 on Build GRCh38
rs778797237 Chr.11: 62625760-62625760 on Build GRCh38
rs570095749 Chr.11: 62625900-62625900 on Build GRCh38
rs185154133 Chr.11: 62625613-62625613 on Build GRCh38
rs747948615 Chr.11: 62625801-62625801 on Build GRCh38
rs754829169 Chr.11: 62625749-62625749 on Build GRCh38
rs891453680 Chr.11: 62625628-62625630 on Build GRCh38
rs759447048 Chr.11: 62625779-62625779 on Build GRCh38
rs553565070 Chr.11: 62625773-62625773 on Build GRCh38
rs112023179 Chr.11: 62625985-62625985 on Build GRCh38
rs368019259 Chr.11: 62625968-62625968 on Build GRCh38
rs199853864 Chr.11: 62626019-62626019 on Build GRCh38
rs375587198 Chr.11: 62625881-62625881 on Build GRCh38
rs550198965 Chr.11: 62625874-62625874 on Build GRCh38
rs780588721 Chr.11: 62625831-62625831 on Build GRCh38
rs780455543 Chr.11: 62625938-62625938 on Build GRCh38
rs755542361 Chr.11: 62626037-62626037 on Build GRCh38

View Full Product Details