Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62625975-62626473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GANAB
Entrez Gene ID
23193
Gene Name
glucosidase II alpha subunit
Gene Aliases
G2AN, GIIA, GLUII
UniGene
Hs.595071
Gene Chromosome Location
Chr.11: 62624826-62646726 on Build GRCh38
Transcripts
NM_001278192.1 NM_001278193.1 NM_001278194.1 NM_198334.2 NM_198335.3 XM_017017412.1 XM_017017413.1 XM_017017414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6133714 c.2649T>C p.F883F Chr.11: 62626141-62626141 on Build GRCh38
1298314 c.2526A>T p.G842G Chr.11: 62626433-62626433 on Build GRCh38
6669222 c.2738G>A p.R913Q Chr.11: 62626118-62626118 on Build GRCh38
3953583 c.2598A>G p.S866S Chr.11: 62626361-62626361 on Build GRCh38
4812653 c.2592A>T p.G864G Chr.11: 62626433-62626433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985703436 Chr.11: 62626235-62626235 on Build GRCh38
rs779372884 Chr.11: 62626040-62626040 on Build GRCh38
rs200312082 Chr.11: 62626088-62626088 on Build GRCh38
rs778905557 Chr.11: 62626385-62626385 on Build GRCh38
rs763042174 Chr.11: 62626018-62626018 on Build GRCh38
rs759815795 Chr.11: 62626429-62626429 on Build GRCh38
rs373851834 Chr.11: 62626326-62626326 on Build GRCh38
rs567172979 Chr.11: 62626348-62626348 on Build GRCh38
rs200846285 Chr.11: 62626053-62626053 on Build GRCh38
rs370022389 Chr.11: 62626295-62626295 on Build GRCh38
rs113173056 Chr.11: 62626062-62626062 on Build GRCh38
rs191769278 Chr.11: 62626305-62626305 on Build GRCh38
rs796143596 Chr.11: 62626146-62626146 on Build GRCh38
rs376691473 Chr.11: 62626210-62626210 on Build GRCh38
rs775964626 Chr.11: 62626444-62626444 on Build GRCh38
rs377356565 Chr.11: 62626358-62626358 on Build GRCh38
rs901380152 Chr.11: 62626266-62626266 on Build GRCh38
rs763584288 Chr.11: 62626330-62626330 on Build GRCh38
rs866625049 Chr.11: 62626009-62626009 on Build GRCh38
rs767143750 Chr.11: 62626215-62626218 on Build GRCh38
rs202238090 Chr.11: 62626200-62626200 on Build GRCh38
rs548130698 Chr.11: 62626253-62626253 on Build GRCh38
rs747146421 Chr.11: 62626393-62626393 on Build GRCh38
rs927236391 Chr.11: 62626372-62626372 on Build GRCh38
rs776795874 Chr.11: 62626314-62626314 on Build GRCh38
rs1025741030 Chr.11: 62626279-62626279 on Build GRCh38
rs749062477 Chr.11: 62626093-62626093 on Build GRCh38
rs746547413 Chr.11: 62626041-62626041 on Build GRCh38
rs775692598 Chr.11: 62626118-62626118 on Build GRCh38
rs754185984 Chr.11: 62626034-62626034 on Build GRCh38
rs767566866 Chr.11: 62626131-62626131 on Build GRCh38
rs767655408 Chr.11: 62626022-62626022 on Build GRCh38
rs372681163 Chr.11: 62626044-62626044 on Build GRCh38
rs751369780 Chr.11: 62626194-62626194 on Build GRCh38
rs773316721 Chr.11: 62626182-62626182 on Build GRCh38
rs780813693 Chr.11: 62626204-62626204 on Build GRCh38
rs887968214 Chr.11: 62626374-62626374 on Build GRCh38
rs772083807 Chr.11: 62626389-62626389 on Build GRCh38
rs760636927 Chr.11: 62626309-62626309 on Build GRCh38
rs749116810 Chr.11: 62626285-62626285 on Build GRCh38
rs531070731 Chr.11: 62626075-62626075 on Build GRCh38
rs949762250 Chr.11: 62626290-62626290 on Build GRCh38
rs935904904 Chr.11: 62626248-62626248 on Build GRCh38
rs904175508 Chr.11: 62626243-62626243 on Build GRCh38
rs759848415 Chr.11: 62626198-62626198 on Build GRCh38
rs575897355 Chr.11: 62626021-62626021 on Build GRCh38
rs768762083 Chr.11: 62626108-62626108 on Build GRCh38
rs138894150 Chr.11: 62626239-62626239 on Build GRCh38
rs778997632 Chr.11: 62626297-62626297 on Build GRCh38
rs749999508 Chr.11: 62626350-62626350 on Build GRCh38
rs960195651 Chr.11: 62626378-62626378 on Build GRCh38
rs374848841 Chr.11: 62626446-62626446 on Build GRCh38
rs202077418 Chr.11: 62626036-62626036 on Build GRCh38
rs769317810 Chr.11: 62626081-62626081 on Build GRCh38
rs201907445 Chr.11: 62626137-62626137 on Build GRCh38
rs775583375 Chr.11: 62626015-62626015 on Build GRCh38
rs769999391 Chr.11: 62626441-62626441 on Build GRCh38
rs976156325 Chr.11: 62626208-62626208 on Build GRCh38
rs771167129 Chr.11: 62626313-62626313 on Build GRCh38
rs779832857 Chr.11: 62626284-62626284 on Build GRCh38
rs748057362 Chr.11: 62626303-62626303 on Build GRCh38
rs753140462 Chr.11: 62626199-62626199 on Build GRCh38
rs1027872232 Chr.11: 62626066-62626066 on Build GRCh38
rs886037848 Chr.11: 62626328-62626333 on Build GRCh38
rs756866414 Chr.11: 62626199-62626200 on Build GRCh38
rs749980643 Chr.11: 62626329-62626330 on Build GRCh38
rs765530886 Chr.11: 62626321-62626321 on Build GRCh38
rs998369610 Chr.11: 62626274-62626274 on Build GRCh38
rs185278436 Chr.11: 62626026-62626026 on Build GRCh38
rs766580909 Chr.11: 62626195-62626195 on Build GRCh38
rs369751066 Chr.11: 62626014-62626014 on Build GRCh38
rs114915323 Chr.11: 62626411-62626411 on Build GRCh38
rs777546492 Chr.11: 62626390-62626390 on Build GRCh38
rs747888174 Chr.11: 62626387-62626387 on Build GRCh38
rs756435491 Chr.11: 62626031-62626031 on Build GRCh38
rs774379348 Chr.11: 62626116-62626116 on Build GRCh38
rs965565674 Chr.11: 62626024-62626024 on Build GRCh38
rs764479913 Chr.11: 62626331-62626331 on Build GRCh38
rs755832946 Chr.11: 62626213-62626213 on Build GRCh38
rs146501208 Chr.11: 62626371-62626371 on Build GRCh38
rs770934031 Chr.11: 62626398-62626398 on Build GRCh38
rs868572195 Chr.11: 62626222-62626222 on Build GRCh38
rs550777375 Chr.11: 62626154-62626154 on Build GRCh38
rs148970599 Chr.11: 62626258-62626258 on Build GRCh38
rs769876299 Chr.11: 62626114-62626114 on Build GRCh38
rs754238978 Chr.11: 62626196-62626196 on Build GRCh38
rs763103082 Chr.11: 62626319-62626319 on Build GRCh38
rs766704400 Chr.11: 62626033-62626033 on Build GRCh38
rs754833448 Chr.11: 62626377-62626377 on Build GRCh38
rs756754923 Chr.11: 62626043-62626043 on Build GRCh38
rs370213372 Chr.11: 62626368-62626368 on Build GRCh38
rs932461208 Chr.11: 62626226-62626226 on Build GRCh38
rs199853864 Chr.11: 62626019-62626019 on Build GRCh38
rs753473634 Chr.11: 62626376-62626376 on Build GRCh38
rs1039951308 Chr.11: 62626264-62626264 on Build GRCh38
rs755542361 Chr.11: 62626037-62626037 on Build GRCh38

View Full Product Details