Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62626918-62627444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GANAB
Entrez Gene ID
23193
Gene Name
glucosidase II alpha subunit
Gene Aliases
G2AN, GIIA, GLUII
UniGene
Hs.595071
Gene Chromosome Location
Chr.11: 62624826-62646726 on Build GRCh38
Transcripts
NM_001278192.1 NM_001278193.1 NM_001278194.1 NM_198334.2 NM_198335.3 XM_017017412.1 XM_017017413.1 XM_017017414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1162961 c.2257C>T p.L753L Chr.11: 62627113-62627113 on Build GRCh38
6669227 c.2199C>T p.Y733Y Chr.11: 62627335-62627335 on Build GRCh38
1704147 c.2252T>A p.L751Q Chr.11: 62627348-62627348 on Build GRCh38
6669218 c.2319G>A p.A773A Chr.11: 62627117-62627117 on Build GRCh38
1676102 c.2290G>T p.G764C Chr.11: 62627080-62627080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905289346 Chr.11: 62626996-62626996 on Build GRCh38
rs1014218282 Chr.11: 62627106-62627106 on Build GRCh38
rs192389541 Chr.11: 62627236-62627236 on Build GRCh38
rs764389221 Chr.11: 62627025-62627025 on Build GRCh38
rs765237116 Chr.11: 62627133-62627133 on Build GRCh38
rs746612506 Chr.11: 62627264-62627264 on Build GRCh38
rs757605917 Chr.11: 62627048-62627048 on Build GRCh38
rs868816483 Chr.11: 62627121-62627121 on Build GRCh38
rs745636112 Chr.11: 62627293-62627293 on Build GRCh38
rs750746389 Chr.11: 62627175-62627175 on Build GRCh38
rs769875477 Chr.11: 62627294-62627294 on Build GRCh38
rs137897818 Chr.11: 62627193-62627193 on Build GRCh38
rs371022745 Chr.11: 62627246-62627246 on Build GRCh38
rs896469685 Chr.11: 62627058-62627058 on Build GRCh38
rs147361014 Chr.11: 62627182-62627182 on Build GRCh38
rs530752893 Chr.11: 62626962-62626962 on Build GRCh38
rs201994619 Chr.11: 62627093-62627093 on Build GRCh38
rs971828323 Chr.11: 62627197-62627197 on Build GRCh38
rs778458461 Chr.11: 62627066-62627066 on Build GRCh38
rs781689114 Chr.11: 62626946-62626946 on Build GRCh38
rs772872094 Chr.11: 62627086-62627086 on Build GRCh38
rs747338622 Chr.11: 62626986-62626987 on Build GRCh38
rs747622584 Chr.11: 62627067-62627067 on Build GRCh38
rs1033238356 Chr.11: 62627167-62627167 on Build GRCh38
rs564051409 Chr.11: 62626988-62626988 on Build GRCh38
rs1020057727 Chr.11: 62627071-62627071 on Build GRCh38
rs1050736531 Chr.11: 62626986-62626986 on Build GRCh38
rs763082802 Chr.11: 62627158-62627158 on Build GRCh38
rs7949030 Chr.11: 62627409-62627409 on Build GRCh38
rs775761544 Chr.11: 62627138-62627138 on Build GRCh38
rs368563939 Chr.11: 62627176-62627176 on Build GRCh38
rs771079554 Chr.11: 62627147-62627148 on Build GRCh38
rs751740452 Chr.11: 62627170-62627170 on Build GRCh38
rs758734849 Chr.11: 62627019-62627019 on Build GRCh38
rs1020214808 Chr.11: 62627107-62627107 on Build GRCh38
rs573953588 Chr.11: 62627316-62627316 on Build GRCh38
rs757863937 Chr.11: 62627257-62627257 on Build GRCh38
rs750845882 Chr.11: 62627317-62627317 on Build GRCh38
rs774283742 Chr.11: 62627312-62627312 on Build GRCh38
rs143779086 Chr.11: 62627308-62627308 on Build GRCh38
rs377549675 Chr.11: 62626963-62626963 on Build GRCh38
rs940802827 Chr.11: 62626959-62626959 on Build GRCh38
rs183266881 Chr.11: 62627243-62627243 on Build GRCh38
rs958575447 Chr.11: 62627162-62627162 on Build GRCh38
rs140219733 Chr.11: 62627118-62627118 on Build GRCh38
rs776590206 Chr.11: 62626982-62626982 on Build GRCh38
rs774004006 Chr.11: 62626961-62626961 on Build GRCh38
rs373852510 Chr.11: 62627255-62627255 on Build GRCh38
rs770691650 Chr.11: 62627099-62627099 on Build GRCh38
rs377045951 Chr.11: 62627262-62627262 on Build GRCh38
rs781081597 Chr.11: 62627278-62627278 on Build GRCh38
rs142739405 Chr.11: 62627180-62627180 on Build GRCh38
rs908807407 Chr.11: 62627201-62627201 on Build GRCh38
rs370977625 Chr.11: 62626990-62626990 on Build GRCh38
rs371649150 Chr.11: 62626993-62626993 on Build GRCh38
rs766901948 Chr.11: 62627323-62627323 on Build GRCh38
rs1048839150 Chr.11: 62626952-62626952 on Build GRCh38
rs775532479 Chr.11: 62627305-62627305 on Build GRCh38
rs757578887 Chr.11: 62627171-62627171 on Build GRCh38
rs771755933 Chr.11: 62627085-62627085 on Build GRCh38
rs756597402 Chr.11: 62627059-62627059 on Build GRCh38
rs781236509 Chr.11: 62627069-62627069 on Build GRCh38
rs762160029 Chr.11: 62627313-62627313 on Build GRCh38
rs186826272 Chr.11: 62627271-62627271 on Build GRCh38
rs941587240 Chr.11: 62627235-62627235 on Build GRCh38
rs201217245 Chr.11: 62626953-62626953 on Build GRCh38
rs1010001494 Chr.11: 62627029-62627029 on Build GRCh38
rs760496643 Chr.11: 62626969-62626969 on Build GRCh38
rs1051078758 Chr.11: 62627267-62627267 on Build GRCh38
rs751896515 Chr.11: 62627037-62627037 on Build GRCh38
rs767980518 Chr.11: 62627174-62627174 on Build GRCh38
rs781558089 Chr.11: 62627053-62627053 on Build GRCh38
rs754275909 Chr.11: 62627361-62627361 on Build GRCh38
rs757915417 Chr.11: 62627362-62627362 on Build GRCh38
rs896531060 Chr.11: 62627295-62627295 on Build GRCh38
rs11231166 Chr.11: 62626973-62626973 on Build GRCh38
rs764446223 Chr.11: 62627169-62627169 on Build GRCh38
rs568832916 Chr.11: 62627194-62627194 on Build GRCh38
rs765472949 Chr.11: 62626984-62626984 on Build GRCh38
rs777411181 Chr.11: 62627365-62627365 on Build GRCh38
rs756983818 Chr.11: 62627367-62627367 on Build GRCh38
rs768726456 Chr.11: 62627309-62627309 on Build GRCh38
rs776795590 Chr.11: 62627376-62627378 on Build GRCh38
rs751003975 Chr.11: 62627366-62627366 on Build GRCh38
rs375923197 Chr.11: 62627187-62627187 on Build GRCh38
rs541870201 Chr.11: 62626962-62626963 on Build GRCh38
rs748781912 Chr.11: 62626947-62626947 on Build GRCh38
rs748719281 Chr.11: 62626945-62626945 on Build GRCh38
rs746330393 Chr.11: 62627057-62627057 on Build GRCh38
rs200456484 Chr.11: 62627117-62627117 on Build GRCh38
rs923427983 Chr.11: 62627203-62627203 on Build GRCh38
rs771390500 Chr.11: 62626943-62626943 on Build GRCh38
rs373029944 Chr.11: 62627008-62627008 on Build GRCh38
rs778004655 Chr.11: 62626985-62626986 on Build GRCh38
rs759537157 Chr.11: 62626983-62626983 on Build GRCh38
rs745693187 Chr.11: 62627370-62627370 on Build GRCh38
rs373254816 Chr.11: 62626957-62626957 on Build GRCh38
rs934275463 Chr.11: 62626983-62626984 on Build GRCh38
rs572016346 Chr.11: 62627248-62627248 on Build GRCh38
rs34969464 Chr.11: 62627177-62627178 on Build GRCh38

View Full Product Details