Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62662736-62663243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf98
Entrez Gene ID
102288414
Gene Name
chromosome 11 open reading frame 98
Gene Aliases
C11orf48
UniGene
Hs.9061
Gene Chromosome Location
Chr.11: 62662817-62665216 on Build GRCh38
Transcripts
NM_001286086.1 NR_104414.1 NM_024099.3

Gene Symbol
LBHD1
Entrez Gene ID
79081
Gene Name
LBH domain containing 1
Gene Aliases
C11orf48
UniGene
Hs.9061
Gene Chromosome Location
Chr.11: 62662817-62671769 on Build GRCh38
Transcripts
NM_001286086.1 NR_104414.1 NM_024099.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4508247 c.769C>T p.P257S Chr.11: 62663152-62663152 on Build GRCh38
358555 c.761+1G>T p.? Chr.11: 62663235-62663235 on Build GRCh38
929768 c.769C>A p.P257T Chr.11: 62663152-62663152 on Build GRCh38
3953587 c.762-5C>T p.? Chr.11: 62663164-62663164 on Build GRCh38
6134027 c.792G>C p.*264Y Chr.11: 62663129-62663129 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761736692 Chr.11: 62663222-62663222 on Build GRCh38
rs768263293 Chr.11: 62662818-62662818 on Build GRCh38
rs547669457 Chr.11: 62662895-62662895 on Build GRCh38
rs756324077 Chr.11: 62663139-62663139 on Build GRCh38
rs772519143 Chr.11: 62663078-62663078 on Build GRCh38
rs774029741 Chr.11: 62663207-62663207 on Build GRCh38
rs186673653 Chr.11: 62663093-62663093 on Build GRCh38
rs560413045 Chr.11: 62662881-62662881 on Build GRCh38
rs1025104336 Chr.11: 62662992-62662992 on Build GRCh38
rs987254173 Chr.11: 62663192-62663192 on Build GRCh38
rs597259 Chr.11: 62663187-62663187 on Build GRCh38
rs763951172 Chr.11: 62663103-62663103 on Build GRCh38
rs76217138 Chr.11: 62662780-62662780 on Build GRCh38
rs143302349 Chr.11: 62662843-62662848 on Build GRCh38
rs879363467 Chr.11: 62662823-62662823 on Build GRCh38
rs779942411 Chr.11: 62663142-62663142 on Build GRCh38
rs775269609 Chr.11: 62663098-62663098 on Build GRCh38
rs1047047964 Chr.11: 62663059-62663059 on Build GRCh38
rs866005354 Chr.11: 62662761-62662761 on Build GRCh38
rs781450469 Chr.11: 62663004-62663004 on Build GRCh38
rs985845589 Chr.11: 62662901-62662901 on Build GRCh38
rs201886846 Chr.11: 62663157-62663157 on Build GRCh38
rs774795533 Chr.11: 62663198-62663198 on Build GRCh38
rs748722003 Chr.11: 62663075-62663075 on Build GRCh38
rs773687736 Chr.11: 62663080-62663080 on Build GRCh38
rs750385168 Chr.11: 62663130-62663130 on Build GRCh38
rs767609183 Chr.11: 62663118-62663118 on Build GRCh38
rs971920304 Chr.11: 62662831-62662831 on Build GRCh38
rs892611883 Chr.11: 62662956-62662956 on Build GRCh38
rs7386 Chr.11: 62662863-62662863 on Build GRCh38
rs545071343 Chr.11: 62662829-62662829 on Build GRCh38
rs754143597 Chr.11: 62663087-62663088 on Build GRCh38
rs751270075 Chr.11: 62663109-62663109 on Build GRCh38
rs780394568 Chr.11: 62663014-62663014 on Build GRCh38
rs779084248 Chr.11: 62663057-62663057 on Build GRCh38
rs759563797 Chr.11: 62663006-62663006 on Build GRCh38
rs571353154 Chr.11: 62663151-62663151 on Build GRCh38
rs769081916 Chr.11: 62663048-62663048 on Build GRCh38
rs769359991 Chr.11: 62663190-62663190 on Build GRCh38
rs151001215 Chr.11: 62662828-62662828 on Build GRCh38
rs755148668 Chr.11: 62663094-62663095 on Build GRCh38
rs368100722 Chr.11: 62663085-62663085 on Build GRCh38
rs762453530 Chr.11: 62663099-62663099 on Build GRCh38
rs768140456 Chr.11: 62663206-62663206 on Build GRCh38
rs1053554858 Chr.11: 62662946-62662946 on Build GRCh38
rs773117214 Chr.11: 62663041-62663041 on Build GRCh38
rs184364824 Chr.11: 62663052-62663052 on Build GRCh38
rs761860938 Chr.11: 62663115-62663115 on Build GRCh38
rs762693839 Chr.11: 62663202-62663202 on Build GRCh38
rs376436715 Chr.11: 62663171-62663171 on Build GRCh38
rs888466142 Chr.11: 62663162-62663162 on Build GRCh38
rs756236311 Chr.11: 62663013-62663013 on Build GRCh38
rs565407042 Chr.11: 62662839-62662839 on Build GRCh38
rs562760434 Chr.11: 62663025-62663025 on Build GRCh38
rs192448421 Chr.11: 62662976-62662976 on Build GRCh38
rs940581199 Chr.11: 62662809-62662809 on Build GRCh38
rs545671087 Chr.11: 62662931-62662931 on Build GRCh38
rs779357864 Chr.11: 62663166-62663166 on Build GRCh38
rs772396802 Chr.11: 62663178-62663178 on Build GRCh38
rs878856688 Chr.11: 62662927-62662927 on Build GRCh38
rs760130957 Chr.11: 62662953-62662953 on Build GRCh38
rs541482879 Chr.11: 62662864-62662864 on Build GRCh38
rs895250182 Chr.11: 62662939-62662939 on Build GRCh38
rs773133062 Chr.11: 62663001-62663001 on Build GRCh38
rs374875814 Chr.11: 62662845-62662850 on Build GRCh38
rs191835475 Chr.11: 62663212-62663212 on Build GRCh38
rs549305475 Chr.11: 62662979-62662979 on Build GRCh38

View Full Product Details