Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62665564-62666063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LBHD1
Entrez Gene ID
79081
Gene Name
LBH domain containing 1
Gene Aliases
C11orf48
UniGene
Hs.9061
Gene Chromosome Location
Chr.11: 62662817-62671769 on Build GRCh38
Transcripts
NM_024099.3 NM_001043229.1 XM_005274232.4 NR_004390.1

Gene Symbol
METTL12
Entrez Gene ID
751071
Gene Name
methyltransferase like 12
Gene Aliases
U99HG
UniGene
Hs.640726
Gene Chromosome Location
Chr.11: 62665252-62667451 on Build GRCh38
Transcripts
NM_024099.3 NM_001043229.1 XM_005274232.4 NR_004390.1

Gene Symbol
SNORA57
Entrez Gene ID
692158
Gene Name
small nucleolar RNA, H/ACA box 57
Gene Aliases
U99
UniGene
Hs.687390
Gene Chromosome Location
Chr.11: 62665422-62665570 on Build GRCh38
Transcripts
NM_024099.3 NM_001043229.1 XM_005274232.4 NR_004390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6711576 c.51G>C p.T17T Chr.11: 62665930-62665930 on Build GRCh38
4034845 c.32C>T p.P11L Chr.11: 62665911-62665911 on Build GRCh38
689865 c.17G>T p.R6L Chr.11: 62665896-62665896 on Build GRCh38
112100 c.4_5insC p.A3fs*25 Chr.11: 62665883-62665884 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754080939 Chr.11: 62665606-62665606 on Build GRCh38
rs778460771 Chr.11: 62665641-62665641 on Build GRCh38
rs763963891 Chr.11: 62665606-62665606 on Build GRCh38
rs866507432 Chr.11: 62665672-62665672 on Build GRCh38
rs748978799 Chr.11: 62665926-62665926 on Build GRCh38
rs948715763 Chr.11: 62665796-62665796 on Build GRCh38
rs775059235 Chr.11: 62665847-62665847 on Build GRCh38
rs1010282037 Chr.11: 62665799-62665799 on Build GRCh38
rs752851660 Chr.11: 62665947-62665948 on Build GRCh38
rs534502356 Chr.11: 62666031-62666031 on Build GRCh38
rs574137652 Chr.11: 62665680-62665680 on Build GRCh38
rs985248080 Chr.11: 62665649-62665649 on Build GRCh38
rs909195760 Chr.11: 62666016-62666016 on Build GRCh38
rs530958358 Chr.11: 62665924-62665924 on Build GRCh38
rs929903045 Chr.11: 62666015-62666015 on Build GRCh38
rs918465611 Chr.11: 62665998-62665998 on Build GRCh38
rs765614681 Chr.11: 62665888-62665888 on Build GRCh38
rs965186554 Chr.11: 62665910-62665910 on Build GRCh38
rs562358156 Chr.11: 62665911-62665911 on Build GRCh38
rs1031841302 Chr.11: 62666032-62666032 on Build GRCh38
rs756189579 Chr.11: 62665619-62665619 on Build GRCh38
rs759256836 Chr.11: 62665590-62665590 on Build GRCh38
rs565551567 Chr.11: 62666025-62666025 on Build GRCh38
rs574869674 Chr.11: 62665735-62665735 on Build GRCh38
rs778575685 Chr.11: 62665832-62665833 on Build GRCh38
rs995281368 Chr.11: 62665608-62665608 on Build GRCh38
rs755239414 Chr.11: 62665866-62665867 on Build GRCh38
rs968503086 Chr.11: 62665643-62665643 on Build GRCh38
rs778207562 Chr.11: 62665905-62665905 on Build GRCh38
rs760725613 Chr.11: 62665868-62665868 on Build GRCh38
rs773038049 Chr.11: 62665969-62665969 on Build GRCh38
rs765326358 Chr.11: 62665917-62665919 on Build GRCh38
rs199880114 Chr.11: 62665845-62665845 on Build GRCh38
rs776727574 Chr.11: 62665975-62665975 on Build GRCh38
rs951303285 Chr.11: 62665624-62665624 on Build GRCh38
rs534609307 Chr.11: 62665622-62665622 on Build GRCh38
rs554404678 Chr.11: 62665639-62665639 on Build GRCh38
rs767734152 Chr.11: 62665615-62665615 on Build GRCh38
rs929260139 Chr.11: 62665709-62665710 on Build GRCh38
rs774118809 Chr.11: 62665936-62665936 on Build GRCh38
rs777976706 Chr.11: 62665971-62665995 on Build GRCh38
rs576853975 Chr.11: 62665834-62665834 on Build GRCh38
rs750219059 Chr.11: 62665763-62665763 on Build GRCh38
rs376484943 Chr.11: 62665609-62665609 on Build GRCh38
rs540699308 Chr.11: 62665825-62665825 on Build GRCh38
rs754078168 Chr.11: 62665835-62665835 on Build GRCh38
rs112534450 Chr.11: 62665752-62665752 on Build GRCh38
rs985195641 Chr.11: 62665881-62665881 on Build GRCh38
rs976584632 Chr.11: 62665977-62665977 on Build GRCh38
rs1003009191 Chr.11: 62665794-62665794 on Build GRCh38
rs978527715 Chr.11: 62665724-62665724 on Build GRCh38
rs779186504 Chr.11: 62665841-62665841 on Build GRCh38
rs371004419 Chr.11: 62665995-62665995 on Build GRCh38
rs750520435 Chr.11: 62665618-62665618 on Build GRCh38
rs202151951 Chr.11: 62666016-62666016 on Build GRCh38
rs748024340 Chr.11: 62665939-62665939 on Build GRCh38
rs748912165 Chr.11: 62665851-62665851 on Build GRCh38
rs755588623 Chr.11: 62665917-62665917 on Build GRCh38
rs373978238 Chr.11: 62665974-62665974 on Build GRCh38
rs911041727 Chr.11: 62665676-62665676 on Build GRCh38
rs570771969 Chr.11: 62665889-62665889 on Build GRCh38
rs567910386 Chr.11: 62665977-62666001 on Build GRCh38
rs930698957 Chr.11: 62665837-62665837 on Build GRCh38
rs775175499 Chr.11: 62665602-62665602 on Build GRCh38
rs1044397255 Chr.11: 62665797-62665797 on Build GRCh38
rs185447414 Chr.11: 62665970-62665970 on Build GRCh38
rs181220820 Chr.11: 62665893-62665893 on Build GRCh38
rs546733362 Chr.11: 62666001-62666001 on Build GRCh38
rs376122030 Chr.11: 62665869-62665869 on Build GRCh38
rs777692176 Chr.11: 62665996-62665996 on Build GRCh38
rs965069507 Chr.11: 62665678-62665678 on Build GRCh38
rs543009354 Chr.11: 62665729-62665729 on Build GRCh38
rs769379100 Chr.11: 62665593-62665593 on Build GRCh38
rs774440347 Chr.11: 62665607-62665607 on Build GRCh38
rs371249223 Chr.11: 62665970-62665994 on Build GRCh38
rs758589986 Chr.11: 62665953-62665977 on Build GRCh38
rs199964289 Chr.11: 62665879-62665879 on Build GRCh38
rs774246334 Chr.11: 62665859-62665859 on Build GRCh38
rs755156327 Chr.11: 62665873-62665873 on Build GRCh38
rs899188705 Chr.11: 62665830-62665830 on Build GRCh38
rs759592827 Chr.11: 62665993-62665993 on Build GRCh38
rs760598891 Chr.11: 62665971-62665971 on Build GRCh38
rs372582513 Chr.11: 62665858-62665858 on Build GRCh38
rs767605267 Chr.11: 62665867-62665867 on Build GRCh38
rs1028064462 Chr.11: 62665699-62665699 on Build GRCh38
rs367863343 Chr.11: 62665976-62665976 on Build GRCh38
rs552515929 Chr.11: 62665846-62665846 on Build GRCh38
rs768372431 Chr.11: 62665935-62665935 on Build GRCh38
rs968481749 Chr.11: 62665703-62665703 on Build GRCh38
rs1027971200 Chr.11: 62665927-62665927 on Build GRCh38
rs138999399 Chr.11: 62665842-62665842 on Build GRCh38
rs755354386 Chr.11: 62665840-62665840 on Build GRCh38
rs771887717 Chr.11: 62665955-62665955 on Build GRCh38

View Full Product Details