Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62666404-62666908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LBHD1
Entrez Gene ID
79081
Gene Name
LBH domain containing 1
Gene Aliases
C11orf48
UniGene
Hs.9061
Gene Chromosome Location
Chr.11: 62662817-62671769 on Build GRCh38
Transcripts
NM_024099.3 NM_001043229.1 XM_005274232.4

Gene Symbol
METTL12
Entrez Gene ID
751071
Gene Name
methyltransferase like 12
Gene Aliases
U99HG
UniGene
Hs.640726
Gene Chromosome Location
Chr.11: 62665252-62667451 on Build GRCh38
Transcripts
NM_024099.3 NM_001043229.1 XM_005274232.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1676109 c.571C>T p.Q191* Chr.11: 62666899-62666899 on Build GRCh38
6569976 c.244G>A p.V82M Chr.11: 62666572-62666572 on Build GRCh38
1298330 c.555G>T p.L185F Chr.11: 62666883-62666883 on Build GRCh38
2039387 c.573G>T p.Q191H Chr.11: 62666901-62666901 on Build GRCh38
1746402 c.172G>C p.D58H Chr.11: 62666500-62666500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749819388 Chr.11: 62666754-62666754 on Build GRCh38
rs545639180 Chr.11: 62666466-62666466 on Build GRCh38
rs780945893 Chr.11: 62666755-62666756 on Build GRCh38
rs377091669 Chr.11: 62666614-62666614 on Build GRCh38
rs769584090 Chr.11: 62666779-62666779 on Build GRCh38
rs1005348888 Chr.11: 62666518-62666518 on Build GRCh38
rs753626421 Chr.11: 62666644-62666644 on Build GRCh38
rs371558650 Chr.11: 62666869-62666869 on Build GRCh38
rs372150390 Chr.11: 62666509-62666509 on Build GRCh38
rs758568457 Chr.11: 62666657-62666657 on Build GRCh38
rs750641923 Chr.11: 62666748-62666748 on Build GRCh38
rs374573824 Chr.11: 62666760-62666760 on Build GRCh38
rs370558447 Chr.11: 62666681-62666681 on Build GRCh38
rs753195000 Chr.11: 62666840-62666840 on Build GRCh38
rs72927292 Chr.11: 62666426-62666426 on Build GRCh38
rs772597672 Chr.11: 62666469-62666469 on Build GRCh38
rs746570653 Chr.11: 62666757-62666757 on Build GRCh38
rs905553228 Chr.11: 62666880-62666880 on Build GRCh38
rs368690203 Chr.11: 62666807-62666807 on Build GRCh38
rs533414913 Chr.11: 62666711-62666711 on Build GRCh38
rs747051469 Chr.11: 62666814-62666815 on Build GRCh38
rs776163706 Chr.11: 62666758-62666758 on Build GRCh38
rs774649747 Chr.11: 62666459-62666459 on Build GRCh38
rs778114547 Chr.11: 62666520-62666520 on Build GRCh38
rs751859433 Chr.11: 62666665-62666665 on Build GRCh38
rs756860374 Chr.11: 62666500-62666500 on Build GRCh38
rs758550021 Chr.11: 62666597-62666597 on Build GRCh38
rs748425494 Chr.11: 62666515-62666515 on Build GRCh38
rs201779870 Chr.11: 62666460-62666460 on Build GRCh38
rs960009581 Chr.11: 62666612-62666612 on Build GRCh38
rs913182455 Chr.11: 62666843-62666843 on Build GRCh38
rs749161623 Chr.11: 62666872-62666872 on Build GRCh38
rs749424508 Chr.11: 62666454-62666454 on Build GRCh38
rs201025757 Chr.11: 62666742-62666742 on Build GRCh38
rs368095657 Chr.11: 62666865-62666865 on Build GRCh38
rs550893088 Chr.11: 62666824-62666824 on Build GRCh38
rs757327696 Chr.11: 62666740-62666740 on Build GRCh38
rs11231181 Chr.11: 62666701-62666701 on Build GRCh38
rs202170779 Chr.11: 62666496-62666496 on Build GRCh38
rs755334407 Chr.11: 62666838-62666838 on Build GRCh38
rs751624812 Chr.11: 62666746-62666746 on Build GRCh38
rs759348507 Chr.11: 62666647-62666647 on Build GRCh38
rs935850025 Chr.11: 62666786-62666786 on Build GRCh38
rs781450776 Chr.11: 62666673-62666673 on Build GRCh38
rs201286649 Chr.11: 62666472-62666472 on Build GRCh38
rs760477963 Chr.11: 62666576-62666576 on Build GRCh38
rs766022997 Chr.11: 62666579-62666579 on Build GRCh38
rs865863586 Chr.11: 62666800-62666800 on Build GRCh38
rs188883781 Chr.11: 62666463-62666463 on Build GRCh38
rs567917506 Chr.11: 62666785-62666785 on Build GRCh38
rs766219934 Chr.11: 62666642-62666642 on Build GRCh38
rs768240000 Chr.11: 62666540-62666540 on Build GRCh38
rs754824488 Chr.11: 62666585-62666585 on Build GRCh38
rs756496301 Chr.11: 62666675-62666675 on Build GRCh38
rs763255599 Chr.11: 62666715-62666715 on Build GRCh38
rs759777728 Chr.11: 62666477-62666477 on Build GRCh38
rs775569393 Chr.11: 62666726-62666726 on Build GRCh38
rs746257828 Chr.11: 62666674-62666674 on Build GRCh38
rs746500626 Chr.11: 62666702-62666702 on Build GRCh38
rs201738445 Chr.11: 62666842-62666842 on Build GRCh38
rs771424123 Chr.11: 62666613-62666613 on Build GRCh38
rs757368095 Chr.11: 62666666-62666666 on Build GRCh38
rs763451990 Chr.11: 62666495-62666495 on Build GRCh38
rs137922656 Chr.11: 62666625-62666625 on Build GRCh38
rs556929101 Chr.11: 62666484-62666484 on Build GRCh38
rs544901596 Chr.11: 62666682-62666682 on Build GRCh38
rs111425217 Chr.11: 62666440-62666440 on Build GRCh38
rs200276860 Chr.11: 62666588-62666588 on Build GRCh38
rs118053937 Chr.11: 62666753-62666753 on Build GRCh38
rs761102979 Chr.11: 62666481-62666481 on Build GRCh38
rs761289453 Chr.11: 62666560-62666560 on Build GRCh38
rs753016276 Chr.11: 62666650-62666651 on Build GRCh38
rs199971805 Chr.11: 62666884-62666884 on Build GRCh38
rs773666009 Chr.11: 62666470-62666470 on Build GRCh38
rs771411834 Chr.11: 62666538-62666538 on Build GRCh38
rs747447715 Chr.11: 62666524-62666524 on Build GRCh38
rs764336072 Chr.11: 62666717-62666722 on Build GRCh38
rs375255311 Chr.11: 62666883-62666883 on Build GRCh38
rs769505563 Chr.11: 62666764-62666764 on Build GRCh38
rs547965574 Chr.11: 62666458-62666458 on Build GRCh38
rs752670920 Chr.11: 62666653-62666653 on Build GRCh38
rs759890869 Chr.11: 62666539-62666539 on Build GRCh38
rs541913227 Chr.11: 62666618-62666618 on Build GRCh38
rs747167884 Chr.11: 62666608-62666608 on Build GRCh38
rs530577932 Chr.11: 62666806-62666806 on Build GRCh38
rs759310093 Chr.11: 62666723-62666723 on Build GRCh38
rs750265298 Chr.11: 62666783-62666785 on Build GRCh38
rs976128423 Chr.11: 62666594-62666594 on Build GRCh38
rs752869045 Chr.11: 62666590-62666590 on Build GRCh38
rs375107020 Chr.11: 62666503-62666503 on Build GRCh38
rs772858211 Chr.11: 62666632-62666632 on Build GRCh38
rs778202351 Chr.11: 62666601-62666601 on Build GRCh38
rs932466674 Chr.11: 62666622-62666622 on Build GRCh38
rs1027754065 Chr.11: 62666727-62666727 on Build GRCh38
rs750614380 Chr.11: 62666811-62666811 on Build GRCh38
rs76078023 Chr.11: 62666755-62666755 on Build GRCh38
rs765604886 Chr.11: 62666614-62666614 on Build GRCh38
rs767575262 Chr.11: 62666808-62666808 on Build GRCh38
rs369948826 Chr.11: 62666565-62666565 on Build GRCh38
rs751224800 Chr.11: 62666499-62666499 on Build GRCh38
rs757275785 Chr.11: 62666750-62666757 on Build GRCh38
rs777929810 Chr.11: 62666664-62666664 on Build GRCh38
rs751735040 Chr.11: 62666739-62666739 on Build GRCh38
rs371427770 Chr.11: 62666837-62666837 on Build GRCh38
rs192379593 Chr.11: 62666640-62666640 on Build GRCh38
rs779457794 Chr.11: 62666839-62666839 on Build GRCh38
rs771555436 Chr.11: 62666629-62666629 on Build GRCh38
rs199842069 Chr.11: 62666745-62666745 on Build GRCh38
rs775374841 Chr.11: 62666777-62666777 on Build GRCh38
rs374848905 Chr.11: 62666691-62666691 on Build GRCh38
rs762579166 Chr.11: 62666488-62666488 on Build GRCh38
rs562774549 Chr.11: 62666564-62666564 on Build GRCh38
rs777003067 Chr.11: 62666485-62666485 on Build GRCh38
rs776034294 Chr.11: 62666623-62666623 on Build GRCh38
rs750069893 Chr.11: 62666575-62666575 on Build GRCh38
rs764287011 Chr.11: 62666735-62666735 on Build GRCh38
rs767322545 Chr.11: 62666501-62666501 on Build GRCh38
rs376560159 Chr.11: 62666652-62666652 on Build GRCh38
rs570436114 Chr.11: 62666502-62666502 on Build GRCh38
rs370220110 Chr.11: 62666619-62666619 on Build GRCh38
rs764164185 Chr.11: 62666799-62666799 on Build GRCh38
rs773682549 Chr.11: 62666546-62666546 on Build GRCh38
rs879037551 Chr.11: 62666776-62666776 on Build GRCh38
rs755092361 Chr.11: 62666511-62666511 on Build GRCh38
rs199630913 Chr.11: 62666847-62666847 on Build GRCh38
rs369330577 Chr.11: 62666441-62666441 on Build GRCh38
rs200778480 Chr.11: 62666581-62666581 on Build GRCh38
rs778818724 Chr.11: 62666513-62666513 on Build GRCh38
rs751155890 Chr.11: 62666425-62666425 on Build GRCh38

View Full Product Details