Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62908995-62909490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRM1
Entrez Gene ID
1128
Gene Name
cholinergic receptor muscarinic 1
Gene Aliases
HM1, M1, M1R
UniGene
Hs.632119
Gene Chromosome Location
Chr.11: 62908675-62921861 on Build GRCh38
Transcripts
NM_000738.2 XM_011544742.2

SNP Detail

SNP ID Chromosome Location
rs547306257 Chr.11: 62909283-62909283 on Build GRCh38
rs1019952664 Chr.11: 62909309-62909309 on Build GRCh38
rs988496566 Chr.11: 62909354-62909354 on Build GRCh38
rs920592103 Chr.11: 62909439-62909439 on Build GRCh38
rs1047809931 Chr.11: 62909449-62909449 on Build GRCh38
rs892942288 Chr.11: 62909220-62909220 on Build GRCh38
rs559492833 Chr.11: 62909238-62909238 on Build GRCh38
rs138086507 Chr.11: 62909419-62909419 on Build GRCh38
rs753608313 Chr.11: 62909462-62909462 on Build GRCh38
rs565560808 Chr.11: 62909359-62909359 on Build GRCh38
rs992985717 Chr.11: 62909373-62909373 on Build GRCh38
rs1011467091 Chr.11: 62909221-62909221 on Build GRCh38
rs116161975 Chr.11: 62909343-62909343 on Build GRCh38
rs886486403 Chr.11: 62909451-62909451 on Build GRCh38
rs374302795 Chr.11: 62909158-62909158 on Build GRCh38
rs1044372723 Chr.11: 62909274-62909274 on Build GRCh38
rs560884278 Chr.11: 62909310-62909310 on Build GRCh38
rs574418695 Chr.11: 62909060-62909061 on Build GRCh38
rs1007599214 Chr.11: 62909277-62909277 on Build GRCh38
rs939876253 Chr.11: 62909457-62909457 on Build GRCh38
rs1031880224 Chr.11: 62909363-62909363 on Build GRCh38
rs930710144 Chr.11: 62909282-62909282 on Build GRCh38
rs921033494 Chr.11: 62909135-62909135 on Build GRCh38
rs1004082855 Chr.11: 62909035-62909035 on Build GRCh38
rs552102877 Chr.11: 62909348-62909348 on Build GRCh38
rs577310699 Chr.11: 62909097-62909097 on Build GRCh38
rs762265426 Chr.11: 62909287-62909287 on Build GRCh38
rs147408879 Chr.11: 62909376-62909376 on Build GRCh38
rs527557687 Chr.11: 62909281-62909281 on Build GRCh38
rs1057503299 Chr.11: 62909231-62909231 on Build GRCh38
rs138644279 Chr.11: 62909302-62909302 on Build GRCh38
rs879374813 Chr.11: 62909398-62909398 on Build GRCh38
rs765916967 Chr.11: 62909353-62909353 on Build GRCh38
rs1019310308 Chr.11: 62909070-62909070 on Build GRCh38
rs750908456 Chr.11: 62909331-62909331 on Build GRCh38
rs746730719 Chr.11: 62909356-62909356 on Build GRCh38
rs901067074 Chr.11: 62909312-62909312 on Build GRCh38
rs951322867 Chr.11: 62909024-62909024 on Build GRCh38
rs546141018 Chr.11: 62909214-62909214 on Build GRCh38
rs919391550 Chr.11: 62909044-62909044 on Build GRCh38
rs934942398 Chr.11: 62909226-62909226 on Build GRCh38
rs934369900 Chr.11: 62909213-62909213 on Build GRCh38
rs11823728 Chr.11: 62909330-62909330 on Build GRCh38
rs772133173 Chr.11: 62909384-62909384 on Build GRCh38
rs930762532 Chr.11: 62909445-62909445 on Build GRCh38
rs980532318 Chr.11: 62909431-62909431 on Build GRCh38
rs373825611 Chr.11: 62909098-62909098 on Build GRCh38
rs956193344 Chr.11: 62909368-62909368 on Build GRCh38
rs1052889614 Chr.11: 62909215-62909215 on Build GRCh38
rs764424665 Chr.11: 62909166-62909166 on Build GRCh38
rs916132497 Chr.11: 62909120-62909120 on Build GRCh38
rs751930184 Chr.11: 62909264-62909264 on Build GRCh38
rs142847086 Chr.11: 62909342-62909342 on Build GRCh38
rs965425957 Chr.11: 62909071-62909071 on Build GRCh38
rs917467451 Chr.11: 62909239-62909239 on Build GRCh38
rs140743882 Chr.11: 62909128-62909128 on Build GRCh38

View Full Product Details