Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63635444-63635947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL3
Entrez Gene ID
25923
Gene Name
atlastin GTPase 3
Gene Aliases
HSN1F
UniGene
Hs.356719
Gene Chromosome Location
Chr.11: 63624082-63671974 on Build GRCh38
Transcripts
NM_001290048.1 NM_015459.4 XM_006718493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4664077 c.1012C>A p.P338T Chr.11: 63635557-63635557 on Build GRCh38
139907 c.1025C>T p.S342F Chr.11: 63635544-63635544 on Build GRCh38
1183931 c.989A>C p.K330T Chr.11: 63635580-63635580 on Build GRCh38
6894560 c.1106C>T p.S369F Chr.11: 63635544-63635544 on Build GRCh38
4664078 c.1093C>A p.P365T Chr.11: 63635557-63635557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023662320 Chr.11: 63635882-63635882 on Build GRCh38
rs117830205 Chr.11: 63635813-63635813 on Build GRCh38
rs556252870 Chr.11: 63635472-63635472 on Build GRCh38
rs780994050 Chr.11: 63635875-63635875 on Build GRCh38
rs199945115 Chr.11: 63635524-63635524 on Build GRCh38
rs1050082087 Chr.11: 63635723-63635723 on Build GRCh38
rs767260184 Chr.11: 63635572-63635572 on Build GRCh38
rs187363534 Chr.11: 63635900-63635900 on Build GRCh38
rs754735689 Chr.11: 63635619-63635619 on Build GRCh38
rs34109274 Chr.11: 63635775-63635776 on Build GRCh38
rs190159572 Chr.11: 63635508-63635508 on Build GRCh38
rs528921459 Chr.11: 63635891-63635891 on Build GRCh38
rs975689206 Chr.11: 63635636-63635636 on Build GRCh38
rs1004292829 Chr.11: 63635895-63635895 on Build GRCh38
rs74851651 Chr.11: 63635604-63635604 on Build GRCh38
rs948667364 Chr.11: 63635819-63635819 on Build GRCh38
rs1040722063 Chr.11: 63635824-63635825 on Build GRCh38
rs746679687 Chr.11: 63635635-63635635 on Build GRCh38
rs758542166 Chr.11: 63635630-63635630 on Build GRCh38
rs370486647 Chr.11: 63635482-63635482 on Build GRCh38
rs61928161 Chr.11: 63635646-63635646 on Build GRCh38
rs952313313 Chr.11: 63635576-63635576 on Build GRCh38
rs572184919 Chr.11: 63635569-63635569 on Build GRCh38
rs978374249 Chr.11: 63635670-63635670 on Build GRCh38
rs951423011 Chr.11: 63635517-63635517 on Build GRCh38
rs780875902 Chr.11: 63635652-63635652 on Build GRCh38
rs747149705 Chr.11: 63635520-63635520 on Build GRCh38
rs1026475969 Chr.11: 63635570-63635570 on Build GRCh38
rs928635419 Chr.11: 63635899-63635899 on Build GRCh38
rs558698105 Chr.11: 63635531-63635531 on Build GRCh38
rs970321937 Chr.11: 63635886-63635886 on Build GRCh38
rs934737096 Chr.11: 63635647-63635647 on Build GRCh38
rs368447357 Chr.11: 63635902-63635902 on Build GRCh38
rs766282029 Chr.11: 63635614-63635614 on Build GRCh38
rs748322025 Chr.11: 63635492-63635492 on Build GRCh38
rs769826594 Chr.11: 63635867-63635867 on Build GRCh38
rs752594187 Chr.11: 63635623-63635623 on Build GRCh38
rs754751102 Chr.11: 63635489-63635489 on Build GRCh38
rs777984545 Chr.11: 63635516-63635516 on Build GRCh38
rs529203879 Chr.11: 63635804-63635804 on Build GRCh38
rs896942457 Chr.11: 63635830-63635830 on Build GRCh38
rs753802919 Chr.11: 63635618-63635618 on Build GRCh38
rs1056925343 Chr.11: 63635801-63635801 on Build GRCh38
rs899051027 Chr.11: 63635866-63635866 on Build GRCh38
rs370251222 Chr.11: 63635638-63635638 on Build GRCh38
rs182263771 Chr.11: 63635716-63635716 on Build GRCh38
rs1026950281 Chr.11: 63635877-63635877 on Build GRCh38
rs878931465 Chr.11: 63635634-63635634 on Build GRCh38
rs1027087649 Chr.11: 63635497-63635497 on Build GRCh38
rs772706990 Chr.11: 63635597-63635597 on Build GRCh38
rs764953578 Chr.11: 63635622-63635622 on Build GRCh38
rs373751961 Chr.11: 63635566-63635566 on Build GRCh38
rs1052922142 Chr.11: 63635648-63635648 on Build GRCh38
rs757595908 Chr.11: 63635588-63635589 on Build GRCh38
rs751902951 Chr.11: 63635577-63635579 on Build GRCh38
rs201973274 Chr.11: 63635535-63635535 on Build GRCh38
rs773989806 Chr.11: 63635549-63635549 on Build GRCh38
rs779155610 Chr.11: 63635490-63635490 on Build GRCh38
rs947330096 Chr.11: 63635714-63635714 on Build GRCh38
rs1007988303 Chr.11: 63635479-63635479 on Build GRCh38
rs560871854 Chr.11: 63635737-63635737 on Build GRCh38
rs889480258 Chr.11: 63635728-63635728 on Build GRCh38
rs1026487510 Chr.11: 63635844-63635844 on Build GRCh38
rs914365377 Chr.11: 63635697-63635697 on Build GRCh38
rs924366007 Chr.11: 63635693-63635693 on Build GRCh38
rs1044308719 Chr.11: 63635820-63635820 on Build GRCh38
rs745762711 Chr.11: 63635832-63635832 on Build GRCh38

View Full Product Details