Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63651826-63652326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL3
Entrez Gene ID
25923
Gene Name
atlastin GTPase 3
Gene Aliases
HSN1F
UniGene
Hs.356719
Gene Chromosome Location
Chr.11: 63624082-63671974 on Build GRCh38
Transcripts
NM_001290048.1 NM_015459.4 XM_006718493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6422086 c.530A>G p.N177S Chr.11: 63651967-63651967 on Build GRCh38
6422087 c.611A>G p.N204S Chr.11: 63651967-63651967 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs942754069 Chr.11: 63651909-63651909 on Build GRCh38
rs971907747 Chr.11: 63652079-63652079 on Build GRCh38
rs499942 Chr.11: 63651997-63651997 on Build GRCh38
rs757867402 Chr.11: 63651989-63651989 on Build GRCh38
rs769188221 Chr.11: 63651921-63651921 on Build GRCh38
rs533114101 Chr.11: 63651920-63651920 on Build GRCh38
rs541724894 Chr.11: 63652255-63652255 on Build GRCh38
rs1051083452 Chr.11: 63651859-63651859 on Build GRCh38
rs184718120 Chr.11: 63652063-63652063 on Build GRCh38
rs978258987 Chr.11: 63652064-63652064 on Build GRCh38
rs553867064 Chr.11: 63652070-63652070 on Build GRCh38
rs542730325 Chr.11: 63652150-63652150 on Build GRCh38
rs556701807 Chr.11: 63652182-63652182 on Build GRCh38
rs774742655 Chr.11: 63651922-63651922 on Build GRCh38
rs3758646 Chr.11: 63652234-63652234 on Build GRCh38
rs756300796 Chr.11: 63652029-63652029 on Build GRCh38
rs758193180 Chr.11: 63652003-63652003 on Build GRCh38
rs558373567 Chr.11: 63651981-63651981 on Build GRCh38
rs538455700 Chr.11: 63651950-63651950 on Build GRCh38
rs776366099 Chr.11: 63651940-63651940 on Build GRCh38
rs995783775 Chr.11: 63652260-63652260 on Build GRCh38
rs1013288149 Chr.11: 63652015-63652015 on Build GRCh38
rs923202598 Chr.11: 63652104-63652104 on Build GRCh38
rs756193857 Chr.11: 63652239-63652239 on Build GRCh38
rs942637912 Chr.11: 63652230-63652230 on Build GRCh38
rs765020653 Chr.11: 63651983-63651983 on Build GRCh38
rs911348286 Chr.11: 63651874-63651874 on Build GRCh38
rs940006079 Chr.11: 63652203-63652203 on Build GRCh38
rs576648160 Chr.11: 63652195-63652195 on Build GRCh38
rs545613470 Chr.11: 63652202-63652202 on Build GRCh38
rs893309308 Chr.11: 63651930-63651930 on Build GRCh38
rs573713387 Chr.11: 63652057-63652057 on Build GRCh38
rs954645040 Chr.11: 63652143-63652143 on Build GRCh38
rs780193918 Chr.11: 63652033-63652033 on Build GRCh38
rs768000338 Chr.11: 63651932-63651932 on Build GRCh38
rs995315271 Chr.11: 63651933-63651933 on Build GRCh38
rs774694716 Chr.11: 63651904-63651906 on Build GRCh38
rs962200525 Chr.11: 63652075-63652075 on Build GRCh38
rs752431263 Chr.11: 63652001-63652001 on Build GRCh38
rs1033970885 Chr.11: 63652013-63652013 on Build GRCh38
rs752230117 Chr.11: 63651987-63651987 on Build GRCh38
rs759709098 Chr.11: 63652136-63652136 on Build GRCh38
rs1037095437 Chr.11: 63651897-63651897 on Build GRCh38
rs545160035 Chr.11: 63651992-63651992 on Build GRCh38
rs745732615 Chr.11: 63652016-63652016 on Build GRCh38
rs762198713 Chr.11: 63651931-63651931 on Build GRCh38
rs780949051 Chr.11: 63652007-63652007 on Build GRCh38
rs1005931530 Chr.11: 63652058-63652058 on Build GRCh38
rs372237930 Chr.11: 63652254-63652254 on Build GRCh38
rs143027971 Chr.11: 63652101-63652101 on Build GRCh38

View Full Product Details