Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63763866-63764360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf95
Entrez Gene ID
65998
Gene Name
chromosome 11 open reading frame 95
Gene Aliases
-
UniGene
Hs.191073
Gene Chromosome Location
Chr.11: 63759892-63768641 on Build GRCh38
Transcripts
NM_001144936.1

SNP Detail

SNP ID Chromosome Location
rs913592824 Chr.11: 63764053-63764053 on Build GRCh38
rs754621403 Chr.11: 63763988-63763988 on Build GRCh38
rs769983648 Chr.11: 63764144-63764166 on Build GRCh38
rs990609708 Chr.11: 63763981-63763981 on Build GRCh38
rs978974665 Chr.11: 63764248-63764248 on Build GRCh38
rs747555071 Chr.11: 63764042-63764044 on Build GRCh38
rs982317461 Chr.11: 63763957-63763957 on Build GRCh38
rs890526326 Chr.11: 63763889-63763889 on Build GRCh38
rs998199128 Chr.11: 63763944-63763944 on Build GRCh38
rs746254947 Chr.11: 63764048-63764062 on Build GRCh38
rs549520004 Chr.11: 63764114-63764114 on Build GRCh38
rs1021212037 Chr.11: 63763947-63763947 on Build GRCh38
rs951320782 Chr.11: 63763956-63763956 on Build GRCh38
rs1035304794 Chr.11: 63764195-63764195 on Build GRCh38
rs946377880 Chr.11: 63764220-63764220 on Build GRCh38
rs1007070724 Chr.11: 63764073-63764075 on Build GRCh38
rs909697957 Chr.11: 63763988-63763989 on Build GRCh38
rs988324934 Chr.11: 63764202-63764202 on Build GRCh38
rs745679828 Chr.11: 63764249-63764249 on Build GRCh38
rs545728124 Chr.11: 63763970-63763970 on Build GRCh38
rs928147830 Chr.11: 63763964-63763964 on Build GRCh38
rs771459139 Chr.11: 63764042-63764047 on Build GRCh38
rs563153953 Chr.11: 63764117-63764117 on Build GRCh38
rs892051805 Chr.11: 63763918-63763918 on Build GRCh38
rs901253804 Chr.11: 63764099-63764099 on Build GRCh38
rs775846054 Chr.11: 63764293-63764301 on Build GRCh38
rs984029818 Chr.11: 63763963-63763963 on Build GRCh38
rs1007589813 Chr.11: 63764062-63764062 on Build GRCh38
rs775546125 Chr.11: 63764324-63764324 on Build GRCh38
rs1007620955 Chr.11: 63763907-63763907 on Build GRCh38
rs958063360 Chr.11: 63763928-63763928 on Build GRCh38
rs1039100316 Chr.11: 63764072-63764072 on Build GRCh38
rs190405599 Chr.11: 63764018-63764018 on Build GRCh38
rs951405221 Chr.11: 63764123-63764123 on Build GRCh38
rs1052105260 Chr.11: 63764048-63764048 on Build GRCh38
rs763302363 Chr.11: 63764330-63764330 on Build GRCh38
rs960480880 Chr.11: 63764201-63764201 on Build GRCh38
rs1043352201 Chr.11: 63764056-63764056 on Build GRCh38
rs373286923 Chr.11: 63763909-63763909 on Build GRCh38
rs1057496545 Chr.11: 63763917-63763917 on Build GRCh38
rs939045007 Chr.11: 63763916-63763916 on Build GRCh38
rs909541580 Chr.11: 63764016-63764016 on Build GRCh38
rs1005399374 Chr.11: 63764143-63764143 on Build GRCh38
rs571924031 Chr.11: 63764013-63764013 on Build GRCh38
rs1044813240 Chr.11: 63763893-63763893 on Build GRCh38
rs1010533026 Chr.11: 63763921-63763922 on Build GRCh38
rs1050929438 Chr.11: 63764059-63764059 on Build GRCh38
rs770868392 Chr.11: 63764216-63764216 on Build GRCh38
rs889654374 Chr.11: 63764060-63764060 on Build GRCh38
rs1025629022 Chr.11: 63764120-63764120 on Build GRCh38
rs1006956000 Chr.11: 63763936-63763936 on Build GRCh38
rs552076149 Chr.11: 63764304-63764304 on Build GRCh38
rs1018373045 Chr.11: 63763938-63763939 on Build GRCh38
rs377366978 Chr.11: 63763996-63763996 on Build GRCh38
rs918523894 Chr.11: 63764045-63764045 on Build GRCh38
rs959562251 Chr.11: 63764145-63764145 on Build GRCh38
rs904721152 Chr.11: 63763934-63763934 on Build GRCh38
rs529843477 Chr.11: 63764086-63764086 on Build GRCh38
rs967258879 Chr.11: 63763951-63763951 on Build GRCh38
rs781626817 Chr.11: 63764041-63764042 on Build GRCh38
rs868715678 Chr.11: 63764154-63764154 on Build GRCh38
rs1021598502 Chr.11: 63764101-63764101 on Build GRCh38
rs980634775 Chr.11: 63764155-63764155 on Build GRCh38
rs532240968 Chr.11: 63764146-63764146 on Build GRCh38
rs959094290 Chr.11: 63763978-63763978 on Build GRCh38
rs750760709 Chr.11: 63763911-63763911 on Build GRCh38
rs926195118 Chr.11: 63764264-63764264 on Build GRCh38
rs1010928537 Chr.11: 63764085-63764085 on Build GRCh38
rs1037332886 Chr.11: 63764026-63764026 on Build GRCh38
rs913550628 Chr.11: 63764207-63764207 on Build GRCh38
rs560809792 Chr.11: 63764054-63764054 on Build GRCh38
rs976803702 Chr.11: 63764166-63764166 on Build GRCh38
rs564843259 Chr.11: 63763986-63763986 on Build GRCh38
rs966975503 Chr.11: 63764103-63764103 on Build GRCh38
rs1016821177 Chr.11: 63764144-63764144 on Build GRCh38
rs965723681 Chr.11: 63763938-63763938 on Build GRCh38
rs1035185086 Chr.11: 63764130-63764130 on Build GRCh38
rs563538561 Chr.11: 63764058-63764060 on Build GRCh38
rs1011538756 Chr.11: 63763929-63763929 on Build GRCh38
rs1039863734 Chr.11: 63764292-63764293 on Build GRCh38
rs865977403 Chr.11: 63764291-63764291 on Build GRCh38
rs776339558 Chr.11: 63764219-63764219 on Build GRCh38
rs893766550 Chr.11: 63764083-63764083 on Build GRCh38

View Full Product Details