Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64118474-64118971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLRT1
Entrez Gene ID
23769
Gene Name
fibronectin leucine rich transmembrane protein 1
Gene Aliases
SPG68
UniGene
Hs.584876
Gene Chromosome Location
Chr.11: 64035970-64119183 on Build GRCh38
Transcripts
NM_013280.4 XM_005273861.1 XM_011544883.1 NM_014067.3 XM_005273939.4 XM_006718521.3 XM_006718522.2 XM_011544970.2

Gene Symbol
MACROD1
Entrez Gene ID
28992
Gene Name
MACRO domain containing 1
Gene Aliases
LRP16
UniGene
Hs.602898
Gene Chromosome Location
Chr.11: 63998554-64166652 on Build GRCh38
Transcripts
NM_013280.4 XM_005273861.1 XM_011544883.1 NM_014067.3 XM_005273939.4 XM_006718521.3 XM_006718522.2 XM_011544970.2

SNP Detail

SNP ID Chromosome Location
rs761392849 Chr.11: 64118924-64118924 on Build GRCh38
rs915558999 Chr.11: 64118638-64118638 on Build GRCh38
rs993356233 Chr.11: 64118904-64118904 on Build GRCh38
rs777159936 Chr.11: 64118755-64118755 on Build GRCh38
rs962114872 Chr.11: 64118716-64118716 on Build GRCh38
rs766420782 Chr.11: 64118660-64118660 on Build GRCh38
rs398075989 Chr.11: 64118938-64118938 on Build GRCh38
rs1023239539 Chr.11: 64118499-64118499 on Build GRCh38
rs939509788 Chr.11: 64118687-64118687 on Build GRCh38
rs773385188 Chr.11: 64118733-64118733 on Build GRCh38
rs973765341 Chr.11: 64118507-64118507 on Build GRCh38
rs973188009 Chr.11: 64118729-64118729 on Build GRCh38
rs969716623 Chr.11: 64118940-64118940 on Build GRCh38
rs564377790 Chr.11: 64118606-64118606 on Build GRCh38
rs969055393 Chr.11: 64118506-64118506 on Build GRCh38
rs760797014 Chr.11: 64118571-64118571 on Build GRCh38
rs202054903 Chr.11: 64118913-64118913 on Build GRCh38
rs370302486 Chr.11: 64118937-64118937 on Build GRCh38
rs398016367 Chr.11: 64118606-64118606 on Build GRCh38
rs539777849 Chr.11: 64118945-64118945 on Build GRCh38
rs866670323 Chr.11: 64118692-64118692 on Build GRCh38
rs371030473 Chr.11: 64118936-64118936 on Build GRCh38
rs377157389 Chr.11: 64118923-64118923 on Build GRCh38
rs879165333 Chr.11: 64118595-64118596 on Build GRCh38
rs1007417839 Chr.11: 64118563-64118563 on Build GRCh38
rs984489335 Chr.11: 64118624-64118624 on Build GRCh38
rs577833595 Chr.11: 64118658-64118658 on Build GRCh38
rs984033497 Chr.11: 64118561-64118561 on Build GRCh38
rs1040270213 Chr.11: 64118876-64118876 on Build GRCh38
rs878892115 Chr.11: 64118596-64118596 on Build GRCh38
rs965847677 Chr.11: 64118598-64118598 on Build GRCh38
rs759381240 Chr.11: 64118752-64118752 on Build GRCh38
rs868702912 Chr.11: 64118546-64118546 on Build GRCh38
rs901733670 Chr.11: 64118886-64118886 on Build GRCh38
rs112755142 Chr.11: 64118758-64118758 on Build GRCh38
rs556668400 Chr.11: 64118677-64118677 on Build GRCh38
rs1018767533 Chr.11: 64118570-64118570 on Build GRCh38
rs1053005201 Chr.11: 64118697-64118699 on Build GRCh38
rs951855851 Chr.11: 64118606-64118607 on Build GRCh38
rs931981027 Chr.11: 64118757-64118757 on Build GRCh38
rs1044095455 Chr.11: 64118501-64118501 on Build GRCh38
rs921914098 Chr.11: 64118746-64118746 on Build GRCh38
rs908069417 Chr.11: 64118704-64118704 on Build GRCh38
rs144040219 Chr.11: 64118732-64118732 on Build GRCh38
rs10570373 Chr.11: 64118595-64118595 on Build GRCh38
rs528466299 Chr.11: 64118803-64118803 on Build GRCh38
rs969665659 Chr.11: 64118655-64118655 on Build GRCh38
rs368370823 Chr.11: 64118799-64118799 on Build GRCh38
rs1026070531 Chr.11: 64118605-64118605 on Build GRCh38
rs5792312 Chr.11: 64118928-64118928 on Build GRCh38
rs1043795670 Chr.11: 64118794-64118794 on Build GRCh38
rs63644561 Chr.11: 64118939-64118939 on Build GRCh38
rs548315776 Chr.11: 64118869-64118869 on Build GRCh38
rs952206772 Chr.11: 64118553-64118553 on Build GRCh38

View Full Product Details