Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64207268-64207765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT3
Entrez Gene ID
83706
Gene Name
fermitin family member 3
Gene Aliases
KIND3, MIG-2, MIG2B, UNC112C, URP2, URP2SF
UniGene
Hs.180535
Gene Chromosome Location
Chr.11: 64206665-64223891 on Build GRCh38
Transcripts
NM_031471.5 NM_178443.2 XM_011545294.2 XM_011545295.2 XM_017018398.1 XM_017018399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6663182 c.36C>T p.I12I Chr.11: 64207400-64207400 on Build GRCh38
5816069 c.56G>T p.R19L Chr.11: 64207420-64207420 on Build GRCh38
6350196 c.47G>A p.W16* Chr.11: 64207411-64207411 on Build GRCh38
1188279 c.59T>G p.V20G Chr.11: 64207423-64207423 on Build GRCh38
2162980 c.6G>A p.A2A Chr.11: 64207370-64207370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948838429 Chr.11: 64207467-64207468 on Build GRCh38
rs910823177 Chr.11: 64207710-64207710 on Build GRCh38
rs375595381 Chr.11: 64207401-64207401 on Build GRCh38
rs892554921 Chr.11: 64207524-64207524 on Build GRCh38
rs375241843 Chr.11: 64207510-64207510 on Build GRCh38
rs889203319 Chr.11: 64207300-64207300 on Build GRCh38
rs551546723 Chr.11: 64207369-64207369 on Build GRCh38
rs759870657 Chr.11: 64207350-64207350 on Build GRCh38
rs879119442 Chr.11: 64207311-64207311 on Build GRCh38
rs536057240 Chr.11: 64207678-64207678 on Build GRCh38
rs759179590 Chr.11: 64207498-64207498 on Build GRCh38
rs142815441 Chr.11: 64207523-64207523 on Build GRCh38
rs768712216 Chr.11: 64207567-64207567 on Build GRCh38
rs563036460 Chr.11: 64207575-64207575 on Build GRCh38
rs766140675 Chr.11: 64207496-64207496 on Build GRCh38
rs753418361 Chr.11: 64207457-64207457 on Build GRCh38
rs753093402 Chr.11: 64207366-64207366 on Build GRCh38
rs201247015 Chr.11: 64207341-64207341 on Build GRCh38
rs147311995 Chr.11: 64207451-64207451 on Build GRCh38
rs747046546 Chr.11: 64207419-64207419 on Build GRCh38
rs369873469 Chr.11: 64207562-64207562 on Build GRCh38
rs368140133 Chr.11: 64207538-64207538 on Build GRCh38
rs923459714 Chr.11: 64207692-64207692 on Build GRCh38
rs547477693 Chr.11: 64207634-64207634 on Build GRCh38
rs770220477 Chr.11: 64207484-64207484 on Build GRCh38
rs144256756 Chr.11: 64207450-64207450 on Build GRCh38
rs200960603 Chr.11: 64207488-64207488 on Build GRCh38
rs200509898 Chr.11: 64207389-64207389 on Build GRCh38
rs146533229 Chr.11: 64207615-64207615 on Build GRCh38
rs745962281 Chr.11: 64207483-64207483 on Build GRCh38
rs1033292454 Chr.11: 64207576-64207576 on Build GRCh38
rs71468660 Chr.11: 64207692-64207693 on Build GRCh38
rs774681649 Chr.11: 64207331-64207331 on Build GRCh38
rs764571646 Chr.11: 64207491-64207491 on Build GRCh38
rs773145008 Chr.11: 64207341-64207342 on Build GRCh38
rs759997761 Chr.11: 64207426-64207426 on Build GRCh38
rs749388890 Chr.11: 64207527-64207527 on Build GRCh38
rs149000560 Chr.11: 64207494-64207494 on Build GRCh38
rs369062120 Chr.11: 64207508-64207508 on Build GRCh38
rs1001382865 Chr.11: 64207301-64207301 on Build GRCh38
rs986353899 Chr.11: 64207705-64207705 on Build GRCh38
rs144961137 Chr.11: 64207469-64207469 on Build GRCh38
rs772711731 Chr.11: 64207493-64207493 on Build GRCh38
rs776483974 Chr.11: 64207570-64207570 on Build GRCh38
rs182893243 Chr.11: 64207595-64207595 on Build GRCh38
rs890149276 Chr.11: 64207721-64207721 on Build GRCh38
rs375314540 Chr.11: 64207554-64207554 on Build GRCh38
rs371003332 Chr.11: 64207400-64207400 on Build GRCh38
rs772165226 Chr.11: 64207718-64207718 on Build GRCh38
rs1007694822 Chr.11: 64207736-64207736 on Build GRCh38
rs879171486 Chr.11: 64207621-64207621 on Build GRCh38
rs560752986 Chr.11: 64207555-64207555 on Build GRCh38
rs1017427707 Chr.11: 64207535-64207535 on Build GRCh38
rs920303282 Chr.11: 64207374-64207374 on Build GRCh38
rs749840857 Chr.11: 64207382-64207382 on Build GRCh38
rs765757409 Chr.11: 64207437-64207437 on Build GRCh38
rs71468661 Chr.11: 64207696-64207696 on Build GRCh38
rs368108733 Chr.11: 64207342-64207342 on Build GRCh38
rs751379709 Chr.11: 64207520-64207520 on Build GRCh38
rs765072715 Chr.11: 64207502-64207502 on Build GRCh38
rs771554159 Chr.11: 64207566-64207566 on Build GRCh38
rs933547121 Chr.11: 64207696-64207696 on Build GRCh38
rs145854519 Chr.11: 64207490-64207490 on Build GRCh38
rs543307968 Chr.11: 64207573-64207573 on Build GRCh38
rs1005760705 Chr.11: 64207320-64207320 on Build GRCh38
rs773550225 Chr.11: 64207417-64207417 on Build GRCh38
rs772115690 Chr.11: 64207416-64207416 on Build GRCh38
rs139520045 Chr.11: 64207439-64207439 on Build GRCh38
rs527398808 Chr.11: 64207632-64207632 on Build GRCh38
rs121918296 Chr.11: 64207412-64207412 on Build GRCh38
rs931727572 Chr.11: 64207423-64207423 on Build GRCh38
rs771929762 Chr.11: 64207327-64207328 on Build GRCh38
rs368232843 Chr.11: 64207475-64207475 on Build GRCh38
rs936699931 Chr.11: 64207509-64207509 on Build GRCh38
rs769924203 Chr.11: 64207542-64207542 on Build GRCh38
rs150500299 Chr.11: 64207420-64207420 on Build GRCh38
rs1053798800 Chr.11: 64207712-64207712 on Build GRCh38
rs575642594 Chr.11: 64207503-64207503 on Build GRCh38
rs147987989 Chr.11: 64207391-64207391 on Build GRCh38
rs371970022 Chr.11: 64207635-64207635 on Build GRCh38
rs754398780 Chr.11: 64207343-64207343 on Build GRCh38
rs1037202592 Chr.11: 64207659-64207659 on Build GRCh38
rs956717316 Chr.11: 64207340-64207340 on Build GRCh38
rs767213325 Chr.11: 64207370-64207370 on Build GRCh38
rs141640835 Chr.11: 64207392-64207392 on Build GRCh38
rs772357755 Chr.11: 64207335-64207335 on Build GRCh38
rs1032163277 Chr.11: 64207328-64207328 on Build GRCh38
rs777090294 Chr.11: 64207422-64207422 on Build GRCh38
rs1014367318 Chr.11: 64207316-64207327 on Build GRCh38
rs754901103 Chr.11: 64207460-64207460 on Build GRCh38
rs188350808 Chr.11: 64207596-64207596 on Build GRCh38
rs1009780479 Chr.11: 64207541-64207541 on Build GRCh38
rs1015858275 Chr.11: 64207333-64207333 on Build GRCh38
rs747984187 Chr.11: 64207409-64207409 on Build GRCh38
rs972225595 Chr.11: 64207655-64207655 on Build GRCh38
rs554032088 Chr.11: 64207448-64207448 on Build GRCh38
rs948790986 Chr.11: 64207585-64207585 on Build GRCh38
rs980396974 Chr.11: 64207387-64207387 on Build GRCh38
rs140913422 Chr.11: 64207468-64207468 on Build GRCh38
rs776644484 Chr.11: 64207482-64207482 on Build GRCh38
rs182351045 Chr.11: 64207480-64207480 on Build GRCh38
rs780406517 Chr.11: 64207551-64207551 on Build GRCh38
rs577691940 Chr.11: 64207534-64207534 on Build GRCh38
rs778995279 Chr.11: 64207462-64207462 on Build GRCh38
rs369998418 Chr.11: 64207316-64207316 on Build GRCh38
rs529425713 Chr.11: 64207571-64207571 on Build GRCh38
rs750152628 Chr.11: 64207463-64207463 on Build GRCh38
rs748355995 Chr.11: 64207711-64207711 on Build GRCh38
rs945502164 Chr.11: 64207720-64207720 on Build GRCh38
rs376680568 Chr.11: 64207358-64207358 on Build GRCh38
rs778799174 Chr.11: 64207405-64207405 on Build GRCh38
rs369784905 Chr.11: 64207385-64207385 on Build GRCh38
rs765945446 Chr.11: 64207456-64207456 on Build GRCh38

View Full Product Details