Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64303618-64304115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK4-TEX40
Entrez Gene ID
106780802
Gene Name
KCNK4-TEX40 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 64291722-64304769 on Build GRCh38
Transcripts
NR_133662.1 NM_001039496.1

Gene Symbol
TEX40
Entrez Gene ID
25858
Gene Name
testis expressed 40
Gene Aliases
C11orf20
UniGene
Hs.97174
Gene Chromosome Location
Chr.11: 64300391-64304767 on Build GRCh38
Transcripts
NR_133662.1 NM_001039496.1

SNP Detail

SNP ID Chromosome Location
rs1005556027 Chr.11: 64304079-64304079 on Build GRCh38
rs375128394 Chr.11: 64303863-64303863 on Build GRCh38
rs763901112 Chr.11: 64303769-64303769 on Build GRCh38
rs769179250 Chr.11: 64304005-64304005 on Build GRCh38
rs953886839 Chr.11: 64303910-64303910 on Build GRCh38
rs753819880 Chr.11: 64303824-64303824 on Build GRCh38
rs979827941 Chr.11: 64303964-64303964 on Build GRCh38
rs908693986 Chr.11: 64303755-64303755 on Build GRCh38
rs758831368 Chr.11: 64303838-64303838 on Build GRCh38
rs3016144 Chr.11: 64303749-64303749 on Build GRCh38
rs893783268 Chr.11: 64303650-64303650 on Build GRCh38
rs951800175 Chr.11: 64303976-64303976 on Build GRCh38
rs188230000 Chr.11: 64304010-64304010 on Build GRCh38
rs111486531 Chr.11: 64303837-64303837 on Build GRCh38
rs908723679 Chr.11: 64304015-64304015 on Build GRCh38
rs117629442 Chr.11: 64303800-64303800 on Build GRCh38
rs970139322 Chr.11: 64303944-64303944 on Build GRCh38
rs983186721 Chr.11: 64303992-64303992 on Build GRCh38
rs932629698 Chr.11: 64303819-64303819 on Build GRCh38
rs902630772 Chr.11: 64303676-64303676 on Build GRCh38
rs542275914 Chr.11: 64303832-64303832 on Build GRCh38
rs577353961 Chr.11: 64303729-64303729 on Build GRCh38
rs1024188481 Chr.11: 64303926-64303926 on Build GRCh38
rs564431474 Chr.11: 64303979-64303979 on Build GRCh38
rs368607200 Chr.11: 64303850-64303850 on Build GRCh38
rs932887952 Chr.11: 64304052-64304052 on Build GRCh38
rs1049777712 Chr.11: 64304069-64304069 on Build GRCh38
rs774809731 Chr.11: 64303760-64303760 on Build GRCh38
rs953146677 Chr.11: 64303670-64303670 on Build GRCh38
rs537785303 Chr.11: 64303661-64303661 on Build GRCh38
rs370841141 Chr.11: 64303724-64303724 on Build GRCh38
rs930374370 Chr.11: 64303738-64303738 on Build GRCh38
rs986321655 Chr.11: 64303678-64303678 on Build GRCh38
rs761579740 Chr.11: 64303780-64303780 on Build GRCh38
rs977410411 Chr.11: 64304044-64304044 on Build GRCh38
rs769578342 Chr.11: 64303751-64303751 on Build GRCh38
rs748432428 Chr.11: 64303731-64303731 on Build GRCh38
rs544551400 Chr.11: 64303941-64303941 on Build GRCh38
rs573575828 Chr.11: 64303913-64303913 on Build GRCh38
rs939413907 Chr.11: 64303981-64303981 on Build GRCh38
rs375612985 Chr.11: 64303746-64303746 on Build GRCh38
rs1015222440 Chr.11: 64304083-64304083 on Build GRCh38
rs940180221 Chr.11: 64304025-64304025 on Build GRCh38
rs749399787 Chr.11: 64303723-64303723 on Build GRCh38
rs1027322344 Chr.11: 64303973-64303973 on Build GRCh38
rs371416872 Chr.11: 64303820-64303820 on Build GRCh38
rs202192649 Chr.11: 64303798-64303798 on Build GRCh38
rs748722693 Chr.11: 64303866-64303866 on Build GRCh38
rs985760939 Chr.11: 64303912-64303912 on Build GRCh38
rs762548736 Chr.11: 64303761-64303761 on Build GRCh38
rs372378268 Chr.11: 64303858-64303858 on Build GRCh38
rs779266935 Chr.11: 64303836-64303836 on Build GRCh38
rs772616151 Chr.11: 64303740-64303740 on Build GRCh38
rs367587832 Chr.11: 64303818-64303818 on Build GRCh38
rs745584569 Chr.11: 64303747-64303747 on Build GRCh38
rs1003590279 Chr.11: 64303733-64303733 on Build GRCh38
rs868444493 Chr.11: 64303802-64303802 on Build GRCh38
rs891615486 Chr.11: 64303873-64303873 on Build GRCh38
rs922844414 Chr.11: 64304046-64304046 on Build GRCh38
rs566102497 Chr.11: 64304073-64304073 on Build GRCh38
rs1042862508 Chr.11: 64303675-64303675 on Build GRCh38
rs558059948 Chr.11: 64303673-64303673 on Build GRCh38
rs895306504 Chr.11: 64304026-64304026 on Build GRCh38
rs533406934 Chr.11: 64303987-64303987 on Build GRCh38
rs1027780314 Chr.11: 64303660-64303660 on Build GRCh38
rs1018424280 Chr.11: 64303686-64303686 on Build GRCh38
rs377684493 Chr.11: 64303770-64303770 on Build GRCh38
rs1042355955 Chr.11: 64303950-64303950 on Build GRCh38
rs781038205 Chr.11: 64303856-64303856 on Build GRCh38
rs374082033 Chr.11: 64303809-64303809 on Build GRCh38
rs759361472 Chr.11: 64303919-64303919 on Build GRCh38

View Full Product Details