Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64304453-64304947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK4-TEX40
Entrez Gene ID
106780802
Gene Name
KCNK4-TEX40 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 64291722-64304769 on Build GRCh38
Transcripts
NR_133662.1 NM_001039496.1

Gene Symbol
TEX40
Entrez Gene ID
25858
Gene Name
testis expressed 40
Gene Aliases
C11orf20
UniGene
Hs.97174
Gene Chromosome Location
Chr.11: 64300391-64304767 on Build GRCh38
Transcripts
NR_133662.1 NM_001039496.1

SNP Detail

SNP ID Chromosome Location
rs988814019 Chr.11: 64304802-64304802 on Build GRCh38
rs768268486 Chr.11: 64304511-64304511 on Build GRCh38
rs3217060 Chr.11: 64304830-64304831 on Build GRCh38
rs754757530 Chr.11: 64304532-64304534 on Build GRCh38
rs779555813 Chr.11: 64304601-64304601 on Build GRCh38
rs572282220 Chr.11: 64304923-64304923 on Build GRCh38
rs926166703 Chr.11: 64304761-64304761 on Build GRCh38
rs772010947 Chr.11: 64304530-64304530 on Build GRCh38
rs887445352 Chr.11: 64304588-64304588 on Build GRCh38
rs752830093 Chr.11: 64304576-64304576 on Build GRCh38
rs940584459 Chr.11: 64304896-64304896 on Build GRCh38
rs916589693 Chr.11: 64304753-64304754 on Build GRCh38
rs1022105342 Chr.11: 64304616-64304616 on Build GRCh38
rs948214239 Chr.11: 64304504-64304504 on Build GRCh38
rs553420226 Chr.11: 64304540-64304540 on Build GRCh38
rs771957826 Chr.11: 64304672-64304672 on Build GRCh38
rs774146866 Chr.11: 64304528-64304528 on Build GRCh38
rs925966255 Chr.11: 64304483-64304483 on Build GRCh38
rs927167789 Chr.11: 64304831-64304831 on Build GRCh38
rs866155782 Chr.11: 64304883-64304884 on Build GRCh38
rs749034466 Chr.11: 64304619-64304619 on Build GRCh38
rs371993262 Chr.11: 64304560-64304560 on Build GRCh38
rs758921326 Chr.11: 64304580-64304580 on Build GRCh38
rs766477307 Chr.11: 64304556-64304556 on Build GRCh38
rs887948109 Chr.11: 64304514-64304514 on Build GRCh38
rs182296875 Chr.11: 64304879-64304879 on Build GRCh38
rs544413071 Chr.11: 64304719-64304719 on Build GRCh38
rs940876963 Chr.11: 64304803-64304803 on Build GRCh38
rs1036051727 Chr.11: 64304910-64304910 on Build GRCh38
rs760492297 Chr.11: 64304541-64304541 on Build GRCh38
rs200513166 Chr.11: 64304584-64304584 on Build GRCh38
rs747302993 Chr.11: 64304575-64304578 on Build GRCh38
rs950192092 Chr.11: 64304813-64304813 on Build GRCh38
rs375321956 Chr.11: 64304568-64304568 on Build GRCh38
rs1022493089 Chr.11: 64304917-64304917 on Build GRCh38
rs778449994 Chr.11: 64304582-64304582 on Build GRCh38
rs192327290 Chr.11: 64304726-64304726 on Build GRCh38
rs762296238 Chr.11: 64304604-64304604 on Build GRCh38
rs933566759 Chr.11: 64304859-64304859 on Build GRCh38
rs768193699 Chr.11: 64304621-64304621 on Build GRCh38
rs867277514 Chr.11: 64304561-64304561 on Build GRCh38
rs542083860 Chr.11: 64304715-64304715 on Build GRCh38
rs1006613072 Chr.11: 64304904-64304904 on Build GRCh38
rs369708415 Chr.11: 64304538-64304538 on Build GRCh38
rs573178727 Chr.11: 64304559-64304559 on Build GRCh38
rs765053386 Chr.11: 64304618-64304623 on Build GRCh38
rs2286613 Chr.11: 64304772-64304772 on Build GRCh38
rs747679722 Chr.11: 64304629-64304629 on Build GRCh38
rs751935028 Chr.11: 64304583-64304583 on Build GRCh38
rs913208137 Chr.11: 64304808-64304808 on Build GRCh38
rs1004516850 Chr.11: 64304602-64304602 on Build GRCh38
rs773565627 Chr.11: 64304525-64304525 on Build GRCh38
rs889472952 Chr.11: 64304903-64304903 on Build GRCh38
rs954324002 Chr.11: 64304701-64304701 on Build GRCh38
rs887728238 Chr.11: 64304883-64304883 on Build GRCh38
rs761669359 Chr.11: 64304529-64304529 on Build GRCh38
rs41294371 Chr.11: 64304884-64304885 on Build GRCh38
rs1046203715 Chr.11: 64304816-64304816 on Build GRCh38
rs373270751 Chr.11: 64304696-64304696 on Build GRCh38
rs778333684 Chr.11: 64304624-64304624 on Build GRCh38
rs1007053694 Chr.11: 64304613-64304613 on Build GRCh38
rs948126505 Chr.11: 64304763-64304763 on Build GRCh38
rs772943039 Chr.11: 64304531-64304531 on Build GRCh38
rs937118172 Chr.11: 64304850-64304850 on Build GRCh38
rs760711554 Chr.11: 64304618-64304618 on Build GRCh38
rs905680916 Chr.11: 64304575-64304575 on Build GRCh38
rs773172143 Chr.11: 64304681-64304681 on Build GRCh38

View Full Product Details