Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64305525-64306043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRRA
Entrez Gene ID
2101
Gene Name
estrogen related receptor alpha
Gene Aliases
ERR1, ERRa, ERRalpha, ESRL1, NR3B1
UniGene
Hs.110849
Gene Chromosome Location
Chr.11: 64305528-64316743 on Build GRCh38
Transcripts
NM_001282451.1 NM_004451.4

SNP Detail

SNP ID Chromosome Location
rs371705706 Chr.11: 64305979-64305979 on Build GRCh38
rs575697371 Chr.11: 64305941-64305941 on Build GRCh38
rs113980709 Chr.11: 64305685-64305685 on Build GRCh38
rs41294378 Chr.11: 64305865-64305865 on Build GRCh38
rs917584059 Chr.11: 64305847-64305847 on Build GRCh38
rs1031949466 Chr.11: 64305781-64305781 on Build GRCh38
rs904673409 Chr.11: 64306017-64306017 on Build GRCh38
rs897811404 Chr.11: 64305665-64305665 on Build GRCh38
rs897780359 Chr.11: 64305869-64305869 on Build GRCh38
rs1007647523 Chr.11: 64305732-64305732 on Build GRCh38
rs926435308 Chr.11: 64305855-64305855 on Build GRCh38
rs557943953 Chr.11: 64305999-64305999 on Build GRCh38
rs1037652823 Chr.11: 64305663-64305664 on Build GRCh38
rs987731308 Chr.11: 64305801-64305801 on Build GRCh38
rs929215925 Chr.11: 64305877-64305877 on Build GRCh38
rs1045260207 Chr.11: 64305610-64305610 on Build GRCh38
rs935029958 Chr.11: 64305581-64305581 on Build GRCh38
rs949114817 Chr.11: 64305852-64305852 on Build GRCh38
rs1017720295 Chr.11: 64305927-64305927 on Build GRCh38
rs776716277 Chr.11: 64305623-64305623 on Build GRCh38
rs890512006 Chr.11: 64305731-64305731 on Build GRCh38
rs890480791 Chr.11: 64305921-64305921 on Build GRCh38
rs555435448 Chr.11: 64305889-64305889 on Build GRCh38
rs948160584 Chr.11: 64305840-64305840 on Build GRCh38
rs993537024 Chr.11: 64305668-64305668 on Build GRCh38
rs956363426 Chr.11: 64305799-64305799 on Build GRCh38
rs1030413087 Chr.11: 64305687-64305687 on Build GRCh38
rs577799651 Chr.11: 64306001-64306001 on Build GRCh38
rs376330846 Chr.11: 64305853-64305853 on Build GRCh38
rs941896497 Chr.11: 64305858-64305858 on Build GRCh38
rs979412472 Chr.11: 64305776-64305776 on Build GRCh38
rs949145075 Chr.11: 64305597-64305597 on Build GRCh38
rs1008030042 Chr.11: 64305926-64305926 on Build GRCh38
rs924914621 Chr.11: 64305551-64305551 on Build GRCh38
rs917697236 Chr.11: 64305594-64305594 on Build GRCh38
rs968929447 Chr.11: 64305756-64305756 on Build GRCh38
rs1018167245 Chr.11: 64305754-64305754 on Build GRCh38
rs566802559 Chr.11: 64305631-64305631 on Build GRCh38
rs554089174 Chr.11: 64305586-64305586 on Build GRCh38
rs41294376 Chr.11: 64305854-64305854 on Build GRCh38

View Full Product Details