Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64307085-64307573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRRA
Entrez Gene ID
2101
Gene Name
estrogen related receptor alpha
Gene Aliases
ERR1, ERRa, ERRalpha, ESRL1, NR3B1
UniGene
Hs.110849
Gene Chromosome Location
Chr.11: 64305528-64316743 on Build GRCh38
Transcripts
NM_001282450.1 NM_001282451.1 NM_004451.4 XM_017017313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7366655 c.321C>G p.I107M Chr.11: 64307500-64307500 on Build GRCh38
3869839 c.114C>T p.A38A Chr.11: 64307293-64307293 on Build GRCh38
5617874 c.77C>A p.S26Y Chr.11: 64307256-64307256 on Build GRCh38
2163412 c.284C>A p.S95Y Chr.11: 64307463-64307463 on Build GRCh38
1191886 c.56G>A p.S19N Chr.11: 64307235-64307235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779933190 Chr.11: 64307366-64307366 on Build GRCh38
rs768035615 Chr.11: 64307312-64307312 on Build GRCh38
rs373721261 Chr.11: 64307441-64307441 on Build GRCh38
rs773620669 Chr.11: 64307300-64307300 on Build GRCh38
rs747209844 Chr.11: 64307407-64307407 on Build GRCh38
rs374574706 Chr.11: 64307259-64307259 on Build GRCh38
rs763774214 Chr.11: 64307440-64307440 on Build GRCh38
rs778566959 Chr.11: 64307170-64307170 on Build GRCh38
rs774437764 Chr.11: 64307308-64307308 on Build GRCh38
rs772101514 Chr.11: 64307393-64307393 on Build GRCh38
rs944405327 Chr.11: 64307530-64307530 on Build GRCh38
rs981951323 Chr.11: 64307124-64307124 on Build GRCh38
rs771441085 Chr.11: 64307425-64307425 on Build GRCh38
rs766797384 Chr.11: 64307322-64307322 on Build GRCh38
rs866624106 Chr.11: 64307459-64307459 on Build GRCh38
rs189599481 Chr.11: 64307133-64307133 on Build GRCh38
rs373399001 Chr.11: 64307201-64307201 on Build GRCh38
rs764333747 Chr.11: 64307255-64307255 on Build GRCh38
rs747624545 Chr.11: 64307176-64307176 on Build GRCh38
rs756063614 Chr.11: 64307503-64307503 on Build GRCh38
rs759516822 Chr.11: 64307470-64307470 on Build GRCh38
rs749450239 Chr.11: 64307292-64307292 on Build GRCh38
rs374570016 Chr.11: 64307543-64307543 on Build GRCh38
rs748235199 Chr.11: 64307377-64307377 on Build GRCh38
rs770105556 Chr.11: 64307229-64307229 on Build GRCh38
rs560892872 Chr.11: 64307352-64307352 on Build GRCh38
rs576087229 Chr.11: 64307143-64307143 on Build GRCh38
rs200261687 Chr.11: 64307272-64307272 on Build GRCh38
rs774575272 Chr.11: 64307256-64307256 on Build GRCh38
rs754206501 Chr.11: 64307325-64307325 on Build GRCh38
rs568939011 Chr.11: 64307375-64307375 on Build GRCh38
rs757990720 Chr.11: 64307327-64307327 on Build GRCh38
rs377634093 Chr.11: 64307260-64307260 on Build GRCh38
rs761377564 Chr.11: 64307445-64307445 on Build GRCh38
rs762319495 Chr.11: 64307258-64307258 on Build GRCh38
rs549790798 Chr.11: 64307374-64307374 on Build GRCh38
rs752386768 Chr.11: 64307350-64307351 on Build GRCh38
rs773764691 Chr.11: 64307403-64307403 on Build GRCh38
rs763158722 Chr.11: 64307150-64307150 on Build GRCh38
rs769011427 Chr.11: 64307293-64307293 on Build GRCh38
rs377457765 Chr.11: 64307177-64307177 on Build GRCh38
rs754868069 Chr.11: 64307525-64307525 on Build GRCh38
rs780168774 Chr.11: 64307520-64307520 on Build GRCh38
rs374554380 Chr.11: 64307329-64307329 on Build GRCh38
rs769763318 Chr.11: 64307285-64307285 on Build GRCh38
rs1032848222 Chr.11: 64307275-64307275 on Build GRCh38
rs780126062 Chr.11: 64307230-64307230 on Build GRCh38
rs768541509 Chr.11: 64307277-64307277 on Build GRCh38
rs367797877 Chr.11: 64307482-64307482 on Build GRCh38
rs763105612 Chr.11: 64307234-64307234 on Build GRCh38
rs117285599 Chr.11: 64307271-64307271 on Build GRCh38
rs927465480 Chr.11: 64307142-64307142 on Build GRCh38
rs752166239 Chr.11: 64307156-64307156 on Build GRCh38
rs750762788 Chr.11: 64307162-64307162 on Build GRCh38
rs749114932 Chr.11: 64307372-64307372 on Build GRCh38
rs752404387 Chr.11: 64307268-64307268 on Build GRCh38
rs764846318 Chr.11: 64307266-64307266 on Build GRCh38
rs756476016 Chr.11: 64307263-64307263 on Build GRCh38
rs745675889 Chr.11: 64307214-64307214 on Build GRCh38
rs781549015 Chr.11: 64307161-64307161 on Build GRCh38
rs35472129 Chr.11: 64307396-64307397 on Build GRCh38
rs376539921 Chr.11: 64307431-64307431 on Build GRCh38
rs745647545 Chr.11: 64307284-64307284 on Build GRCh38
rs757950233 Chr.11: 64307186-64307186 on Build GRCh38
rs761087370 Chr.11: 64307313-64307313 on Build GRCh38
rs61739389 Chr.11: 64307303-64307303 on Build GRCh38
rs950108650 Chr.11: 64307114-64307114 on Build GRCh38
rs201997221 Chr.11: 64307437-64307437 on Build GRCh38
rs779182187 Chr.11: 64307528-64307528 on Build GRCh38
rs752779312 Chr.11: 64307144-64307144 on Build GRCh38
rs765798256 Chr.11: 64307547-64307547 on Build GRCh38
rs878989820 Chr.11: 64307247-64307247 on Build GRCh38
rs530019281 Chr.11: 64307365-64307365 on Build GRCh38
rs751043001 Chr.11: 64307332-64307332 on Build GRCh38
rs761105169 Chr.11: 64307404-64307404 on Build GRCh38
rs781171709 Chr.11: 64307351-64307351 on Build GRCh38
rs370988249 Chr.11: 64307498-64307498 on Build GRCh38
rs781025976 Chr.11: 64307276-64307276 on Build GRCh38
rs775847343 Chr.11: 64307286-64307286 on Build GRCh38
rs753654303 Chr.11: 64307524-64307524 on Build GRCh38
rs369905424 Chr.11: 64307210-64307210 on Build GRCh38
rs4930694 Chr.11: 64307151-64307151 on Build GRCh38
rs756759935 Chr.11: 64307350-64307350 on Build GRCh38

View Full Product Details