Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64341148-64341639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88B
Entrez Gene ID
283234
Gene Name
coiled-coil domain containing 88B
Gene Aliases
BRLZ, CCDC88, HKRP3, gipie
UniGene
Hs.98564
Gene Chromosome Location
Chr.11: 64340203-64357534 on Build GRCh38
Transcripts
NM_032251.5 XM_006718519.3 XM_011544957.2 XM_011544959.2 XM_011544960.2 XM_017017601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2163471 c.364A>C p.T122P Chr.11: 64341154-64341154 on Build GRCh38
1492657 c.436G>A p.A146T Chr.11: 64341317-64341317 on Build GRCh38
4823683 c.489C>T p.L163L Chr.11: 64341462-64341462 on Build GRCh38
4035113 c.458G>A p.R153Q Chr.11: 64341431-64341431 on Build GRCh38
6876115 c.526C>T p.Q176* Chr.11: 64341499-64341499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961976142 Chr.11: 64341430-64341430 on Build GRCh38
rs375096255 Chr.11: 64341582-64341582 on Build GRCh38
rs772792497 Chr.11: 64341464-64341464 on Build GRCh38
rs750979749 Chr.11: 64341516-64341516 on Build GRCh38
rs575895616 Chr.11: 64341390-64341390 on Build GRCh38
rs766929936 Chr.11: 64341248-64341248 on Build GRCh38
rs776765853 Chr.11: 64341477-64341477 on Build GRCh38
rs747595586 Chr.11: 64341303-64341303 on Build GRCh38
rs761193202 Chr.11: 64341618-64341618 on Build GRCh38
rs536003588 Chr.11: 64341353-64341353 on Build GRCh38
rs374646135 Chr.11: 64341361-64341361 on Build GRCh38
rs776574188 Chr.11: 64341372-64341372 on Build GRCh38
rs1023781906 Chr.11: 64341356-64341356 on Build GRCh38
rs933443506 Chr.11: 64341182-64341182 on Build GRCh38
rs755265664 Chr.11: 64341282-64341282 on Build GRCh38
rs371459691 Chr.11: 64341224-64341224 on Build GRCh38
rs200216096 Chr.11: 64341594-64341594 on Build GRCh38
rs777354647 Chr.11: 64341557-64341557 on Build GRCh38
rs758521714 Chr.11: 64341401-64341401 on Build GRCh38
rs756527659 Chr.11: 64341433-64341433 on Build GRCh38
rs746169612 Chr.11: 64341318-64341318 on Build GRCh38
rs372493314 Chr.11: 64341176-64341176 on Build GRCh38
rs773861608 Chr.11: 64341228-64341228 on Build GRCh38
rs1035604338 Chr.11: 64341411-64341411 on Build GRCh38
rs760506272 Chr.11: 64341274-64341274 on Build GRCh38
rs1021906628 Chr.11: 64341333-64341333 on Build GRCh38
rs770612437 Chr.11: 64341565-64341565 on Build GRCh38
rs774055112 Chr.11: 64341352-64341352 on Build GRCh38
rs781493402 Chr.11: 64341210-64341211 on Build GRCh38
rs746367594 Chr.11: 64341431-64341431 on Build GRCh38
rs753639035 Chr.11: 64341276-64341276 on Build GRCh38
rs367964927 Chr.11: 64341610-64341610 on Build GRCh38
rs1047290992 Chr.11: 64341187-64341187 on Build GRCh38
rs578157759 Chr.11: 64341219-64341219 on Build GRCh38
rs142786551 Chr.11: 64341599-64341599 on Build GRCh38
rs779212070 Chr.11: 64341283-64341283 on Build GRCh38
rs766236393 Chr.11: 64341275-64341275 on Build GRCh38
rs377292763 Chr.11: 64341471-64341471 on Build GRCh38
rs199674677 Chr.11: 64341446-64341446 on Build GRCh38
rs775381249 Chr.11: 64341489-64341489 on Build GRCh38
rs751666917 Chr.11: 64341502-64341502 on Build GRCh38
rs370989286 Chr.11: 64341552-64341552 on Build GRCh38
rs752965257 Chr.11: 64341291-64341291 on Build GRCh38
rs971949522 Chr.11: 64341459-64341459 on Build GRCh38
rs772463081 Chr.11: 64341175-64341175 on Build GRCh38
rs758073998 Chr.11: 64341556-64341556 on Build GRCh38
rs764269192 Chr.11: 64341406-64341406 on Build GRCh38
rs781555613 Chr.11: 64341316-64341316 on Build GRCh38
rs376267018 Chr.11: 64341201-64341201 on Build GRCh38
rs200711256 Chr.11: 64341588-64341588 on Build GRCh38
rs201265638 Chr.11: 64341499-64341499 on Build GRCh38
rs201011301 Chr.11: 64341591-64341591 on Build GRCh38
rs768300049 Chr.11: 64341338-64341338 on Build GRCh38
rs756583196 Chr.11: 64341532-64341532 on Build GRCh38
rs747461484 Chr.11: 64341200-64341200 on Build GRCh38
rs759521744 Chr.11: 64341480-64341480 on Build GRCh38
rs771584414 Chr.11: 64341457-64341457 on Build GRCh38
rs975002585 Chr.11: 64341196-64341196 on Build GRCh38
rs765159929 Chr.11: 64341483-64341483 on Build GRCh38
rs750462536 Chr.11: 64341613-64341613 on Build GRCh38
rs751540151 Chr.11: 64341418-64341418 on Build GRCh38
rs780428012 Chr.11: 64341550-64341550 on Build GRCh38
rs753020371 Chr.11: 64341398-64341398 on Build GRCh38
rs779903561 Chr.11: 64341564-64341589 on Build GRCh38
rs140757172 Chr.11: 64341293-64341293 on Build GRCh38
rs780372875 Chr.11: 64341436-64341436 on Build GRCh38
rs746657628 Chr.11: 64341563-64341563 on Build GRCh38
rs766170572 Chr.11: 64341371-64341371 on Build GRCh38
rs767574030 Chr.11: 64341514-64341514 on Build GRCh38
rs762002647 Chr.11: 64341609-64341609 on Build GRCh38
rs567118296 Chr.11: 64341296-64341296 on Build GRCh38
rs556067934 Chr.11: 64341564-64341566 on Build GRCh38
rs756154661 Chr.11: 64341415-64341415 on Build GRCh38
rs781610595 Chr.11: 64341428-64341428 on Build GRCh38
rs778414269 Chr.11: 64341195-64341195 on Build GRCh38
rs578081356 Chr.11: 64341505-64341505 on Build GRCh38
rs1008150967 Chr.11: 64341211-64341211 on Build GRCh38
rs758565396 Chr.11: 64341294-64341294 on Build GRCh38
rs767696733 Chr.11: 64341574-64341574 on Build GRCh38
rs547619685 Chr.11: 64341263-64341263 on Build GRCh38
rs947516415 Chr.11: 64341191-64341191 on Build GRCh38
rs750535053 Chr.11: 64341575-64341575 on Build GRCh38
rs556044699 Chr.11: 64341355-64341355 on Build GRCh38
rs757792174 Chr.11: 64341427-64341427 on Build GRCh38
rs757562370 Chr.11: 64341194-64341196 on Build GRCh38
rs751807015 Chr.11: 64341237-64341237 on Build GRCh38
rs760698705 Chr.11: 64341337-64341337 on Build GRCh38
rs761244341 Chr.11: 64341246-64341246 on Build GRCh38
rs776834402 Chr.11: 64341210-64341210 on Build GRCh38
rs746009798 Chr.11: 64341222-64341224 on Build GRCh38
rs150457686 Chr.11: 64341484-64341484 on Build GRCh38
rs765112896 Chr.11: 64341394-64341394 on Build GRCh38
rs771298358 Chr.11: 64341307-64341307 on Build GRCh38

View Full Product Details