Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64826566-64827064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPG
Entrez Gene ID
55561
Gene Name
CDC42 binding protein kinase gamma
Gene Aliases
DMPK2, HSMDPKIN, KAPPA-200, MRCKG, MRCKgamma
UniGene
Hs.293590
Gene Chromosome Location
Chr.11: 64823809-64844685 on Build GRCh38
Transcripts
NM_017525.2 XM_011545155.2 XM_011545156.2 XM_011545157.2 XM_011545158.2 XM_011545159.2 XM_011545160.2 XM_017017996.1 XM_017017999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6412677 c.4484G>T p.S1495I Chr.11: 64826700-64826700 on Build GRCh38
4444356 c.4465C>T p.R1489C Chr.11: 64826719-64826719 on Build GRCh38
3869868 c.4486G>A p.E1496K Chr.11: 64826698-64826698 on Build GRCh38
5724090 c.4417C>T p.R1473C Chr.11: 64826767-64826767 on Build GRCh38
6393923 c.4434G>C p.Q1478H Chr.11: 64826750-64826750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770683579 Chr.11: 64827029-64827030 on Build GRCh38
rs939289220 Chr.11: 64826859-64826859 on Build GRCh38
rs374393124 Chr.11: 64826896-64826896 on Build GRCh38
rs765806049 Chr.11: 64826686-64826686 on Build GRCh38
rs2957159 Chr.11: 64826713-64826713 on Build GRCh38
rs765276102 Chr.11: 64826789-64826789 on Build GRCh38
rs748128816 Chr.11: 64826742-64826743 on Build GRCh38
rs527870287 Chr.11: 64826772-64826772 on Build GRCh38
rs757443824 Chr.11: 64826841-64826841 on Build GRCh38
rs763947851 Chr.11: 64826838-64826838 on Build GRCh38
rs968097316 Chr.11: 64826852-64826852 on Build GRCh38
rs554762176 Chr.11: 64826993-64826993 on Build GRCh38
rs367859198 Chr.11: 64826667-64826667 on Build GRCh38
rs201808737 Chr.11: 64826665-64826665 on Build GRCh38
rs781438449 Chr.11: 64826842-64826842 on Build GRCh38
rs753990804 Chr.11: 64826633-64826633 on Build GRCh38
rs915308256 Chr.11: 64826851-64826851 on Build GRCh38
rs367649320 Chr.11: 64826762-64826762 on Build GRCh38
rs752541965 Chr.11: 64826720-64826720 on Build GRCh38
rs1009126838 Chr.11: 64826802-64826802 on Build GRCh38
rs751855035 Chr.11: 64826839-64826839 on Build GRCh38
rs765223133 Chr.11: 64826699-64826699 on Build GRCh38
rs1042582372 Chr.11: 64826707-64826707 on Build GRCh38
rs777723007 Chr.11: 64826789-64826789 on Build GRCh38
rs865838329 Chr.11: 64826788-64826788 on Build GRCh38
rs923912806 Chr.11: 64826858-64826858 on Build GRCh38
rs898196532 Chr.11: 64826758-64826758 on Build GRCh38
rs770491188 Chr.11: 64827030-64827030 on Build GRCh38
rs776112523 Chr.11: 64827040-64827053 on Build GRCh38
rs755087734 Chr.11: 64826632-64826633 on Build GRCh38
rs200512542 Chr.11: 64826673-64826673 on Build GRCh38
rs1043022202 Chr.11: 64826892-64826892 on Build GRCh38
rs538659781 Chr.11: 64826914-64826914 on Build GRCh38
rs775153521 Chr.11: 64826756-64826756 on Build GRCh38
rs770901698 Chr.11: 64826625-64826625 on Build GRCh38
rs547633009 Chr.11: 64826849-64826849 on Build GRCh38
rs371690004 Chr.11: 64826690-64826690 on Build GRCh38
rs894960056 Chr.11: 64826709-64826709 on Build GRCh38
rs747125782 Chr.11: 64826596-64826596 on Build GRCh38
rs1009160727 Chr.11: 64826931-64826931 on Build GRCh38
rs371150322 Chr.11: 64826649-64826649 on Build GRCh38
rs753856418 Chr.11: 64826689-64826689 on Build GRCh38
rs766004484 Chr.11: 64826775-64826775 on Build GRCh38
rs869151434 Chr.11: 64826613-64826613 on Build GRCh38
rs756129538 Chr.11: 64827015-64827015 on Build GRCh38
rs1022609680 Chr.11: 64826987-64826987 on Build GRCh38
rs747191755 Chr.11: 64826727-64826727 on Build GRCh38
rs529702150 Chr.11: 64826855-64826855 on Build GRCh38
rs1042345780 Chr.11: 64826664-64826664 on Build GRCh38
rs1019199701 Chr.11: 64826945-64826945 on Build GRCh38
rs376711604 Chr.11: 64826825-64826825 on Build GRCh38
rs1019125296 Chr.11: 64826815-64826815 on Build GRCh38
rs760294420 Chr.11: 64826674-64826674 on Build GRCh38
rs915225559 Chr.11: 64826867-64826867 on Build GRCh38
rs180725021 Chr.11: 64826895-64826895 on Build GRCh38
rs567524682 Chr.11: 64826850-64826850 on Build GRCh38
rs915252166 Chr.11: 64826615-64826615 on Build GRCh38
rs534917236 Chr.11: 64826962-64826962 on Build GRCh38
rs61744174 Chr.11: 64826740-64826740 on Build GRCh38
rs780599553 Chr.11: 64827017-64827017 on Build GRCh38
rs369359774 Chr.11: 64826932-64826933 on Build GRCh38
rs772065901 Chr.11: 64826767-64826767 on Build GRCh38
rs767655576 Chr.11: 64826918-64826918 on Build GRCh38
rs1012556662 Chr.11: 64826978-64826978 on Build GRCh38
rs746766642 Chr.11: 64827015-64827015 on Build GRCh38
rs769051055 Chr.11: 64827027-64827027 on Build GRCh38
rs565606476 Chr.11: 64826760-64826760 on Build GRCh38
rs900707789 Chr.11: 64826961-64826961 on Build GRCh38
rs947015866 Chr.11: 64826889-64826889 on Build GRCh38
rs779087727 Chr.11: 64826737-64826738 on Build GRCh38
rs758205938 Chr.11: 64826721-64826721 on Build GRCh38
rs772987595 Chr.11: 64826765-64826765 on Build GRCh38
rs895542130 Chr.11: 64826929-64826930 on Build GRCh38
rs759171305 Chr.11: 64827035-64827035 on Build GRCh38
rs371404482 Chr.11: 64826787-64826787 on Build GRCh38
rs558560424 Chr.11: 64826919-64826919 on Build GRCh38
rs776034595 Chr.11: 64826783-64826783 on Build GRCh38
rs647145 Chr.11: 64826921-64826921 on Build GRCh38
rs758334833 Chr.11: 64826836-64826836 on Build GRCh38
rs778198482 Chr.11: 64826722-64826722 on Build GRCh38
rs569525623 Chr.11: 64826898-64826898 on Build GRCh38
rs975191908 Chr.11: 64826608-64826608 on Build GRCh38
rs531751681 Chr.11: 64826731-64826731 on Build GRCh38
rs745913569 Chr.11: 64826736-64826736 on Build GRCh38
rs1022404879 Chr.11: 64826967-64826967 on Build GRCh38
rs777259807 Chr.11: 64826633-64826633 on Build GRCh38
rs992000358 Chr.11: 64826868-64826868 on Build GRCh38
rs748502915 Chr.11: 64827029-64827029 on Build GRCh38
rs61886081 Chr.11: 64826611-64826611 on Build GRCh38
rs763265252 Chr.11: 64826644-64826644 on Build GRCh38
rs916424733 Chr.11: 64826881-64826881 on Build GRCh38
rs745939677 Chr.11: 64827004-64827004 on Build GRCh38
rs770194301 Chr.11: 64826636-64826636 on Build GRCh38
rs769422630 Chr.11: 64827037-64827037 on Build GRCh38
rs772687112 Chr.11: 64826672-64826672 on Build GRCh38
rs950933484 Chr.11: 64826791-64826791 on Build GRCh38
rs646709 Chr.11: 64827018-64827018 on Build GRCh38
rs890773660 Chr.11: 64826929-64826929 on Build GRCh38
rs776285558 Chr.11: 64827033-64827033 on Build GRCh38
rs781107734 Chr.11: 64826735-64826735 on Build GRCh38

View Full Product Details