Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64854071-64854606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD1
Entrez Gene ID
10938
Gene Name
EH domain containing 1
Gene Aliases
H-PAST, HPAST1, PAST, PAST1
UniGene
Hs.523774
Gene Chromosome Location
Chr.11: 64852727-64879713 on Build GRCh38
Transcripts
NM_001282444.1 NM_001282445.1 NM_006795.3 XM_011544739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5743476 c.1336C>T p.P446S Chr.11: 64854602-64854602 on Build GRCh38
5499879 c.1367C>T p.S456F Chr.11: 64854571-64854571 on Build GRCh38
2164621 c.1569G>A p.P523P Chr.11: 64854369-64854369 on Build GRCh38
6652409 c.1495G>A p.E499K Chr.11: 64854443-64854443 on Build GRCh38
6296779 c.1388C>A p.T463K Chr.11: 64854550-64854550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769943543 Chr.11: 64854299-64854299 on Build GRCh38
rs201518487 Chr.11: 64854383-64854383 on Build GRCh38
rs567807578 Chr.11: 64854463-64854463 on Build GRCh38
rs762558324 Chr.11: 64854379-64854379 on Build GRCh38
rs748553155 Chr.11: 64854357-64854357 on Build GRCh38
rs370715286 Chr.11: 64854301-64854301 on Build GRCh38
rs780161304 Chr.11: 64854328-64854328 on Build GRCh38
rs556269920 Chr.11: 64854513-64854513 on Build GRCh38
rs370511627 Chr.11: 64854322-64854322 on Build GRCh38
rs566884714 Chr.11: 64854180-64854180 on Build GRCh38
rs771446626 Chr.11: 64854509-64854509 on Build GRCh38
rs368623232 Chr.11: 64854221-64854221 on Build GRCh38
rs267603107 Chr.11: 64854392-64854392 on Build GRCh38
rs150171959 Chr.11: 64854459-64854459 on Build GRCh38
rs755070502 Chr.11: 64854416-64854416 on Build GRCh38
rs116092297 Chr.11: 64854297-64854297 on Build GRCh38
rs565074634 Chr.11: 64854165-64854165 on Build GRCh38
rs759301415 Chr.11: 64854365-64854365 on Build GRCh38
rs748991388 Chr.11: 64854519-64854519 on Build GRCh38
rs8855 Chr.11: 64854101-64854101 on Build GRCh38
rs781734567 Chr.11: 64854579-64854579 on Build GRCh38
rs750305737 Chr.11: 64854456-64854456 on Build GRCh38
rs373695316 Chr.11: 64854279-64854279 on Build GRCh38
rs920241270 Chr.11: 64854420-64854420 on Build GRCh38
rs375831742 Chr.11: 64854311-64854311 on Build GRCh38
rs757811551 Chr.11: 64854577-64854577 on Build GRCh38
rs775315983 Chr.11: 64854444-64854444 on Build GRCh38
rs969701520 Chr.11: 64854105-64854105 on Build GRCh38
rs763660911 Chr.11: 64854380-64854380 on Build GRCh38
rs774868370 Chr.11: 64854346-64854346 on Build GRCh38
rs768335405 Chr.11: 64854447-64854447 on Build GRCh38
rs760582220 Chr.11: 64854549-64854549 on Build GRCh38
rs958886354 Chr.11: 64854473-64854473 on Build GRCh38
rs577914472 Chr.11: 64854278-64854278 on Build GRCh38
rs755469852 Chr.11: 64854310-64854310 on Build GRCh38
rs777466279 Chr.11: 64854574-64854574 on Build GRCh38
rs116537817 Chr.11: 64854213-64854213 on Build GRCh38
rs370911440 Chr.11: 64854454-64854454 on Build GRCh38
rs773707020 Chr.11: 64854450-64854450 on Build GRCh38
rs556266157 Chr.11: 64854174-64854174 on Build GRCh38
rs749381158 Chr.11: 64854329-64854329 on Build GRCh38
rs754018251 Chr.11: 64854411-64854411 on Build GRCh38
rs778148331 Chr.11: 64854501-64854501 on Build GRCh38
rs985353293 Chr.11: 64854507-64854507 on Build GRCh38
rs138745282 Chr.11: 64854302-64854302 on Build GRCh38
rs761231096 Chr.11: 64854540-64854540 on Build GRCh38
rs868241191 Chr.11: 64854232-64854232 on Build GRCh38
rs535925350 Chr.11: 64854207-64854207 on Build GRCh38
rs766201999 Chr.11: 64854555-64854555 on Build GRCh38
rs575944576 Chr.11: 64854300-64854300 on Build GRCh38
rs758470055 Chr.11: 64854316-64854316 on Build GRCh38
rs370182623 Chr.11: 64854431-64854431 on Build GRCh38
rs374529326 Chr.11: 64854432-64854432 on Build GRCh38
rs769559617 Chr.11: 64854567-64854568 on Build GRCh38
rs762168746 Chr.11: 64854309-64854309 on Build GRCh38
rs759813362 Chr.11: 64854560-64854560 on Build GRCh38
rs534163322 Chr.11: 64854281-64854281 on Build GRCh38
rs558563641 Chr.11: 64854571-64854571 on Build GRCh38
rs954952083 Chr.11: 64854347-64854347 on Build GRCh38
rs549419275 Chr.11: 64854381-64854381 on Build GRCh38
rs758446769 Chr.11: 64854495-64854495 on Build GRCh38
rs752706189 Chr.11: 64854474-64854474 on Build GRCh38
rs554327697 Chr.11: 64854293-64854293 on Build GRCh38
rs768406850 Chr.11: 64854522-64854522 on Build GRCh38
rs772454605 Chr.11: 64854433-64854433 on Build GRCh38
rs541253559 Chr.11: 64854318-64854318 on Build GRCh38
rs762647797 Chr.11: 64854446-64854446 on Build GRCh38
rs369765857 Chr.11: 64854277-64854277 on Build GRCh38
rs200964887 Chr.11: 64854390-64854390 on Build GRCh38
rs576194809 Chr.11: 64854541-64854541 on Build GRCh38
rs764970881 Chr.11: 64854369-64854369 on Build GRCh38
rs745821544 Chr.11: 64854505-64854513 on Build GRCh38
rs747511970 Chr.11: 64854505-64854505 on Build GRCh38
rs200832610 Chr.11: 64854298-64854298 on Build GRCh38
rs551660619 Chr.11: 64854107-64854107 on Build GRCh38
rs569201757 Chr.11: 64854220-64854220 on Build GRCh38
rs984652057 Chr.11: 64854123-64854123 on Build GRCh38
rs547602653 Chr.11: 64854172-64854172 on Build GRCh38
rs376757467 Chr.11: 64854263-64854263 on Build GRCh38
rs3897618 Chr.11: 64854384-64854384 on Build GRCh38
rs534200860 Chr.11: 64854435-64854435 on Build GRCh38
rs764141773 Chr.11: 64854317-64854317 on Build GRCh38
rs1032477936 Chr.11: 64854233-64854233 on Build GRCh38
rs758441856 Chr.11: 64854567-64854567 on Build GRCh38
rs367709253 Chr.11: 64854441-64854441 on Build GRCh38
rs554087346 Chr.11: 64854453-64854453 on Build GRCh38
rs565216224 Chr.11: 64854304-64854304 on Build GRCh38
rs752251630 Chr.11: 64854315-64854315 on Build GRCh38
rs910451434 Chr.11: 64854580-64854580 on Build GRCh38
rs940666057 Chr.11: 64854116-64854116 on Build GRCh38
rs775411023 Chr.11: 64854582-64854582 on Build GRCh38
rs758380064 Chr.11: 64854289-64854289 on Build GRCh38
rs754106431 Chr.11: 64854467-64854467 on Build GRCh38
rs535486154 Chr.11: 64854576-64854576 on Build GRCh38
rs529387634 Chr.11: 64854370-64854370 on Build GRCh38
rs755160517 Chr.11: 64854470-64854470 on Build GRCh38
rs1036307074 Chr.11: 64854128-64854128 on Build GRCh38
rs934794441 Chr.11: 64854280-64854280 on Build GRCh38
rs377232695 Chr.11: 64854393-64854393 on Build GRCh38
rs772535208 Chr.11: 64854362-64854362 on Build GRCh38
rs538160490 Chr.11: 64854243-64854243 on Build GRCh38
rs181239788 Chr.11: 64854166-64854166 on Build GRCh38
rs202195664 Chr.11: 64854561-64854561 on Build GRCh38
rs747349369 Chr.11: 64854437-64854437 on Build GRCh38
rs138617869 Chr.11: 64854471-64854471 on Build GRCh38
rs753625953 Chr.11: 64854558-64854558 on Build GRCh38
rs768654185 Chr.11: 64854341-64854341 on Build GRCh38
rs1043264835 Chr.11: 64854274-64854275 on Build GRCh38
rs34740136 Chr.11: 64854546-64854546 on Build GRCh38
rs897745909 Chr.11: 64854523-64854523 on Build GRCh38
rs775227378 Chr.11: 64854372-64854372 on Build GRCh38
rs573676955 Chr.11: 64854294-64854294 on Build GRCh38
rs909230028 Chr.11: 64854106-64854106 on Build GRCh38
rs773992056 Chr.11: 64854539-64854539 on Build GRCh38
rs199602914 Chr.11: 64854325-64854325 on Build GRCh38
rs1016088375 Chr.11: 64854145-64854145 on Build GRCh38
rs972156852 Chr.11: 64854203-64854203 on Build GRCh38
rs746154578 Chr.11: 64854324-64854324 on Build GRCh38

View Full Product Details