Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64894440-64894936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG2A
Entrez Gene ID
23130
Gene Name
autophagy related 2A
Gene Aliases
-
UniGene
Hs.370671
Gene Chromosome Location
Chr.11: 64894532-64917250 on Build GRCh38
Transcripts
NM_015104.2 XM_005273849.3 XM_005273850.3 XM_011544863.2 XM_011544864.2 XM_011544865.2 XM_011544868.2

SNP Detail

SNP ID Chromosome Location
rs781618803 Chr.11: 64894888-64894888 on Build GRCh38
rs375229974 Chr.11: 64894840-64894840 on Build GRCh38
rs771551561 Chr.11: 64894902-64894902 on Build GRCh38
rs748203664 Chr.11: 64894768-64894768 on Build GRCh38
rs897771302 Chr.11: 64894601-64894601 on Build GRCh38
rs768203544 Chr.11: 64894829-64894829 on Build GRCh38
rs748196442 Chr.11: 64894631-64894631 on Build GRCh38
rs761574985 Chr.11: 64894881-64894882 on Build GRCh38
rs1037678541 Chr.11: 64894586-64894586 on Build GRCh38
rs1008997525 Chr.11: 64894874-64894874 on Build GRCh38
rs778857576 Chr.11: 64894614-64894614 on Build GRCh38
rs760687412 Chr.11: 64894852-64894852 on Build GRCh38
rs770154457 Chr.11: 64894892-64894892 on Build GRCh38
rs534459597 Chr.11: 64894778-64894778 on Build GRCh38
rs924425577 Chr.11: 64894537-64894537 on Build GRCh38
rs772474586 Chr.11: 64894649-64894649 on Build GRCh38
rs565927848 Chr.11: 64894764-64894764 on Build GRCh38
rs779139100 Chr.11: 64894767-64894767 on Build GRCh38
rs750432360 Chr.11: 64894846-64894846 on Build GRCh38
rs182319197 Chr.11: 64894486-64894486 on Build GRCh38
rs757762198 Chr.11: 64894887-64894887 on Build GRCh38
rs370910848 Chr.11: 64894619-64894619 on Build GRCh38
rs756230557 Chr.11: 64894583-64894583 on Build GRCh38
rs200510974 Chr.11: 64894738-64894738 on Build GRCh38
rs537013037 Chr.11: 64894862-64894862 on Build GRCh38
rs902795385 Chr.11: 64894897-64894897 on Build GRCh38
rs200063859 Chr.11: 64894855-64894855 on Build GRCh38
rs761324656 Chr.11: 64894721-64894721 on Build GRCh38
rs772926397 Chr.11: 64894907-64894907 on Build GRCh38
rs746237202 Chr.11: 64894891-64894891 on Build GRCh38
rs970230004 Chr.11: 64894741-64894741 on Build GRCh38
rs576977881 Chr.11: 64894493-64894493 on Build GRCh38
rs774222990 Chr.11: 64894868-64894868 on Build GRCh38
rs1037626493 Chr.11: 64894848-64894848 on Build GRCh38
rs751032846 Chr.11: 64894720-64894720 on Build GRCh38
rs890306421 Chr.11: 64894677-64894677 on Build GRCh38
rs779984288 Chr.11: 64894743-64894743 on Build GRCh38
rs768603032 Chr.11: 64894819-64894819 on Build GRCh38
rs773913867 Chr.11: 64894896-64894896 on Build GRCh38
rs777819723 Chr.11: 64894875-64894875 on Build GRCh38
rs547862788 Chr.11: 64894818-64894818 on Build GRCh38
rs772155188 Chr.11: 64894777-64894777 on Build GRCh38
rs756012179 Chr.11: 64894740-64894740 on Build GRCh38
rs1014197530 Chr.11: 64894908-64894908 on Build GRCh38
rs773720054 Chr.11: 64894688-64894688 on Build GRCh38
rs747495712 Chr.11: 64894797-64894797 on Build GRCh38
rs534891434 Chr.11: 64894650-64894650 on Build GRCh38
rs565379422 Chr.11: 64894540-64894540 on Build GRCh38
rs556994668 Chr.11: 64894906-64894906 on Build GRCh38
rs567755503 Chr.11: 64894853-64894853 on Build GRCh38
rs941781856 Chr.11: 64894831-64894831 on Build GRCh38
rs993422514 Chr.11: 64894618-64894618 on Build GRCh38
rs751536085 Chr.11: 64894802-64894802 on Build GRCh38
rs866189486 Chr.11: 64894496-64894496 on Build GRCh38
rs761259756 Chr.11: 64894839-64894839 on Build GRCh38
rs1021205875 Chr.11: 64894744-64894744 on Build GRCh38
rs754181426 Chr.11: 64894751-64894751 on Build GRCh38
rs565998317 Chr.11: 64894589-64894589 on Build GRCh38
rs866531708 Chr.11: 64894723-64894723 on Build GRCh38
rs762578757 Chr.11: 64894706-64894706 on Build GRCh38
rs747579646 Chr.11: 64894877-64894877 on Build GRCh38
rs771340699 Chr.11: 64894807-64894807 on Build GRCh38
rs938507311 Chr.11: 64894558-64894558 on Build GRCh38
rs765412974 Chr.11: 64894856-64894856 on Build GRCh38
rs748512903 Chr.11: 64894838-64894838 on Build GRCh38
rs1051657342 Chr.11: 64894637-64894637 on Build GRCh38
rs759826034 Chr.11: 64894813-64894813 on Build GRCh38
rs552312911 Chr.11: 64894762-64894762 on Build GRCh38
rs775174706 Chr.11: 64894701-64894701 on Build GRCh38
rs185714284 Chr.11: 64894707-64894707 on Build GRCh38
rs993558166 Chr.11: 64894643-64894643 on Build GRCh38
rs772855188 Chr.11: 64894903-64894903 on Build GRCh38
rs77692680 Chr.11: 64894498-64894498 on Build GRCh38
rs776972204 Chr.11: 64894809-64894809 on Build GRCh38
rs780451059 Chr.11: 64894566-64894566 on Build GRCh38
rs776653277 Chr.11: 64894472-64894472 on Build GRCh38
rs201943955 Chr.11: 64894748-64894748 on Build GRCh38
rs993003508 Chr.11: 64894539-64894539 on Build GRCh38
rs890358586 Chr.11: 64894864-64894864 on Build GRCh38
rs753699654 Chr.11: 64894854-64894854 on Build GRCh38
rs900680651 Chr.11: 64894884-64894884 on Build GRCh38
rs527745375 Chr.11: 64894580-64894580 on Build GRCh38
rs948868462 Chr.11: 64894550-64894550 on Build GRCh38
rs78213205 Chr.11: 64894734-64894734 on Build GRCh38
rs956003175 Chr.11: 64894733-64894733 on Build GRCh38
rs758460317 Chr.11: 64894869-64894869 on Build GRCh38
rs924894921 Chr.11: 64894495-64894495 on Build GRCh38
rs890509803 Chr.11: 64894868-64894868 on Build GRCh38
rs561311535 Chr.11: 64894689-64894689 on Build GRCh38
rs537534402 Chr.11: 64894704-64894707 on Build GRCh38
rs1037369026 Chr.11: 64894600-64894600 on Build GRCh38
rs948638520 Chr.11: 64894585-64894585 on Build GRCh38
rs1056993618 Chr.11: 64894571-64894571 on Build GRCh38

View Full Product Details