Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64896622-64897021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG2A
Entrez Gene ID
23130
Gene Name
autophagy related 2A
Gene Aliases
-
UniGene
Hs.370671
Gene Chromosome Location
Chr.11: 64894532-64917250 on Build GRCh38
Transcripts
NM_015104.2 XM_005273849.3 XM_005273850.3 XM_011544863.2 XM_011544864.2 XM_011544865.2 XM_011544866.2 XM_011544868.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7188665 c.5265C>G p.V1755V Chr.11: 64896755-64896755 on Build GRCh38
4914131 c.5226C>A p.P1742P Chr.11: 64896794-64896794 on Build GRCh38
6606645 c.5251A>G p.M1751V Chr.11: 64896769-64896769 on Build GRCh38
4915847 c.5210G>T p.R1737L Chr.11: 64896810-64896810 on Build GRCh38
689644 c.5191G>T p.E1731* Chr.11: 64896829-64896829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749022607 Chr.11: 64896819-64896819 on Build GRCh38
rs747790597 Chr.11: 64896654-64896654 on Build GRCh38
rs376512254 Chr.11: 64896954-64896955 on Build GRCh38
rs746563475 Chr.11: 64896793-64896793 on Build GRCh38
rs2509734 Chr.11: 64896980-64896980 on Build GRCh38
rs369521423 Chr.11: 64896700-64896700 on Build GRCh38
rs566467183 Chr.11: 64896863-64896863 on Build GRCh38
rs866713435 Chr.11: 64896883-64896883 on Build GRCh38
rs777427525 Chr.11: 64896659-64896659 on Build GRCh38
rs1048318649 Chr.11: 64896997-64896997 on Build GRCh38
rs995441262 Chr.11: 64896972-64896972 on Build GRCh38
rs774394054 Chr.11: 64896829-64896829 on Build GRCh38
rs780559476 Chr.11: 64896888-64896888 on Build GRCh38
rs770926724 Chr.11: 64896697-64896697 on Build GRCh38
rs186029133 Chr.11: 64896970-64896970 on Build GRCh38
rs1004421288 Chr.11: 64896911-64896911 on Build GRCh38
rs1013985578 Chr.11: 64896727-64896727 on Build GRCh38
rs751153590 Chr.11: 64896872-64896872 on Build GRCh38
rs750554511 Chr.11: 64896896-64896898 on Build GRCh38
rs116700761 Chr.11: 64896962-64896962 on Build GRCh38
rs781265243 Chr.11: 64896891-64896902 on Build GRCh38
rs775743582 Chr.11: 64896717-64896717 on Build GRCh38
rs778386503 Chr.11: 64896762-64896762 on Build GRCh38
rs180755321 Chr.11: 64896815-64896815 on Build GRCh38
rs931743501 Chr.11: 64896683-64896683 on Build GRCh38
rs779916115 Chr.11: 64896918-64896918 on Build GRCh38
rs763882132 Chr.11: 64896866-64896866 on Build GRCh38
rs758140381 Chr.11: 64896778-64896778 on Build GRCh38
rs965789744 Chr.11: 64896689-64896689 on Build GRCh38
rs1050180584 Chr.11: 64896696-64896696 on Build GRCh38
rs137960072 Chr.11: 64896830-64896830 on Build GRCh38
rs532962074 Chr.11: 64896788-64896788 on Build GRCh38
rs756394295 Chr.11: 64896645-64896645 on Build GRCh38
rs756853132 Chr.11: 64896885-64896885 on Build GRCh38
rs770861079 Chr.11: 64896966-64896966 on Build GRCh38
rs756549544 Chr.11: 64896755-64896755 on Build GRCh38
rs756694584 Chr.11: 64896794-64896794 on Build GRCh38
rs764231451 Chr.11: 64896720-64896720 on Build GRCh38
rs202081004 Chr.11: 64896767-64896767 on Build GRCh38
rs773836517 Chr.11: 64896835-64896835 on Build GRCh38
rs771775485 Chr.11: 64896658-64896658 on Build GRCh38
rs759695622 Chr.11: 64896721-64896721 on Build GRCh38
rs150740265 Chr.11: 64896722-64896722 on Build GRCh38
rs368862661 Chr.11: 64896646-64896646 on Build GRCh38
rs377170791 Chr.11: 64896761-64896761 on Build GRCh38
rs754040545 Chr.11: 64896668-64896668 on Build GRCh38
rs754279269 Chr.11: 64896859-64896859 on Build GRCh38
rs759420148 Chr.11: 64896707-64896707 on Build GRCh38
rs112253332 Chr.11: 64896643-64896643 on Build GRCh38
rs750864750 Chr.11: 64896747-64896747 on Build GRCh38
rs769558897 Chr.11: 64896712-64896712 on Build GRCh38
rs781243276 Chr.11: 64896795-64896795 on Build GRCh38
rs763153887 Chr.11: 64896719-64896719 on Build GRCh38
rs757557132 Chr.11: 64896880-64896881 on Build GRCh38
rs768887348 Chr.11: 64896820-64896820 on Build GRCh38
rs111792976 Chr.11: 64896912-64896912 on Build GRCh38
rs143510901 Chr.11: 64896809-64896809 on Build GRCh38
rs766936924 Chr.11: 64896845-64896845 on Build GRCh38
rs756055733 Chr.11: 64896915-64896915 on Build GRCh38
rs369146463 Chr.11: 64896881-64896881 on Build GRCh38
rs745306302 Chr.11: 64896903-64896903 on Build GRCh38
rs761091944 Chr.11: 64896844-64896844 on Build GRCh38
rs541670712 Chr.11: 64896653-64896653 on Build GRCh38
rs745775934 Chr.11: 64896797-64896797 on Build GRCh38
rs925109437 Chr.11: 64896699-64896699 on Build GRCh38

View Full Product Details