Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55934846-55935445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKA
Entrez Gene ID
1606
Gene Name
diacylglycerol kinase alpha
Gene Aliases
DAGK, DAGK1, DGK-alpha
UniGene
Hs.524488
Gene Chromosome Location
Chr.12: 55931076-55954023 on Build GRCh38
Transcripts
NM_001345.4 NM_201444.2 NM_201445.1 NM_201554.1 XM_005268689.2 XM_011537991.2 XM_011537993.2 XM_011537995.2 XM_017018901.1 XM_017018902.1 XM_017018903.1 XM_017018904.1 XM_017018905.1 XM_017018906.1 XM_017018907.1

SNP Detail

SNP ID Chromosome Location
rs922998259 Chr.12: 55935281-55935281 on Build GRCh38
rs1046509434 Chr.12: 55935032-55935032 on Build GRCh38
rs554053709 Chr.12: 55934977-55934977 on Build GRCh38
rs1042931711 Chr.12: 55935228-55935228 on Build GRCh38
rs914191532 Chr.12: 55935347-55935347 on Build GRCh38
rs796123120 Chr.12: 55935316-55935316 on Build GRCh38
rs767515981 Chr.12: 55935211-55935214 on Build GRCh38
rs910783105 Chr.12: 55934924-55934924 on Build GRCh38
rs969637043 Chr.12: 55934976-55934976 on Build GRCh38
rs936910637 Chr.12: 55934950-55934950 on Build GRCh38
rs1016135197 Chr.12: 55935212-55935212 on Build GRCh38
rs529867069 Chr.12: 55935307-55935307 on Build GRCh38
rs1009579718 Chr.12: 55935029-55935029 on Build GRCh38
rs936827231 Chr.12: 55935305-55935305 on Build GRCh38
rs114200298 Chr.12: 55934962-55934962 on Build GRCh38
rs956930507 Chr.12: 55935071-55935071 on Build GRCh38
rs78149473 Chr.12: 55935158-55935158 on Build GRCh38
rs181262747 Chr.12: 55935114-55935114 on Build GRCh38
rs971745343 Chr.12: 55935267-55935267 on Build GRCh38
rs1005710154 Chr.12: 55934946-55934946 on Build GRCh38
rs766808974 Chr.12: 55935127-55935127 on Build GRCh38
rs901049273 Chr.12: 55935301-55935301 on Build GRCh38
rs186134087 Chr.12: 55935311-55935311 on Build GRCh38
rs1041367298 Chr.12: 55935287-55935287 on Build GRCh38
rs1046457470 Chr.12: 55935363-55935363 on Build GRCh38
rs933485204 Chr.12: 55934904-55934904 on Build GRCh38
rs1046720911 Chr.12: 55935358-55935358 on Build GRCh38
rs756994284 Chr.12: 55934991-55934991 on Build GRCh38
rs888222163 Chr.12: 55935391-55935391 on Build GRCh38
rs1054006318 Chr.12: 55935344-55935344 on Build GRCh38
rs7308505 Chr.12: 55934915-55934915 on Build GRCh38
rs867838916 Chr.12: 55935254-55935254 on Build GRCh38
rs60022764 Chr.12: 55935414-55935414 on Build GRCh38
rs961653467 Chr.12: 55935249-55935249 on Build GRCh38
rs750753309 Chr.12: 55935255-55935255 on Build GRCh38
rs952492140 Chr.12: 55934935-55934935 on Build GRCh38
rs755518328 Chr.12: 55935164-55935164 on Build GRCh38
rs117439217 Chr.12: 55935302-55935302 on Build GRCh38
rs34587885 Chr.12: 55934871-55934872 on Build GRCh38
rs954515433 Chr.12: 55935297-55935297 on Build GRCh38

View Full Product Details