Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56639798-56640211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5B
Entrez Gene ID
506
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
Gene Aliases
ATPMB, ATPSB, HEL-S-271
UniGene
Hs.406510
Gene Chromosome Location
Chr.12: 56638175-56646068 on Build GRCh38
Transcripts
NM_001686.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5506409 c.1216C>T p.R406C Chr.12: 56640051-56640051 on Build GRCh38
6137412 c.1263C>T p.A421A Chr.12: 56640004-56640004 on Build GRCh38
5983710 c.1093G>C p.D365H Chr.12: 56640174-56640174 on Build GRCh38
3772695 c.1287+2T>G p.? Chr.12: 56639978-56639978 on Build GRCh38
7202594 c.1248_1256delGCATTACGA p.E416_Y418delEHY Chr.12: 56640011-56640019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752724870 Chr.12: 56639977-56639977 on Build GRCh38
rs545428507 Chr.12: 56639831-56639832 on Build GRCh38
rs753990283 Chr.12: 56639949-56639949 on Build GRCh38
rs528493009 Chr.12: 56639918-56639918 on Build GRCh38
rs765193597 Chr.12: 56639965-56639965 on Build GRCh38
rs755679536 Chr.12: 56640050-56640050 on Build GRCh38
rs766580897 Chr.12: 56639948-56639948 on Build GRCh38
rs764133649 Chr.12: 56640168-56640168 on Build GRCh38
rs775419624 Chr.12: 56640153-56640153 on Build GRCh38
rs781006746 Chr.12: 56640041-56640041 on Build GRCh38
rs933537224 Chr.12: 56640185-56640185 on Build GRCh38
rs748738029 Chr.12: 56640064-56640064 on Build GRCh38
rs771224329 Chr.12: 56640115-56640115 on Build GRCh38
rs745460216 Chr.12: 56640046-56640046 on Build GRCh38
rs966693186 Chr.12: 56639924-56639924 on Build GRCh38
rs779399792 Chr.12: 56640052-56640052 on Build GRCh38
rs12229457 Chr.12: 56639896-56639896 on Build GRCh38
rs546628597 Chr.12: 56639961-56639961 on Build GRCh38
rs764722110 Chr.12: 56640067-56640067 on Build GRCh38
rs747376704 Chr.12: 56640110-56640110 on Build GRCh38
rs923436881 Chr.12: 56640144-56640144 on Build GRCh38
rs943527181 Chr.12: 56640138-56640138 on Build GRCh38
rs202065156 Chr.12: 56640118-56640118 on Build GRCh38
rs780570761 Chr.12: 56639933-56639934 on Build GRCh38
rs368274816 Chr.12: 56640142-56640142 on Build GRCh38
rs943241795 Chr.12: 56640079-56640079 on Build GRCh38
rs751485472 Chr.12: 56640172-56640172 on Build GRCh38
rs545332621 Chr.12: 56639875-56639875 on Build GRCh38
rs777770019 Chr.12: 56640012-56640012 on Build GRCh38
rs758249534 Chr.12: 56639986-56639986 on Build GRCh38
rs959911699 Chr.12: 56639835-56639835 on Build GRCh38
rs1051700160 Chr.12: 56640033-56640033 on Build GRCh38
rs925433137 Chr.12: 56640020-56640020 on Build GRCh38
rs568588220 Chr.12: 56640039-56640039 on Build GRCh38
rs554853671 Chr.12: 56639874-56639874 on Build GRCh38
rs757138996 Chr.12: 56640181-56640181 on Build GRCh38
rs763014861 Chr.12: 56640160-56640160 on Build GRCh38
rs151237275 Chr.12: 56640013-56640013 on Build GRCh38
rs988689562 Chr.12: 56639838-56639838 on Build GRCh38
rs763772844 Chr.12: 56640109-56640109 on Build GRCh38

View Full Product Details