Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57806279-57806701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVIL
Entrez Gene ID
10677
Gene Name
advillin
Gene Aliases
ADVIL, DOC6, p92
UniGene
Hs.584854
Gene Chromosome Location
Chr.12: 57797372-57818704 on Build GRCh38
Transcripts
NM_006576.3 XM_017018710.1 XM_017018711.1 XM_017018712.1 XM_017018713.1 XM_017018714.1 XM_017018715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
942247 c.1637G>A p.R546Q Chr.12: 57806394-57806394 on Build GRCh38
694909 c.1617C>A p.S539S Chr.12: 57806414-57806414 on Build GRCh38
3463935 c.1604C>T p.S535F Chr.12: 57806427-57806427 on Build GRCh38
2156263 c.1574C>G p.T525S Chr.12: 57806457-57806457 on Build GRCh38
2156262 c.1573A>T p.T525S Chr.12: 57806458-57806458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944740649 Chr.12: 57806342-57806342 on Build GRCh38
rs1048351035 Chr.12: 57806534-57806534 on Build GRCh38
rs983632064 Chr.12: 57806520-57806520 on Build GRCh38
rs928555449 Chr.12: 57806326-57806326 on Build GRCh38
rs750487437 Chr.12: 57806509-57806509 on Build GRCh38
rs775358186 Chr.12: 57806307-57806308 on Build GRCh38
rs779545561 Chr.12: 57806552-57806552 on Build GRCh38
rs778333984 Chr.12: 57806443-57806443 on Build GRCh38
rs771805994 Chr.12: 57806551-57806551 on Build GRCh38
rs776512313 Chr.12: 57806579-57806579 on Build GRCh38
rs552650655 Chr.12: 57806526-57806526 on Build GRCh38
rs745526301 Chr.12: 57806449-57806449 on Build GRCh38
rs1035573250 Chr.12: 57806644-57806644 on Build GRCh38
rs780497014 Chr.12: 57806405-57806405 on Build GRCh38
rs376322015 Chr.12: 57806576-57806576 on Build GRCh38
rs755557542 Chr.12: 57806411-57806411 on Build GRCh38
rs886646616 Chr.12: 57806566-57806566 on Build GRCh38
rs746892386 Chr.12: 57806570-57806570 on Build GRCh38
rs1047851 Chr.12: 57806401-57806401 on Build GRCh38
rs771916309 Chr.12: 57806348-57806348 on Build GRCh38
rs549145073 Chr.12: 57806394-57806394 on Build GRCh38
rs367900033 Chr.12: 57806512-57806512 on Build GRCh38
rs781638853 Chr.12: 57806431-57806431 on Build GRCh38
rs771540578 Chr.12: 57806538-57806538 on Build GRCh38
rs368889532 Chr.12: 57806587-57806587 on Build GRCh38
rs756548425 Chr.12: 57806542-57806542 on Build GRCh38
rs150473391 Chr.12: 57806501-57806501 on Build GRCh38
rs779905314 Chr.12: 57806360-57806360 on Build GRCh38
rs960237279 Chr.12: 57806658-57806658 on Build GRCh38
rs969064372 Chr.12: 57806675-57806675 on Build GRCh38
rs187759826 Chr.12: 57806617-57806617 on Build GRCh38
rs930926767 Chr.12: 57806314-57806314 on Build GRCh38
rs771588357 Chr.12: 57806459-57806459 on Build GRCh38
rs761954969 Chr.12: 57806395-57806395 on Build GRCh38
rs769002961 Chr.12: 57806330-57806330 on Build GRCh38
rs113363824 Chr.12: 57806627-57806627 on Build GRCh38
rs375036175 Chr.12: 57806341-57806341 on Build GRCh38
rs771131780 Chr.12: 57806543-57806544 on Build GRCh38
rs766484991 Chr.12: 57806513-57806513 on Build GRCh38
rs755293608 Chr.12: 57806529-57806529 on Build GRCh38
rs760701700 Chr.12: 57806370-57806370 on Build GRCh38
rs773985981 Chr.12: 57806340-57806340 on Build GRCh38
rs775382103 Chr.12: 57806460-57806460 on Build GRCh38
rs747554651 Chr.12: 57806324-57806324 on Build GRCh38
rs748861123 Chr.12: 57806433-57806433 on Build GRCh38
rs369830395 Chr.12: 57806669-57806669 on Build GRCh38
rs762043345 Chr.12: 57806494-57806494 on Build GRCh38
rs371474877 Chr.12: 57806333-57806333 on Build GRCh38
rs761448851 Chr.12: 57806421-57806421 on Build GRCh38
rs770606856 Chr.12: 57806337-57806337 on Build GRCh38
rs201497225 Chr.12: 57806410-57806410 on Build GRCh38
rs769802237 Chr.12: 57806584-57806584 on Build GRCh38
rs776706732 Chr.12: 57806470-57806470 on Build GRCh38
rs775083813 Chr.12: 57806358-57806358 on Build GRCh38
rs1052236272 Chr.12: 57806609-57806609 on Build GRCh38
rs373501648 Chr.12: 57806309-57806309 on Build GRCh38
rs960653179 Chr.12: 57806408-57806408 on Build GRCh38
rs760369242 Chr.12: 57806463-57806463 on Build GRCh38
rs770410961 Chr.12: 57806436-57806436 on Build GRCh38
rs762907011 Chr.12: 57806588-57806588 on Build GRCh38
rs1023322969 Chr.12: 57806643-57806643 on Build GRCh38
rs1040352926 Chr.12: 57806345-57806345 on Build GRCh38
rs768285344 Chr.12: 57806582-57806583 on Build GRCh38
rs777062181 Chr.12: 57806608-57806608 on Build GRCh38
rs771986471 Chr.12: 57806301-57806302 on Build GRCh38
rs748769512 Chr.12: 57806334-57806334 on Build GRCh38
rs778427686 Chr.12: 57806544-57806544 on Build GRCh38
rs376981876 Chr.12: 57806323-57806323 on Build GRCh38
rs1039261270 Chr.12: 57806477-57806477 on Build GRCh38
rs759776451 Chr.12: 57806654-57806657 on Build GRCh38
rs768679508 Chr.12: 57806466-57806466 on Build GRCh38
rs763893013 Chr.12: 57806372-57806372 on Build GRCh38
rs367636607 Chr.12: 57806398-57806398 on Build GRCh38
rs372000272 Chr.12: 57806539-57806539 on Build GRCh38
rs745314355 Chr.12: 57806548-57806548 on Build GRCh38

View Full Product Details