Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55360674-55361194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM71E2
Entrez Gene ID
284418
Gene Name
family with sequence similarity 71 member E2
Gene Aliases
C19orf16
UniGene
Hs.528319
Gene Chromosome Location
Chr.19: 55354908-55363252 on Build GRCh38
Transcripts
NM_001145402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1612845 c.713C>T p.S238L Chr.19: 55361045-55361045 on Build GRCh38
1001493 c.756G>T p.K252N Chr.19: 55361002-55361002 on Build GRCh38
4411312 c.803G>A p.R268H Chr.19: 55360861-55360861 on Build GRCh38
3389395 c.694T>A p.S232T Chr.19: 55361064-55361064 on Build GRCh38
5697516 c.758+3G>A p.? Chr.19: 55360997-55360997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750049835 Chr.19: 55360698-55360698 on Build GRCh38
rs545227943 Chr.19: 55361030-55361030 on Build GRCh38
rs556971271 Chr.19: 55360977-55360977 on Build GRCh38
rs961708545 Chr.19: 55360811-55360811 on Build GRCh38
rs903239328 Chr.19: 55361026-55361026 on Build GRCh38
rs530006264 Chr.19: 55360709-55360709 on Build GRCh38
rs1043533047 Chr.19: 55361010-55361010 on Build GRCh38
rs1027797482 Chr.19: 55361125-55361126 on Build GRCh38
rs746278294 Chr.19: 55361130-55361131 on Build GRCh38
rs960935602 Chr.19: 55360752-55360752 on Build GRCh38
rs907589486 Chr.19: 55360801-55360801 on Build GRCh38
rs199736290 Chr.19: 55361066-55361066 on Build GRCh38
rs1017817404 Chr.19: 55360888-55360889 on Build GRCh38
rs1053124643 Chr.19: 55361045-55361045 on Build GRCh38
rs544399847 Chr.19: 55361068-55361068 on Build GRCh38
rs946535170 Chr.19: 55361007-55361007 on Build GRCh38
rs984510874 Chr.19: 55360786-55360786 on Build GRCh38
rs576407381 Chr.19: 55361162-55361162 on Build GRCh38
rs1023927360 Chr.19: 55360775-55360775 on Build GRCh38
rs781413847 Chr.19: 55361074-55361074 on Build GRCh38
rs1013025647 Chr.19: 55360763-55360763 on Build GRCh38
rs181946545 Chr.19: 55360888-55360888 on Build GRCh38
rs973209343 Chr.19: 55360889-55360889 on Build GRCh38
rs977109883 Chr.19: 55360894-55360894 on Build GRCh38
rs775900006 Chr.19: 55361148-55361149 on Build GRCh38
rs754884224 Chr.19: 55360780-55360780 on Build GRCh38
rs375100584 Chr.19: 55361164-55361164 on Build GRCh38
rs1054947400 Chr.19: 55360968-55360968 on Build GRCh38
rs536758422 Chr.19: 55360905-55360905 on Build GRCh38
rs149167516 Chr.19: 55361108-55361108 on Build GRCh38
rs781189798 Chr.19: 55360848-55360848 on Build GRCh38
rs56109413 Chr.19: 55360978-55360978 on Build GRCh38
rs532687400 Chr.19: 55361166-55361166 on Build GRCh38
rs540209151 Chr.19: 55361120-55361120 on Build GRCh38
rs760088753 Chr.19: 55360994-55360994 on Build GRCh38
rs759045596 Chr.19: 55361022-55361022 on Build GRCh38
rs939300682 Chr.19: 55360943-55360943 on Build GRCh38
rs954961216 Chr.19: 55360827-55360827 on Build GRCh38
rs897799 Chr.19: 55360823-55360823 on Build GRCh38
rs745969178 Chr.19: 55361083-55361083 on Build GRCh38
rs368182481 Chr.19: 55361044-55361044 on Build GRCh38
rs774826823 Chr.19: 55360936-55360936 on Build GRCh38
rs537071202 Chr.19: 55361125-55361125 on Build GRCh38
rs939246955 Chr.19: 55360720-55360720 on Build GRCh38
rs986797326 Chr.19: 55361171-55361171 on Build GRCh38
rs747232835 Chr.19: 55360840-55360840 on Build GRCh38
rs961800541 Chr.19: 55361126-55361126 on Build GRCh38
rs1012146614 Chr.19: 55360773-55360773 on Build GRCh38
rs752177095 Chr.19: 55360983-55360983 on Build GRCh38
rs748478533 Chr.19: 55361138-55361138 on Build GRCh38
rs560947882 Chr.19: 55360695-55360695 on Build GRCh38
rs558481472 Chr.19: 55361040-55361040 on Build GRCh38
rs770519796 Chr.19: 55361146-55361146 on Build GRCh38
rs370067875 Chr.19: 55360745-55360745 on Build GRCh38
rs777519348 Chr.19: 55360836-55360836 on Build GRCh38
rs920346019 Chr.19: 55360897-55360897 on Build GRCh38
rs373807482 Chr.19: 55360783-55360783 on Build GRCh38
rs1002944674 Chr.19: 55361028-55361028 on Build GRCh38
rs763033600 Chr.19: 55360925-55360925 on Build GRCh38
rs1054897171 Chr.19: 55360733-55360733 on Build GRCh38
rs145275620 Chr.19: 55361093-55361093 on Build GRCh38
rs745907282 Chr.19: 55360883-55360883 on Build GRCh38
rs375497462 Chr.19: 55360921-55360921 on Build GRCh38
rs752891493 Chr.19: 55360719-55360719 on Build GRCh38
rs1009126954 Chr.19: 55360862-55360862 on Build GRCh38
rs538878452 Chr.19: 55360785-55360785 on Build GRCh38
rs947502690 Chr.19: 55360993-55360993 on Build GRCh38
rs780260883 Chr.19: 55361094-55361094 on Build GRCh38
rs770331885 Chr.19: 55361145-55361148 on Build GRCh38
rs575349061 Chr.19: 55361058-55361058 on Build GRCh38
rs774591011 Chr.19: 55361125-55361125 on Build GRCh38
rs932251351 Chr.19: 55360892-55360892 on Build GRCh38
rs370353415 Chr.19: 55360715-55360715 on Build GRCh38
rs2124918 Chr.19: 55361029-55361029 on Build GRCh38
rs34051292 Chr.19: 55361145-55361145 on Build GRCh38
rs11084397 Chr.19: 55361163-55361163 on Build GRCh38
rs1023517181 Chr.19: 55361073-55361073 on Build GRCh38
rs1002851114 Chr.19: 55360723-55360723 on Build GRCh38
rs770432748 Chr.19: 55360884-55360884 on Build GRCh38
rs773173127 Chr.19: 55360861-55360861 on Build GRCh38
rs927603589 Chr.19: 55360940-55360940 on Build GRCh38
rs536220038 Chr.19: 55360986-55360986 on Build GRCh38
rs185276069 Chr.19: 55361025-55361025 on Build GRCh38
rs369460300 Chr.19: 55361167-55361167 on Build GRCh38
rs771182591 Chr.19: 55360844-55360844 on Build GRCh38
rs1034957575 Chr.19: 55360744-55360744 on Build GRCh38

View Full Product Details