Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24191105-24191512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM228A
Entrez Gene ID
653140
Gene Name
family with sequence similarity 228 member A
Gene Aliases
C2orf84
UniGene
Hs.741736
Gene Chromosome Location
Chr.2: 24175043-24191698 on Build GRCh38
Transcripts
NM_001040710.2

SNP Detail

SNP ID Chromosome Location
rs888082158 Chr.2: 24191191-24191191 on Build GRCh38
rs189945749 Chr.2: 24191429-24191429 on Build GRCh38
rs543082149 Chr.2: 24191284-24191284 on Build GRCh38
rs893085371 Chr.2: 24191485-24191485 on Build GRCh38
rs941066750 Chr.2: 24191193-24191193 on Build GRCh38
rs532832096 Chr.2: 24191273-24191273 on Build GRCh38
rs920673786 Chr.2: 24191354-24191354 on Build GRCh38
rs1033337437 Chr.2: 24191428-24191428 on Build GRCh38
rs996511070 Chr.2: 24191272-24191272 on Build GRCh38
rs941022250 Chr.2: 24191373-24191373 on Build GRCh38
rs140657173 Chr.2: 24191258-24191258 on Build GRCh38
rs930174294 Chr.2: 24191326-24191326 on Build GRCh38
rs532324876 Chr.2: 24191421-24191421 on Build GRCh38
rs1039453266 Chr.2: 24191194-24191194 on Build GRCh38
rs997942168 Chr.2: 24191218-24191218 on Build GRCh38
rs575044654 Chr.2: 24191188-24191188 on Build GRCh38
rs928113998 Chr.2: 24191309-24191309 on Build GRCh38
rs1045627972 Chr.2: 24191408-24191408 on Build GRCh38
rs1047329748 Chr.2: 24191189-24191189 on Build GRCh38
rs996573704 Chr.2: 24191481-24191481 on Build GRCh38
rs899502071 Chr.2: 24191401-24191401 on Build GRCh38
rs989532999 Chr.2: 24191283-24191283 on Build GRCh38
rs892837485 Chr.2: 24191271-24191271 on Build GRCh38
rs1039353096 Chr.2: 24191391-24191391 on Build GRCh38
rs915881822 Chr.2: 24191397-24191397 on Build GRCh38
rs933703682 Chr.2: 24191407-24191407 on Build GRCh38
rs542432030 Chr.2: 24191203-24191203 on Build GRCh38
rs1049873563 Chr.2: 24191487-24191487 on Build GRCh38
rs7564420 Chr.2: 24191133-24191133 on Build GRCh38
rs910159373 Chr.2: 24191369-24191369 on Build GRCh38
rs1054187769 Chr.2: 24191268-24191268 on Build GRCh38
rs537253879 Chr.2: 24191489-24191489 on Build GRCh38
rs1009119987 Chr.2: 24191292-24191292 on Build GRCh38
rs564009120 Chr.2: 24191451-24191452 on Build GRCh38
rs866128217 Chr.2: 24191289-24191289 on Build GRCh38
rs765945785 Chr.2: 24191287-24191287 on Build GRCh38
rs772976621 Chr.2: 24191247-24191247 on Build GRCh38
rs535291867 Chr.2: 24191140-24191140 on Build GRCh38
rs892893805 Chr.2: 24191446-24191446 on Build GRCh38
rs972149266 Chr.2: 24191333-24191333 on Build GRCh38
rs547625655 Chr.2: 24191422-24191422 on Build GRCh38
rs1050843492 Chr.2: 24191412-24191412 on Build GRCh38
rs878870951 Chr.2: 24191341-24191341 on Build GRCh38
rs113331166 Chr.2: 24191304-24191304 on Build GRCh38
rs991903449 Chr.2: 24191395-24191395 on Build GRCh38
rs192791758 Chr.2: 24191262-24191262 on Build GRCh38
rs958259242 Chr.2: 24191275-24191275 on Build GRCh38
rs930630216 Chr.2: 24191150-24191150 on Build GRCh38
rs371349880 Chr.2: 24191132-24191132 on Build GRCh38
rs184252996 Chr.2: 24191363-24191363 on Build GRCh38
rs564093433 Chr.2: 24191230-24191230 on Build GRCh38
rs998721292 Chr.2: 24191427-24191427 on Build GRCh38
rs899623142 Chr.2: 24191213-24191213 on Build GRCh38
rs903035396 Chr.2: 24191242-24191242 on Build GRCh38
rs1029461793 Chr.2: 24191219-24191219 on Build GRCh38
rs540796984 Chr.2: 24191217-24191217 on Build GRCh38

View Full Product Details