Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37928830-37929325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN2
Entrez Gene ID
151393
Gene Name
regulator of microtubule dynamics 2
Gene Aliases
BLOCK18, FAM82A, FAM82A1, PRO34163, PYST9371, RMD-2, RMD2, RMD4
UniGene
Hs.591566
Gene Chromosome Location
Chr.2: 37920789-38068032 on Build GRCh38
Transcripts
NM_001170791.2 NM_001170792.2 NM_001322211.1 NM_001322212.1 XM_011532616.2 XM_011532617.2 XM_011532618.2 XM_011532619.2 XM_017003476.1 XM_017003477.1 XM_017003478.1 XM_017003479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4417170 c.25T>C p.L9L Chr.2: 37929302-37929302 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754222747 Chr.2: 37928879-37928879 on Build GRCh38
rs999037614 Chr.2: 37928892-37928892 on Build GRCh38
rs746337421 Chr.2: 37928897-37928897 on Build GRCh38
rs779512212 Chr.2: 37928895-37928895 on Build GRCh38
rs1015232938 Chr.2: 37928965-37928965 on Build GRCh38
rs185097292 Chr.2: 37929084-37929084 on Build GRCh38
rs778361466 Chr.2: 37929021-37929021 on Build GRCh38
rs889435080 Chr.2: 37929290-37929290 on Build GRCh38
rs961210897 Chr.2: 37929012-37929012 on Build GRCh38
rs769009514 Chr.2: 37929063-37929063 on Build GRCh38
rs1041739523 Chr.2: 37928851-37928851 on Build GRCh38
rs971174856 Chr.2: 37928939-37928939 on Build GRCh38
rs144614322 Chr.2: 37929283-37929283 on Build GRCh38
rs1008134975 Chr.2: 37928995-37928995 on Build GRCh38
rs1011329825 Chr.2: 37929277-37929277 on Build GRCh38
rs982434392 Chr.2: 37928944-37928944 on Build GRCh38
rs762745631 Chr.2: 37929092-37929092 on Build GRCh38
rs961510088 Chr.2: 37929304-37929304 on Build GRCh38
rs770821121 Chr.2: 37929101-37929101 on Build GRCh38
rs932465652 Chr.2: 37929043-37929043 on Build GRCh38
rs1028641466 Chr.2: 37929230-37929230 on Build GRCh38
rs772249199 Chr.2: 37928977-37928977 on Build GRCh38
rs1046614290 Chr.2: 37929126-37929126 on Build GRCh38
rs553434240 Chr.2: 37928874-37928874 on Build GRCh38
rs557983857 Chr.2: 37928947-37928947 on Build GRCh38
rs953976294 Chr.2: 37928992-37928992 on Build GRCh38
rs892807028 Chr.2: 37928921-37928921 on Build GRCh38
rs34303008 Chr.2: 37929057-37929057 on Build GRCh38
rs547955051 Chr.2: 37928925-37928925 on Build GRCh38
rs541004808 Chr.2: 37929082-37929082 on Build GRCh38
rs1049449941 Chr.2: 37929288-37929288 on Build GRCh38
rs776948419 Chr.2: 37929079-37929079 on Build GRCh38
rs954027776 Chr.2: 37929232-37929232 on Build GRCh38
rs1019481362 Chr.2: 37929009-37929009 on Build GRCh38
rs767921451 Chr.2: 37929259-37929259 on Build GRCh38
rs138508519 Chr.2: 37929294-37929294 on Build GRCh38
rs1053289947 Chr.2: 37929212-37929212 on Build GRCh38
rs7565287 Chr.2: 37928990-37928990 on Build GRCh38
rs756465933 Chr.2: 37929265-37929265 on Build GRCh38
rs769413414 Chr.2: 37929071-37929071 on Build GRCh38
rs199894296 Chr.2: 37928997-37928998 on Build GRCh38
rs61743792 Chr.2: 37929302-37929302 on Build GRCh38
rs949636966 Chr.2: 37928876-37928876 on Build GRCh38
rs768537563 Chr.2: 37929139-37929139 on Build GRCh38
rs373130579 Chr.2: 37929266-37929266 on Build GRCh38
rs1015372124 Chr.2: 37929208-37929208 on Build GRCh38
rs561789693 Chr.2: 37929062-37929062 on Build GRCh38
rs1045964675 Chr.2: 37929111-37929111 on Build GRCh38
rs533442979 Chr.2: 37929140-37929140 on Build GRCh38
rs907375843 Chr.2: 37929134-37929134 on Build GRCh38
rs774399095 Chr.2: 37929214-37929214 on Build GRCh38
rs540183952 Chr.2: 37928881-37928881 on Build GRCh38
rs921205844 Chr.2: 37929002-37929005 on Build GRCh38
rs529002953 Chr.2: 37929069-37929069 on Build GRCh38
rs903438298 Chr.2: 37928862-37928862 on Build GRCh38
rs986675717 Chr.2: 37929048-37929048 on Build GRCh38
rs924123174 Chr.2: 37929128-37929128 on Build GRCh38
rs546161992 Chr.2: 37928940-37928940 on Build GRCh38
rs1013497780 Chr.2: 37929103-37929103 on Build GRCh38
rs551609114 Chr.2: 37929264-37929264 on Build GRCh38
rs935486298 Chr.2: 37929200-37929200 on Build GRCh38
rs995213877 Chr.2: 37929218-37929218 on Build GRCh38
rs562677646 Chr.2: 37929095-37929095 on Build GRCh38

View Full Product Details