Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197757008-197757451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOLL
Entrez Gene ID
66037
Gene Name
boule homolog, RNA binding protein
Gene Aliases
BOULE
UniGene
Hs.169797
Gene Chromosome Location
Chr.2: 197726879-197786574 on Build GRCh38
Transcripts
NM_001284358.1 NM_001284361.1 NM_001284362.1 NM_033030.5 NM_197970.2 XM_006712715.3 XM_011511692.2 XM_011511693.2 XM_011511694.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4518399 c.636_636+1GG>AA p.? Chr.2: 197757352-197757353 on Build GRCh38
4089992 c.595A>G p.T199A Chr.2: 197757394-197757394 on Build GRCh38
4518400 c.600_600+1GG>AA p.? Chr.2: 197757352-197757353 on Build GRCh38
4089993 c.559A>G p.T187A Chr.2: 197757394-197757394 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs933056889 Chr.2: 197757282-197757282 on Build GRCh38
rs977583317 Chr.2: 197757259-197757259 on Build GRCh38
rs1035664583 Chr.2: 197757082-197757082 on Build GRCh38
rs543400031 Chr.2: 197757170-197757170 on Build GRCh38
rs865850670 Chr.2: 197757361-197757361 on Build GRCh38
rs929104472 Chr.2: 197757398-197757398 on Build GRCh38
rs960377408 Chr.2: 197757225-197757225 on Build GRCh38
rs920539460 Chr.2: 197757356-197757356 on Build GRCh38
rs147772767 Chr.2: 197757367-197757367 on Build GRCh38
rs774452872 Chr.2: 197757323-197757323 on Build GRCh38
rs991756202 Chr.2: 197757122-197757122 on Build GRCh38
rs529827573 Chr.2: 197757096-197757096 on Build GRCh38
rs887212415 Chr.2: 197757065-197757065 on Build GRCh38
rs1003385175 Chr.2: 197757067-197757067 on Build GRCh38
rs768764288 Chr.2: 197757319-197757319 on Build GRCh38
rs753230341 Chr.2: 197757412-197757412 on Build GRCh38
rs765727894 Chr.2: 197757408-197757408 on Build GRCh38
rs777017173 Chr.2: 197757382-197757382 on Build GRCh38
rs1001925479 Chr.2: 197757199-197757199 on Build GRCh38
rs563296231 Chr.2: 197757189-197757189 on Build GRCh38
rs1020605390 Chr.2: 197757280-197757280 on Build GRCh38
rs758886633 Chr.2: 197757417-197757417 on Build GRCh38
rs749062588 Chr.2: 197757304-197757304 on Build GRCh38
rs1047570451 Chr.2: 197757399-197757399 on Build GRCh38
rs1038188518 Chr.2: 197757142-197757142 on Build GRCh38
rs138957785 Chr.2: 197757325-197757325 on Build GRCh38
rs1013215490 Chr.2: 197757258-197757258 on Build GRCh38
rs772048628 Chr.2: 197757335-197757335 on Build GRCh38
rs551659947 Chr.2: 197757413-197757417 on Build GRCh38
rs1037613473 Chr.2: 197757327-197757327 on Build GRCh38
rs1004162229 Chr.2: 197757127-197757127 on Build GRCh38
rs987636344 Chr.2: 197757292-197757292 on Build GRCh38
rs773284900 Chr.2: 197757350-197757350 on Build GRCh38
rs966913018 Chr.2: 197757281-197757281 on Build GRCh38
rs898356139 Chr.2: 197757177-197757177 on Build GRCh38
rs760862723 Chr.2: 197757357-197757357 on Build GRCh38
rs910180295 Chr.2: 197757301-197757301 on Build GRCh38
rs1033849342 Chr.2: 197757222-197757222 on Build GRCh38
rs375232587 Chr.2: 197757395-197757395 on Build GRCh38
rs199838771 Chr.2: 197757385-197757385 on Build GRCh38

View Full Product Details