Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35627209-35627563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE1
Entrez Gene ID
8904
Gene Name
copine 1
Gene Aliases
COPN1, CPN1
UniGene
Hs.246413
Gene Chromosome Location
Chr.20: 35626031-35664956 on Build GRCh38
Transcripts
NM_001198863.1 NM_003915.5 NM_152925.2 NM_152926.2 NM_152927.2 NM_152928.2 NR_037188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
280106 c.1165C>T p.R389C Chr.20: 35627366-35627366 on Build GRCh38
280105 c.1250C>T p.S417L Chr.20: 35627281-35627281 on Build GRCh38
7238317 c.1130T>G p.I377S Chr.20: 35627401-35627401 on Build GRCh38
4602868 c.1219_1220insT p.A407fs*21 Chr.20: 35627311-35627312 on Build GRCh38
5446387 c.1215_1216insT p.A407fs*21 Chr.20: 35627315-35627316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767313284 Chr.20: 35627438-35627438 on Build GRCh38
rs746060459 Chr.20: 35627386-35627386 on Build GRCh38
rs752476634 Chr.20: 35627451-35627451 on Build GRCh38
rs147762472 Chr.20: 35627281-35627281 on Build GRCh38
rs183483257 Chr.20: 35627525-35627525 on Build GRCh38
rs775874171 Chr.20: 35627285-35627285 on Build GRCh38
rs753375834 Chr.20: 35627280-35627280 on Build GRCh38
rs907772329 Chr.20: 35627347-35627347 on Build GRCh38
rs906174777 Chr.20: 35627302-35627310 on Build GRCh38
rs762381858 Chr.20: 35627326-35627326 on Build GRCh38
rs757272967 Chr.20: 35627373-35627373 on Build GRCh38
rs759299015 Chr.20: 35627349-35627349 on Build GRCh38
rs199971132 Chr.20: 35627422-35627422 on Build GRCh38
rs767040898 Chr.20: 35627249-35627250 on Build GRCh38
rs765969455 Chr.20: 35627334-35627334 on Build GRCh38
rs140353700 Chr.20: 35627386-35627387 on Build GRCh38
rs991658041 Chr.20: 35627355-35627355 on Build GRCh38
rs749361503 Chr.20: 35627252-35627252 on Build GRCh38
rs772421629 Chr.20: 35627284-35627284 on Build GRCh38
rs563028591 Chr.20: 35627523-35627523 on Build GRCh38
rs150317541 Chr.20: 35627306-35627306 on Build GRCh38
rs1046884193 Chr.20: 35627376-35627376 on Build GRCh38
rs1018579147 Chr.20: 35627468-35627468 on Build GRCh38
rs983492743 Chr.20: 35627533-35627533 on Build GRCh38
rs976952231 Chr.20: 35627505-35627505 on Build GRCh38
rs921351437 Chr.20: 35627461-35627461 on Build GRCh38
rs73905908 Chr.20: 35627300-35627300 on Build GRCh38
rs140170187 Chr.20: 35627528-35627528 on Build GRCh38
rs771120968 Chr.20: 35627270-35627270 on Build GRCh38
rs964553536 Chr.20: 35627470-35627470 on Build GRCh38
rs747337175 Chr.20: 35627417-35627417 on Build GRCh38
rs199904497 Chr.20: 35627370-35627370 on Build GRCh38
rs76294482 Chr.20: 35627315-35627315 on Build GRCh38
rs772710552 Chr.20: 35627320-35627320 on Build GRCh38
rs566420424 Chr.20: 35627537-35627537 on Build GRCh38
rs544852809 Chr.20: 35627491-35627491 on Build GRCh38
rs756058775 Chr.20: 35627236-35627236 on Build GRCh38
rs1011146642 Chr.20: 35627453-35627453 on Build GRCh38
rs534828145 Chr.20: 35627466-35627466 on Build GRCh38
rs769147657 Chr.20: 35627418-35627418 on Build GRCh38
rs138915422 Chr.20: 35627359-35627359 on Build GRCh38
rs770311231 Chr.20: 35627425-35627425 on Build GRCh38
rs1003706752 Chr.20: 35627303-35627303 on Build GRCh38
rs1008559415 Chr.20: 35627481-35627481 on Build GRCh38
rs974898227 Chr.20: 35627286-35627286 on Build GRCh38
rs751250213 Chr.20: 35627343-35627343 on Build GRCh38
rs148978209 Chr.20: 35627365-35627365 on Build GRCh38
rs746401145 Chr.20: 35627254-35627254 on Build GRCh38
rs201885779 Chr.20: 35627431-35627431 on Build GRCh38
rs147019139 Chr.20: 35627312-35627313 on Build GRCh38
rs779032581 Chr.20: 35627379-35627379 on Build GRCh38
rs111565842 Chr.20: 35627282-35627282 on Build GRCh38
rs759219394 Chr.20: 35627437-35627437 on Build GRCh38
rs777865462 Chr.20: 35627249-35627249 on Build GRCh38
rs562185178 Chr.20: 35627424-35627424 on Build GRCh38
rs753246224 Chr.20: 35627504-35627504 on Build GRCh38
rs760386348 Chr.20: 35627319-35627320 on Build GRCh38
rs371661762 Chr.20: 35627464-35627464 on Build GRCh38
rs573166041 Chr.20: 35627412-35627412 on Build GRCh38
rs772303395 Chr.20: 35627387-35627387 on Build GRCh38
rs374726269 Chr.20: 35627483-35627483 on Build GRCh38
rs201918169 Chr.20: 35627366-35627366 on Build GRCh38

View Full Product Details