Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15211999-15212589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN7
Entrez Gene ID
23473
Gene Name
calpain 7
Gene Aliases
CALPAIN7, PALBH
UniGene
Hs.595234
Gene Chromosome Location
Chr.3: 15206226-15252916 on Build GRCh38
Transcripts
NM_014296.2 XM_011533555.2 XM_011533556.2 XM_011533557.2 XM_017006043.1 XM_017006044.1 XM_017006045.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6451638 c.111A>G p.A37A Chr.3: 15212112-15212112 on Build GRCh38
5398436 c.124T>C p.Y42H Chr.3: 15212125-15212125 on Build GRCh38
382272 c.109G>A p.A37T Chr.3: 15212110-15212110 on Build GRCh38
5994509 c.175G>A p.E59K Chr.3: 15212176-15212176 on Build GRCh38
1040084 c.129T>C p.A43A Chr.3: 15212130-15212130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751797792 Chr.3: 15212322-15212322 on Build GRCh38
rs759459747 Chr.3: 15212137-15212137 on Build GRCh38
rs565083887 Chr.3: 15212310-15212310 on Build GRCh38
rs748864995 Chr.3: 15212117-15212117 on Build GRCh38
rs371561470 Chr.3: 15212101-15212101 on Build GRCh38
rs188046770 Chr.3: 15212037-15212037 on Build GRCh38
rs545290312 Chr.3: 15212293-15212293 on Build GRCh38
rs147308439 Chr.3: 15212062-15212062 on Build GRCh38
rs764545432 Chr.3: 15212175-15212175 on Build GRCh38
rs755258990 Chr.3: 15212218-15212218 on Build GRCh38
rs758889233 Chr.3: 15212091-15212091 on Build GRCh38
rs891029409 Chr.3: 15212564-15212564 on Build GRCh38
rs955080030 Chr.3: 15212300-15212300 on Build GRCh38
rs528418015 Chr.3: 15212268-15212268 on Build GRCh38
rs745322596 Chr.3: 15212237-15212237 on Build GRCh38
rs942120526 Chr.3: 15212438-15212438 on Build GRCh38
rs898838354 Chr.3: 15212272-15212272 on Build GRCh38
rs770400520 Chr.3: 15212126-15212126 on Build GRCh38
rs569202272 Chr.3: 15212494-15212494 on Build GRCh38
rs538093069 Chr.3: 15212539-15212539 on Build GRCh38
rs780563309 Chr.3: 15212099-15212099 on Build GRCh38
rs375747480 Chr.3: 15212092-15212092 on Build GRCh38
rs186107227 Chr.3: 15212256-15212256 on Build GRCh38
rs144974373 Chr.3: 15212465-15212465 on Build GRCh38
rs898297911 Chr.3: 15212261-15212261 on Build GRCh38
rs780651270 Chr.3: 15212210-15212210 on Build GRCh38
rs574868976 Chr.3: 15212220-15212220 on Build GRCh38
rs938694898 Chr.3: 15212423-15212423 on Build GRCh38
rs754344102 Chr.3: 15212192-15212204 on Build GRCh38
rs1028201232 Chr.3: 15212276-15212276 on Build GRCh38
rs994489484 Chr.3: 15212274-15212274 on Build GRCh38
rs764052756 Chr.3: 15212415-15212415 on Build GRCh38
rs558776718 Chr.3: 15212485-15212485 on Build GRCh38
rs751035768 Chr.3: 15212207-15212207 on Build GRCh38
rs761271512 Chr.3: 15212175-15212176 on Build GRCh38
rs769501079 Chr.3: 15212110-15212110 on Build GRCh38
rs775365951 Chr.3: 15212163-15212163 on Build GRCh38
rs911723949 Chr.3: 15212251-15212251 on Build GRCh38
rs760953519 Chr.3: 15212066-15212066 on Build GRCh38
rs753975969 Chr.3: 15212188-15212188 on Build GRCh38
rs764161167 Chr.3: 15212069-15212069 on Build GRCh38
rs376932100 Chr.3: 15212138-15212138 on Build GRCh38
rs1031411800 Chr.3: 15212338-15212338 on Build GRCh38
rs762924447 Chr.3: 15212409-15212409 on Build GRCh38
rs372827755 Chr.3: 15212059-15212059 on Build GRCh38
rs991739983 Chr.3: 15212076-15212076 on Build GRCh38
rs112637695 Chr.3: 15212413-15212413 on Build GRCh38
rs753969497 Chr.3: 15212073-15212073 on Build GRCh38
rs776967562 Chr.3: 15212264-15212264 on Build GRCh38
rs757244189 Chr.3: 15212083-15212083 on Build GRCh38
rs938663568 Chr.3: 15212159-15212159 on Build GRCh38
rs538954284 Chr.3: 15212483-15212483 on Build GRCh38
rs978699697 Chr.3: 15212418-15212418 on Build GRCh38
rs369177764 Chr.3: 15212243-15212243 on Build GRCh38
rs538916968 Chr.3: 15212032-15212032 on Build GRCh38
rs924607538 Chr.3: 15212450-15212450 on Build GRCh38
rs1008075527 Chr.3: 15212565-15212565 on Build GRCh38
rs779603414 Chr.3: 15212085-15212085 on Build GRCh38
rs750942548 Chr.3: 15212087-15212087 on Build GRCh38
rs767900871 Chr.3: 15212472-15212472 on Build GRCh38
rs955808801 Chr.3: 15212356-15212356 on Build GRCh38
rs943187950 Chr.3: 15212537-15212537 on Build GRCh38
rs762001265 Chr.3: 15212196-15212196 on Build GRCh38
rs751977326 Chr.3: 15212211-15212211 on Build GRCh38
rs375465234 Chr.3: 15212426-15212426 on Build GRCh38
rs765392901 Chr.3: 15212202-15212202 on Build GRCh38
rs772015323 Chr.3: 15212242-15212242 on Build GRCh38
rs530677612 Chr.3: 15212333-15212333 on Build GRCh38
rs760334490 Chr.3: 15212170-15212170 on Build GRCh38
rs748874148 Chr.3: 15212223-15212223 on Build GRCh38
rs540304739 Chr.3: 15212230-15212230 on Build GRCh38
rs911741386 Chr.3: 15212508-15212508 on Build GRCh38
rs374625151 Chr.3: 15212402-15212402 on Build GRCh38
rs778397013 Chr.3: 15212235-15212235 on Build GRCh38
rs907849354 Chr.3: 15212429-15212429 on Build GRCh38
rs192483325 Chr.3: 15212404-15212404 on Build GRCh38
rs77346929 Chr.3: 15212376-15212376 on Build GRCh38
rs180855962 Chr.3: 15212065-15212065 on Build GRCh38
rs1035886632 Chr.3: 15212330-15212330 on Build GRCh38
rs978835488 Chr.3: 15212191-15212191 on Build GRCh38
rs996688499 Chr.3: 15212525-15212525 on Build GRCh38
rs1047513124 Chr.3: 15212275-15212275 on Build GRCh38
rs777242348 Chr.3: 15212113-15212113 on Build GRCh38
rs914733937 Chr.3: 15212382-15212382 on Build GRCh38
rs758981120 Chr.3: 15212209-15212209 on Build GRCh38
rs764404005 Chr.3: 15212176-15212176 on Build GRCh38
rs774324879 Chr.3: 15212135-15212135 on Build GRCh38
rs987498309 Chr.3: 15212493-15212498 on Build GRCh38

View Full Product Details