Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 45084814-45085300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCP1
Entrez Gene ID
64866
Gene Name
CUB domain containing protein 1
Gene Aliases
CD318, SIMA135, TRASK
UniGene
Hs.476093
Gene Chromosome Location
Chr.3: 45082274-45146422 on Build GRCh38
Transcripts
NM_022842.4 XM_011534024.1 XM_017007070.1

SNP Detail

SNP ID Chromosome Location
rs577175008 Chr.3: 45085139-45085139 on Build GRCh38
rs913798775 Chr.3: 45085038-45085038 on Build GRCh38
rs979320988 Chr.3: 45085095-45085095 on Build GRCh38
rs896128089 Chr.3: 45084888-45084888 on Build GRCh38
rs1035612021 Chr.3: 45085086-45085086 on Build GRCh38
rs905042541 Chr.3: 45084846-45084846 on Build GRCh38
rs886175455 Chr.3: 45084983-45084983 on Build GRCh38
rs997047425 Chr.3: 45085267-45085267 on Build GRCh38
rs11721011 Chr.3: 45084912-45084912 on Build GRCh38
rs534865221 Chr.3: 45085135-45085135 on Build GRCh38
rs546949998 Chr.3: 45084852-45084852 on Build GRCh38
rs569863776 Chr.3: 45085046-45085046 on Build GRCh38
rs917713967 Chr.3: 45085160-45085160 on Build GRCh38
rs1056599575 Chr.3: 45085153-45085153 on Build GRCh38
rs141071877 Chr.3: 45084940-45084940 on Build GRCh38
rs1017077438 Chr.3: 45084965-45084965 on Build GRCh38
rs887392928 Chr.3: 45084924-45084924 on Build GRCh38
rs150299181 Chr.3: 45085244-45085244 on Build GRCh38
rs1005785689 Chr.3: 45085222-45085222 on Build GRCh38
rs955536255 Chr.3: 45085012-45085012 on Build GRCh38
rs1056234132 Chr.3: 45084853-45084853 on Build GRCh38
rs745513614 Chr.3: 45084922-45084922 on Build GRCh38
rs768436653 Chr.3: 45084994-45084994 on Build GRCh38
rs1038647823 Chr.3: 45085239-45085239 on Build GRCh38
rs926433073 Chr.3: 45085103-45085103 on Build GRCh38
rs887447053 Chr.3: 45085218-45085218 on Build GRCh38
rs979481771 Chr.3: 45085223-45085223 on Build GRCh38
rs776713013 Chr.3: 45085032-45085032 on Build GRCh38
rs190599509 Chr.3: 45084939-45084939 on Build GRCh38
rs2176804 Chr.3: 45085066-45085066 on Build GRCh38
rs746551149 Chr.3: 45084980-45084980 on Build GRCh38
rs779408049 Chr.3: 45084944-45084944 on Build GRCh38
rs1047359623 Chr.3: 45085182-45085182 on Build GRCh38
rs1001860858 Chr.3: 45085077-45085077 on Build GRCh38
rs928957020 Chr.3: 45085162-45085162 on Build GRCh38
rs547964853 Chr.3: 45084990-45084990 on Build GRCh38
rs1030155047 Chr.3: 45085270-45085270 on Build GRCh38
rs992148486 Chr.3: 45085128-45085128 on Build GRCh38
rs558912501 Chr.3: 45085121-45085121 on Build GRCh38

View Full Product Details