Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129034090-129034364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCC1
Entrez Gene ID
79825
Gene Name
EF-hand and coiled-coil domain containing 1
Gene Aliases
C3orf73, CCDC48
UniGene
Hs.134807
Gene Chromosome Location
Chr.3: 129001604-129040742 on Build GRCh38
Transcripts
NM_024768.2 XM_005247781.3 XM_011513161.2 XM_011513164.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
419848 c.1311C>T p.L437L Chr.3: 129034191-129034191 on Build GRCh38
5007922 c.7_8insA p.Y3fs*1 Chr.3: 129034198-129034199 on Build GRCh38
6572557 c.1382C>G p.A461G Chr.3: 129034262-129034262 on Build GRCh38
1136932 c.1314G>T p.M438I Chr.3: 129034194-129034194 on Build GRCh38
1038030 c.21C>T p.F7F Chr.3: 129034212-129034212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545034238 Chr.3: 129034300-129034300 on Build GRCh38
rs376654866 Chr.3: 129034213-129034213 on Build GRCh38
rs891639434 Chr.3: 129034205-129034205 on Build GRCh38
rs200720072 Chr.3: 129034259-129034259 on Build GRCh38
rs556153454 Chr.3: 129034281-129034281 on Build GRCh38
rs368491173 Chr.3: 129034256-129034256 on Build GRCh38
rs746568535 Chr.3: 129034178-129034178 on Build GRCh38
rs187600174 Chr.3: 129034181-129034181 on Build GRCh38
rs929642404 Chr.3: 129034112-129034112 on Build GRCh38
rs779856577 Chr.3: 129034148-129034148 on Build GRCh38
rs749784255 Chr.3: 129034296-129034296 on Build GRCh38
rs141258380 Chr.3: 129034253-129034253 on Build GRCh38
rs753804517 Chr.3: 129034252-129034252 on Build GRCh38
rs751126757 Chr.3: 129034321-129034323 on Build GRCh38
rs767718057 Chr.3: 129034312-129034312 on Build GRCh38
rs754789473 Chr.3: 129034156-129034156 on Build GRCh38
rs1031483363 Chr.3: 129034192-129034192 on Build GRCh38
rs144276832 Chr.3: 129034212-129034212 on Build GRCh38
rs773324903 Chr.3: 129034317-129034317 on Build GRCh38
rs370363130 Chr.3: 129034182-129034182 on Build GRCh38
rs757375509 Chr.3: 129034132-129034132 on Build GRCh38
rs764358388 Chr.3: 129034128-129034128 on Build GRCh38
rs114087327 Chr.3: 129034297-129034297 on Build GRCh38
rs751857150 Chr.3: 129034131-129034131 on Build GRCh38
rs758066207 Chr.3: 129034273-129034273 on Build GRCh38
rs770228337 Chr.3: 129034194-129034204 on Build GRCh38
rs749020763 Chr.3: 129034149-129034149 on Build GRCh38
rs772767834 Chr.3: 129034174-129034174 on Build GRCh38
rs533958203 Chr.3: 129034160-129034160 on Build GRCh38
rs1032274332 Chr.3: 129034166-129034166 on Build GRCh38
rs760767741 Chr.3: 129034330-129034330 on Build GRCh38
rs773007061 Chr.3: 129034315-129034316 on Build GRCh38
rs766291430 Chr.3: 129034250-129034250 on Build GRCh38
rs537941548 Chr.3: 129034249-129034249 on Build GRCh38
rs781541863 Chr.3: 129034142-129034142 on Build GRCh38
rs949151965 Chr.3: 129034121-129034121 on Build GRCh38
rs1044888423 Chr.3: 129034140-129034140 on Build GRCh38
rs62265318 Chr.3: 129034143-129034143 on Build GRCh38
rs567338824 Chr.3: 129034216-129034216 on Build GRCh38
rs146230336 Chr.3: 129034286-129034286 on Build GRCh38
rs955422908 Chr.3: 129034254-129034254 on Build GRCh38
rs776104584 Chr.3: 129034287-129034287 on Build GRCh38
rs932308023 Chr.3: 129034153-129034153 on Build GRCh38
rs765826655 Chr.3: 129034318-129034319 on Build GRCh38
rs148256726 Chr.3: 129034215-129034215 on Build GRCh38
rs758937555 Chr.3: 129034320-129034321 on Build GRCh38
rs1001168171 Chr.3: 129034144-129034144 on Build GRCh38
rs368394630 Chr.3: 129034179-129034179 on Build GRCh38
rs747767048 Chr.3: 129034170-129034170 on Build GRCh38
rs374178406 Chr.3: 129034146-129034146 on Build GRCh38
rs771668553 Chr.3: 129034173-129034173 on Build GRCh38
rs376688096 Chr.3: 129034113-129034113 on Build GRCh38
rs762622256 Chr.3: 129034316-129034317 on Build GRCh38
rs763415935 Chr.3: 129034114-129034114 on Build GRCh38
rs770539263 Chr.3: 129034270-129034270 on Build GRCh38
rs377018804 Chr.3: 129034261-129034261 on Build GRCh38

View Full Product Details