Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170888829-170889285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF5A2
Entrez Gene ID
56648
Gene Name
eukaryotic translation initiation factor 5A2
Gene Aliases
EIF-5A2, eIF5AII
UniGene
Hs.164144
Gene Chromosome Location
Chr.3: 170888415-170908637 on Build GRCh38
Transcripts
NM_020390.5

SNP Detail

SNP ID Chromosome Location
rs978350653 Chr.3: 170889125-170889125 on Build GRCh38
rs994777305 Chr.3: 170888861-170888861 on Build GRCh38
rs1018198421 Chr.3: 170888954-170888954 on Build GRCh38
rs9812696 Chr.3: 170889078-170889078 on Build GRCh38
rs1043036368 Chr.3: 170889183-170889183 on Build GRCh38
rs13058793 Chr.3: 170889047-170889047 on Build GRCh38
rs929035585 Chr.3: 170888990-170888990 on Build GRCh38
rs550272224 Chr.3: 170889184-170889184 on Build GRCh38
rs192130148 Chr.3: 170889072-170889072 on Build GRCh38
rs1055056941 Chr.3: 170889180-170889180 on Build GRCh38
rs964747947 Chr.3: 170888882-170888882 on Build GRCh38
rs557433728 Chr.3: 170889068-170889072 on Build GRCh38
rs67417067 Chr.3: 170889068-170889069 on Build GRCh38
rs774707360 Chr.3: 170888929-170888929 on Build GRCh38
rs55942418 Chr.3: 170889094-170889094 on Build GRCh38
rs9868209 Chr.3: 170889104-170889104 on Build GRCh38
rs983315576 Chr.3: 170889020-170889020 on Build GRCh38
rs9812695 Chr.3: 170889073-170889073 on Build GRCh38
rs376063208 Chr.3: 170889231-170889231 on Build GRCh38
rs774900302 Chr.3: 170889053-170889054 on Build GRCh38
rs955790255 Chr.3: 170888928-170888928 on Build GRCh38
rs998904136 Chr.3: 170889229-170889229 on Build GRCh38
rs920364273 Chr.3: 170888944-170888944 on Build GRCh38
rs990193134 Chr.3: 170889028-170889028 on Build GRCh38
rs761950554 Chr.3: 170889133-170889133 on Build GRCh38
rs973054674 Chr.3: 170888934-170888934 on Build GRCh38
rs776021660 Chr.3: 170889049-170889053 on Build GRCh38
rs1007156492 Chr.3: 170888948-170888948 on Build GRCh38
rs55706439 Chr.3: 170889071-170889072 on Build GRCh38
rs913354876 Chr.3: 170889035-170889035 on Build GRCh38
rs966559022 Chr.3: 170889117-170889117 on Build GRCh38
rs77669645 Chr.3: 170889068-170889073 on Build GRCh38
rs878900806 Chr.3: 170889165-170889165 on Build GRCh38
rs904651005 Chr.3: 170889193-170889193 on Build GRCh38
rs910783619 Chr.3: 170888873-170888873 on Build GRCh38
rs79641958 Chr.3: 170889065-170889065 on Build GRCh38
rs145717207 Chr.3: 170888959-170888959 on Build GRCh38
rs186829504 Chr.3: 170889048-170889048 on Build GRCh38
rs530377741 Chr.3: 170889156-170889156 on Build GRCh38
rs75632659 Chr.3: 170889050-170889050 on Build GRCh38
rs138240866 Chr.3: 170889128-170889128 on Build GRCh38
rs957372055 Chr.3: 170889013-170889013 on Build GRCh38
rs145925544 Chr.3: 170888926-170888926 on Build GRCh38

View Full Product Details