Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 102892393-102892875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CISD2
Entrez Gene ID
493856
Gene Name
CDGSH iron sulfur domain 2
Gene Aliases
ERIS, Miner1, NAF-1, WFS2, ZCD2
UniGene
Hs.444955
Gene Chromosome Location
Chr.4: 102868978-102892807 on Build GRCh38
Transcripts
NM_001008388.4 NM_001100874.2 NR_047513.1 NR_047515.1

Gene Symbol
SLC9B1
Entrez Gene ID
150159
Gene Name
solute carrier family 9 member B1
Gene Aliases
NHA1, NHEDC1
UniGene
Hs.666728
Gene Chromosome Location
Chr.4: 102885048-103019739 on Build GRCh38
Transcripts
NM_001008388.4 NM_001100874.2 NR_047513.1 NR_047515.1

SNP Detail

SNP ID Chromosome Location
rs6823625 Chr.4: 102892437-102892437 on Build GRCh38
rs542313738 Chr.4: 102892776-102892776 on Build GRCh38
rs1035466651 Chr.4: 102892493-102892493 on Build GRCh38
rs985141688 Chr.4: 102892655-102892658 on Build GRCh38
rs534016541 Chr.4: 102892688-102892688 on Build GRCh38
rs964946715 Chr.4: 102892683-102892683 on Build GRCh38
rs576898706 Chr.4: 102892842-102892842 on Build GRCh38
rs923405719 Chr.4: 102892802-102892802 on Build GRCh38
rs559920232 Chr.4: 102892615-102892615 on Build GRCh38
rs968751485 Chr.4: 102892640-102892640 on Build GRCh38
rs538656604 Chr.4: 102892431-102892431 on Build GRCh38
rs561960425 Chr.4: 102892649-102892649 on Build GRCh38
rs1017905595 Chr.4: 102892663-102892663 on Build GRCh38
rs569044261 Chr.4: 102892528-102892528 on Build GRCh38
rs4699032 Chr.4: 102892724-102892724 on Build GRCh38
rs929414795 Chr.4: 102892841-102892841 on Build GRCh38
rs183447777 Chr.4: 102892415-102892415 on Build GRCh38
rs6846762 Chr.4: 102892577-102892577 on Build GRCh38
rs199627415 Chr.4: 102892661-102892661 on Build GRCh38
rs750886516 Chr.4: 102892668-102892668 on Build GRCh38
rs891567971 Chr.4: 102892527-102892527 on Build GRCh38

View Full Product Details