Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49877050-49877570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISP1
Entrez Gene ID
167
Gene Name
cysteine rich secretory protein 1
Gene Aliases
AEGL1, ARP, CRISP-1, HEL-S-57, HSCRISP1D, HSCRISP1G, HUMARP
UniGene
Hs.109620
Gene Chromosome Location
Chr.6: 49834257-49877096 on Build GRCh38
Transcripts
NM_001205220.1

SNP Detail

SNP ID Chromosome Location
rs540981797 Chr.6: 49877253-49877253 on Build GRCh38
rs553357183 Chr.6: 49877181-49877181 on Build GRCh38
rs187891629 Chr.6: 49877525-49877525 on Build GRCh38
rs941160713 Chr.6: 49877287-49877287 on Build GRCh38
rs1020560797 Chr.6: 49877177-49877177 on Build GRCh38
rs890788928 Chr.6: 49877358-49877358 on Build GRCh38
rs183758593 Chr.6: 49877476-49877476 on Build GRCh38
rs941222908 Chr.6: 49877507-49877507 on Build GRCh38
rs917908529 Chr.6: 49877234-49877234 on Build GRCh38
rs531471086 Chr.6: 49877450-49877450 on Build GRCh38
rs909280310 Chr.6: 49877290-49877290 on Build GRCh38
rs996699542 Chr.6: 49877293-49877293 on Build GRCh38
rs1016765879 Chr.6: 49877095-49877095 on Build GRCh38
rs968205747 Chr.6: 49877148-49877148 on Build GRCh38
rs564080725 Chr.6: 49877258-49877258 on Build GRCh38
rs138062946 Chr.6: 49877110-49877110 on Build GRCh38
rs537077423 Chr.6: 49877478-49877478 on Build GRCh38
rs770966121 Chr.6: 49877513-49877513 on Build GRCh38
rs1033951487 Chr.6: 49877431-49877431 on Build GRCh38
rs746492312 Chr.6: 49877243-49877243 on Build GRCh38
rs968216566 Chr.6: 49877403-49877403 on Build GRCh38
rs917880359 Chr.6: 49877506-49877506 on Build GRCh38
rs551319750 Chr.6: 49877473-49877473 on Build GRCh38
rs189088497 Chr.6: 49877454-49877454 on Build GRCh38
rs7767534 Chr.6: 49877239-49877239 on Build GRCh38
rs979683150 Chr.6: 49877164-49877164 on Build GRCh38
rs940684251 Chr.6: 49877080-49877080 on Build GRCh38
rs145748103 Chr.6: 49877118-49877119 on Build GRCh38
rs1044997758 Chr.6: 49877530-49877530 on Build GRCh38
rs926531193 Chr.6: 49877538-49877538 on Build GRCh38
rs1036315135 Chr.6: 49877099-49877099 on Build GRCh38
rs563425021 Chr.6: 49877404-49877404 on Build GRCh38
rs1020662531 Chr.6: 49877365-49877365 on Build GRCh38
rs1000648939 Chr.6: 49877395-49877395 on Build GRCh38
rs550647100 Chr.6: 49877486-49877486 on Build GRCh38
rs887745463 Chr.6: 49877079-49877079 on Build GRCh38
rs543538529 Chr.6: 49877373-49877373 on Build GRCh38
rs1036751386 Chr.6: 49877331-49877331 on Build GRCh38
rs543714385 Chr.6: 49877119-49877119 on Build GRCh38
rs7763631 Chr.6: 49877109-49877109 on Build GRCh38
rs761851435 Chr.6: 49877334-49877336 on Build GRCh38
rs987894601 Chr.6: 49877140-49877140 on Build GRCh38
rs536888587 Chr.6: 49877141-49877141 on Build GRCh38
rs956434828 Chr.6: 49877156-49877156 on Build GRCh38
rs896379327 Chr.6: 49877100-49877100 on Build GRCh38
rs780705595 Chr.6: 49877289-49877289 on Build GRCh38
rs992546543 Chr.6: 49877182-49877182 on Build GRCh38
rs892194097 Chr.6: 49877408-49877408 on Build GRCh38
rs768116297 Chr.6: 49877288-49877288 on Build GRCh38
rs955322218 Chr.6: 49877130-49877130 on Build GRCh38
rs536417959 Chr.6: 49877508-49877508 on Build GRCh38
rs567766727 Chr.6: 49877502-49877502 on Build GRCh38
rs962413669 Chr.6: 49877495-49877495 on Build GRCh38
rs116784129 Chr.6: 49877361-49877361 on Build GRCh38
rs932025224 Chr.6: 49877078-49877078 on Build GRCh38
rs899648395 Chr.6: 49877519-49877519 on Build GRCh38
rs879551935 Chr.6: 49877133-49877134 on Build GRCh38
rs207466942 Chr.6: 49877199-49877199 on Build GRCh38
rs192832093 Chr.6: 49877467-49877467 on Build GRCh38
rs377706287 Chr.6: 49877354-49877354 on Build GRCh38
rs926829063 Chr.6: 49877165-49877165 on Build GRCh38
rs972102289 Chr.6: 49877501-49877501 on Build GRCh38
rs940729464 Chr.6: 49877322-49877322 on Build GRCh38
rs573418584 Chr.6: 49877534-49877534 on Build GRCh38
rs979568446 Chr.6: 49877411-49877411 on Build GRCh38
rs905076837 Chr.6: 49877367-49877367 on Build GRCh38
rs879470758 Chr.6: 49877463-49877463 on Build GRCh38
rs1009254842 Chr.6: 49877423-49877423 on Build GRCh38
rs929412037 Chr.6: 49877236-49877236 on Build GRCh38
rs149357009 Chr.6: 49877462-49877462 on Build GRCh38
rs13214128 Chr.6: 49877459-49877459 on Build GRCh38
rs1000534412 Chr.6: 49877117-49877118 on Build GRCh38

View Full Product Details