Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 115214517-115215014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADM1
Entrez Gene ID
23705
Gene Name
cell adhesion molecule 1
Gene Aliases
BL2, IGSF4, IGSF4A, NECL2, Necl-2, RA175, ST17, SYNCAM, TSLC1, sTSLC-1, sgIGSF, synCAM1
UniGene
Hs.370510
Gene Chromosome Location
Chr.11: 115173625-115504523 on Build GRCh38
Transcripts
NM_001098517.1 NM_001301043.1 NM_001301044.1 NM_001301045.1 NM_014333.3 XM_005271494.2 XM_017017457.1 XM_017017458.1 XM_017017459.1 XM_017017460.1 XM_017017461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3687096 c.921T>C p.D307D Chr.11: 115214681-115214681 on Build GRCh38
6067652 c.884G>A p.G295E Chr.11: 115214718-115214718 on Build GRCh38
4933347 c.879G>C p.L293L Chr.11: 115214723-115214723 on Build GRCh38
5932729 c.929C>A p.T310K Chr.11: 115214673-115214673 on Build GRCh38
3979140 c.874G>A p.V292I Chr.11: 115214728-115214728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764943948 Chr.11: 115214657-115214657 on Build GRCh38
rs779875593 Chr.11: 115214731-115214731 on Build GRCh38
rs947501184 Chr.11: 115214596-115214596 on Build GRCh38
rs761396235 Chr.11: 115214751-115214751 on Build GRCh38
rs752080208 Chr.11: 115214693-115214693 on Build GRCh38
rs777874833 Chr.11: 115214824-115214824 on Build GRCh38
rs1029133312 Chr.11: 115214937-115214937 on Build GRCh38
rs765359612 Chr.11: 115214700-115214700 on Build GRCh38
rs532788905 Chr.11: 115214639-115214639 on Build GRCh38
rs924736300 Chr.11: 115214988-115214988 on Build GRCh38
rs774927508 Chr.11: 115214559-115214559 on Build GRCh38
rs940777929 Chr.11: 115214885-115214885 on Build GRCh38
rs775406101 Chr.11: 115214635-115214635 on Build GRCh38
rs990198534 Chr.11: 115214637-115214637 on Build GRCh38
rs968747156 Chr.11: 115214947-115214947 on Build GRCh38
rs374153035 Chr.11: 115214704-115214704 on Build GRCh38
rs759431628 Chr.11: 115214551-115214551 on Build GRCh38
rs770088560 Chr.11: 115214623-115214623 on Build GRCh38
rs761329197 Chr.11: 115214650-115214650 on Build GRCh38
rs780716351 Chr.11: 115214591-115214591 on Build GRCh38
rs755007570 Chr.11: 115214604-115214604 on Build GRCh38
rs772994747 Chr.11: 115214765-115214765 on Build GRCh38
rs201871269 Chr.11: 115214702-115214702 on Build GRCh38
rs376941564 Chr.11: 115214631-115214631 on Build GRCh38
rs759894935 Chr.11: 115214789-115214789 on Build GRCh38
rs12576086 Chr.11: 115214866-115214866 on Build GRCh38
rs889347912 Chr.11: 115214941-115214941 on Build GRCh38
rs745943906 Chr.11: 115214712-115214712 on Build GRCh38
rs769241946 Chr.11: 115214763-115214763 on Build GRCh38
rs371635132 Chr.11: 115214618-115214618 on Build GRCh38
rs753202765 Chr.11: 115214808-115214808 on Build GRCh38
rs867521288 Chr.11: 115214730-115214730 on Build GRCh38
rs376680394 Chr.11: 115214813-115214813 on Build GRCh38
rs139051341 Chr.11: 115214629-115214629 on Build GRCh38
rs530800497 Chr.11: 115214912-115214912 on Build GRCh38
rs763864209 Chr.11: 115214566-115214567 on Build GRCh38
rs369076597 Chr.11: 115214573-115214573 on Build GRCh38
rs569853736 Chr.11: 115214942-115214942 on Build GRCh38
rs758904994 Chr.11: 115214582-115214582 on Build GRCh38
rs1049520821 Chr.11: 115214838-115214838 on Build GRCh38
rs150594862 Chr.11: 115214609-115214609 on Build GRCh38
rs762733550 Chr.11: 115214766-115214766 on Build GRCh38
rs528617309 Chr.11: 115214729-115214729 on Build GRCh38
rs184014809 Chr.11: 115214918-115214918 on Build GRCh38
rs745677210 Chr.11: 115214630-115214630 on Build GRCh38
rs757084998 Chr.11: 115214785-115214785 on Build GRCh38
rs746902980 Chr.11: 115214732-115214732 on Build GRCh38
rs753801160 Chr.11: 115214784-115214784 on Build GRCh38
rs574332848 Chr.11: 115214580-115214580 on Build GRCh38
rs997609637 Chr.11: 115214936-115214936 on Build GRCh38
rs765468758 Chr.11: 115214576-115214576 on Build GRCh38
rs369048786 Chr.11: 115214660-115214660 on Build GRCh38
rs755555614 Chr.11: 115214694-115214694 on Build GRCh38
rs766826690 Chr.11: 115214786-115214786 on Build GRCh38
rs763727502 Chr.11: 115214569-115214569 on Build GRCh38
rs753403057 Chr.11: 115214570-115214570 on Build GRCh38
rs997641669 Chr.11: 115214603-115214603 on Build GRCh38
rs896093445 Chr.11: 115214645-115214645 on Build GRCh38
rs934875997 Chr.11: 115214612-115214612 on Build GRCh38
rs965405642 Chr.11: 115214545-115214545 on Build GRCh38
rs763397012 Chr.11: 115214680-115214680 on Build GRCh38
rs1018856549 Chr.11: 115214819-115214819 on Build GRCh38
rs768082832 Chr.11: 115214799-115214799 on Build GRCh38
rs760165224 Chr.11: 115214560-115214560 on Build GRCh38
rs754040804 Chr.11: 115214827-115214827 on Build GRCh38
rs139604912 Chr.11: 115214681-115214681 on Build GRCh38
rs777959081 Chr.11: 115214728-115214728 on Build GRCh38
rs978825597 Chr.11: 115214961-115214961 on Build GRCh38
rs34106103 Chr.11: 115214699-115214699 on Build GRCh38
rs763771913 Chr.11: 115214841-115214841 on Build GRCh38
rs45525440 Chr.11: 115214747-115214747 on Build GRCh38
rs747576693 Chr.11: 115214595-115214595 on Build GRCh38
rs36038019 Chr.11: 115214627-115214628 on Build GRCh38
rs776224513 Chr.11: 115214640-115214640 on Build GRCh38
rs768667228 Chr.11: 115214741-115214741 on Build GRCh38
rs768748078 Chr.11: 115214906-115214906 on Build GRCh38

View Full Product Details