Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 116758958-116759470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUD13
Entrez Gene ID
84811
Gene Name
BUD13 homolog
Gene Aliases
Cwc26, fSAP71
UniGene
Hs.437341
Gene Chromosome Location
Chr.11: 116748170-116772998 on Build GRCh38
Transcripts
NM_001159736.1 NM_032725.3 XM_011543035.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7214989 c.1255-10C>T p.? Chr.11: 116759189-116759189 on Build GRCh38
7357879 c.1360+1G>A p.? Chr.11: 116759073-116759073 on Build GRCh38
5698172 c.1263C>G p.H421Q Chr.11: 116759171-116759171 on Build GRCh38
3375600 c.1297A>G p.T433A Chr.11: 116759137-116759137 on Build GRCh38
6571268 c.1347C>T p.T449T Chr.11: 116759087-116759087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771057423 Chr.11: 116759429-116759429 on Build GRCh38
rs1013414974 Chr.11: 116759095-116759095 on Build GRCh38
rs143075492 Chr.11: 116759183-116759183 on Build GRCh38
rs768816574 Chr.11: 116759101-116759101 on Build GRCh38
rs201570871 Chr.11: 116759091-116759091 on Build GRCh38
rs767550000 Chr.11: 116759163-116759163 on Build GRCh38
rs77429555 Chr.11: 116759032-116759032 on Build GRCh38
rs147463297 Chr.11: 116759415-116759415 on Build GRCh38
rs369731762 Chr.11: 116759125-116759125 on Build GRCh38
rs747603908 Chr.11: 116759197-116759197 on Build GRCh38
rs759491199 Chr.11: 116759134-116759134 on Build GRCh38
rs531659146 Chr.11: 116759323-116759323 on Build GRCh38
rs71833854 Chr.11: 116759018-116759019 on Build GRCh38
rs746534617 Chr.11: 116759038-116759045 on Build GRCh38
rs772578556 Chr.11: 116759084-116759084 on Build GRCh38
rs780503700 Chr.11: 116759191-116759191 on Build GRCh38
rs1039322907 Chr.11: 116759013-116759013 on Build GRCh38
rs955075388 Chr.11: 116759247-116759250 on Build GRCh38
rs542102275 Chr.11: 116759244-116759244 on Build GRCh38
rs752270259 Chr.11: 116759046-116759046 on Build GRCh38
rs752104364 Chr.11: 116759019-116759020 on Build GRCh38
rs4938310 Chr.11: 116759233-116759233 on Build GRCh38
rs763682168 Chr.11: 116759166-116759166 on Build GRCh38
rs779278860 Chr.11: 116759064-116759064 on Build GRCh38
rs74918085 Chr.11: 116759030-116759030 on Build GRCh38
rs371360909 Chr.11: 116759229-116759229 on Build GRCh38
rs910792242 Chr.11: 116758987-116758987 on Build GRCh38
rs754999814 Chr.11: 116759033-116759033 on Build GRCh38
rs544017304 Chr.11: 116759379-116759379 on Build GRCh38
rs975058199 Chr.11: 116759391-116759391 on Build GRCh38
rs546246109 Chr.11: 116759073-116759073 on Build GRCh38
rs942394289 Chr.11: 116758992-116758992 on Build GRCh38
rs755556135 Chr.11: 116759165-116759165 on Build GRCh38
rs928604355 Chr.11: 116759377-116759377 on Build GRCh38
rs371945728 Chr.11: 116759381-116759381 on Build GRCh38
rs756938504 Chr.11: 116759175-116759175 on Build GRCh38
rs17519093 Chr.11: 116759189-116759189 on Build GRCh38
rs909267372 Chr.11: 116759419-116759419 on Build GRCh38
rs756593763 Chr.11: 116759025-116759026 on Build GRCh38
rs35268246 Chr.11: 116759019-116759019 on Build GRCh38
rs774295403 Chr.11: 116759130-116759130 on Build GRCh38
rs565754877 Chr.11: 116759297-116759297 on Build GRCh38
rs116588420 Chr.11: 116759050-116759050 on Build GRCh38
rs747134662 Chr.11: 116759387-116759387 on Build GRCh38
rs748033515 Chr.11: 116759339-116759339 on Build GRCh38
rs778392376 Chr.11: 116759027-116759028 on Build GRCh38
rs937335123 Chr.11: 116759398-116759398 on Build GRCh38
rs532844060 Chr.11: 116759416-116759416 on Build GRCh38
rs746304555 Chr.11: 116759078-116759078 on Build GRCh38
rs182745160 Chr.11: 116759315-116759315 on Build GRCh38
rs987582704 Chr.11: 116759263-116759263 on Build GRCh38
rs374117898 Chr.11: 116759188-116759188 on Build GRCh38
rs535721326 Chr.11: 116759019-116759019 on Build GRCh38
rs56024416 Chr.11: 116759031-116759032 on Build GRCh38
rs200301153 Chr.11: 116759099-116759099 on Build GRCh38
rs77705076 Chr.11: 116759031-116759031 on Build GRCh38
rs910823509 Chr.11: 116759272-116759272 on Build GRCh38
rs981325971 Chr.11: 116759360-116759360 on Build GRCh38
rs200940541 Chr.11: 116759047-116759047 on Build GRCh38
rs1018440349 Chr.11: 116759269-116759269 on Build GRCh38
rs776700062 Chr.11: 116759124-116759124 on Build GRCh38
rs757514920 Chr.11: 116759058-116759058 on Build GRCh38
rs753390831 Chr.11: 116759172-116759172 on Build GRCh38
rs190177392 Chr.11: 116759053-116759053 on Build GRCh38
rs767071640 Chr.11: 116759034-116759034 on Build GRCh38

View Full Product Details