Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117819933-117820472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD2
Entrez Gene ID
486
Gene Name
FXYD domain containing ion transport regulator 2
Gene Aliases
ATP1G1, HOMG2
UniGene
Hs.731865
Gene Chromosome Location
Chr.11: 117820075-117828092 on Build GRCh38
Transcripts
NM_001680.4 NM_021603.3 NM_001204268.1 NM_001243598.2

Gene Symbol
FXYD6-FXYD2
Entrez Gene ID
100533181
Gene Name
FXYD6-FXYD2 readthrough
Gene Aliases
-
UniGene
Hs.744850
Gene Chromosome Location
Chr.11: 117820075-117877031 on Build GRCh38
Transcripts
NM_001680.4 NM_021603.3 NM_001204268.1 NM_001243598.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4779349 c.3G>A p.M1I Chr.11: 117820213-117820213 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1056611576 Chr.11: 117820138-117820138 on Build GRCh38
rs985295506 Chr.11: 117820043-117820043 on Build GRCh38
rs986854821 Chr.11: 117820368-117820368 on Build GRCh38
rs569588241 Chr.11: 117820271-117820271 on Build GRCh38
rs2014536 Chr.11: 117820430-117820430 on Build GRCh38
rs541214280 Chr.11: 117820217-117820217 on Build GRCh38
rs960839981 Chr.11: 117820101-117820101 on Build GRCh38
rs922528097 Chr.11: 117819996-117819996 on Build GRCh38
rs11999 Chr.11: 117820203-117820203 on Build GRCh38
rs78017409 Chr.11: 117820274-117820274 on Build GRCh38
rs1008788691 Chr.11: 117819958-117819958 on Build GRCh38
rs543295885 Chr.11: 117819989-117819989 on Build GRCh38
rs753535073 Chr.11: 117820174-117820174 on Build GRCh38
rs941383526 Chr.11: 117820154-117820154 on Build GRCh38
rs887929436 Chr.11: 117820375-117820375 on Build GRCh38
rs886047704 Chr.11: 117820191-117820191 on Build GRCh38
rs2014537 Chr.11: 117820433-117820433 on Build GRCh38
rs756608243 Chr.11: 117820255-117820255 on Build GRCh38
rs1022502853 Chr.11: 117819985-117819985 on Build GRCh38
rs766787853 Chr.11: 117820327-117820327 on Build GRCh38
rs902147230 Chr.11: 117820236-117820236 on Build GRCh38
rs771522350 Chr.11: 117820128-117820128 on Build GRCh38
rs961572137 Chr.11: 117819973-117819973 on Build GRCh38
rs41313497 Chr.11: 117820230-117820230 on Build GRCh38
rs187180617 Chr.11: 117820325-117820325 on Build GRCh38
rs966467089 Chr.11: 117819990-117819990 on Build GRCh38
rs933780512 Chr.11: 117820008-117820008 on Build GRCh38
rs748340168 Chr.11: 117820381-117820381 on Build GRCh38
rs545584979 Chr.11: 117820165-117820165 on Build GRCh38
rs929928700 Chr.11: 117820366-117820366 on Build GRCh38
rs574815593 Chr.11: 117820155-117820155 on Build GRCh38
rs778421158 Chr.11: 117820369-117820369 on Build GRCh38
rs760117426 Chr.11: 117820005-117820005 on Build GRCh38
rs918614211 Chr.11: 117820354-117820354 on Build GRCh38
rs946438670 Chr.11: 117820270-117820270 on Build GRCh38
rs890290871 Chr.11: 117820198-117820198 on Build GRCh38
rs978693561 Chr.11: 117820044-117820044 on Build GRCh38
rs902199125 Chr.11: 117820110-117820110 on Build GRCh38
rs896749454 Chr.11: 117820151-117820151 on Build GRCh38
rs765100315 Chr.11: 117820210-117820210 on Build GRCh38
rs749646500 Chr.11: 117820437-117820437 on Build GRCh38
rs565539331 Chr.11: 117820308-117820308 on Build GRCh38
rs556928775 Chr.11: 117820049-117820049 on Build GRCh38
rs999299954 Chr.11: 117820253-117820253 on Build GRCh38
rs1014977093 Chr.11: 117820424-117820424 on Build GRCh38
rs1051593449 Chr.11: 117820168-117820168 on Build GRCh38
rs901693142 Chr.11: 117820142-117820142 on Build GRCh38
rs946519509 Chr.11: 117820070-117820070 on Build GRCh38
rs527739331 Chr.11: 117820209-117820209 on Build GRCh38
rs530077413 Chr.11: 117820224-117820224 on Build GRCh38
rs948336709 Chr.11: 117820318-117820318 on Build GRCh38
rs41304349 Chr.11: 117820164-117820164 on Build GRCh38
rs145785816 Chr.11: 117820221-117820221 on Build GRCh38
rs1015442072 Chr.11: 117820194-117820194 on Build GRCh38
rs1006815006 Chr.11: 117820418-117820418 on Build GRCh38
rs1006331061 Chr.11: 117820188-117820188 on Build GRCh38
rs916798039 Chr.11: 117820131-117820131 on Build GRCh38
rs944580820 Chr.11: 117820406-117820406 on Build GRCh38
rs1035198231 Chr.11: 117820064-117820064 on Build GRCh38
rs1029548411 Chr.11: 117820222-117820222 on Build GRCh38
rs1043769879 Chr.11: 117820082-117820082 on Build GRCh38
rs550029549 Chr.11: 117820258-117820258 on Build GRCh38
rs1038825079 Chr.11: 117820417-117820417 on Build GRCh38
rs114952534 Chr.11: 117820301-117820301 on Build GRCh38
rs755001643 Chr.11: 117820309-117820309 on Build GRCh38
rs1038962545 Chr.11: 117820204-117820204 on Build GRCh38
rs911084647 Chr.11: 117820268-117820268 on Build GRCh38
rs758313070 Chr.11: 117820250-117820250 on Build GRCh38
rs900185002 Chr.11: 117819964-117819964 on Build GRCh38
rs574396483 Chr.11: 117819960-117819960 on Build GRCh38
rs890259631 Chr.11: 117819957-117819957 on Build GRCh38
rs1010563814 Chr.11: 117819970-117819970 on Build GRCh38
rs71469992 Chr.11: 117820430-117820433 on Build GRCh38

View Full Product Details